Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98003464-98003715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATN2
Entrez Gene ID
4147
Gene Name
matrilin 2
Gene Aliases
-
UniGene
Hs.189445
Gene Chromosome Location
Chr.8: 97869021-98036720 on Build GRCh38
Transcripts
NM_001317748.1 NM_002380.4 NM_030583.3 XM_005250920.1 XM_017013417.1 XM_017013418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3395461 c.1252G>A p.V418I Chr.8: 98003708-98003708 on Build GRCh38
4883073 c.1205G>T p.R402M Chr.8: 98003661-98003661 on Build GRCh38
1458806 c.1251C>T p.C417C Chr.8: 98003707-98003707 on Build GRCh38
6708856 c.1215C>T p.Y405Y Chr.8: 98003671-98003671 on Build GRCh38
1214628 c.1235G>A p.G412D Chr.8: 98003691-98003691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775599024 Chr.8: 98003658-98003658 on Build GRCh38
rs994928047 Chr.8: 98003536-98003536 on Build GRCh38
rs746527539 Chr.8: 98003598-98003598 on Build GRCh38
rs758054221 Chr.8: 98003593-98003593 on Build GRCh38
rs150407167 Chr.8: 98003509-98003509 on Build GRCh38
rs944734199 Chr.8: 98003518-98003518 on Build GRCh38
rs114175284 Chr.8: 98003535-98003535 on Build GRCh38
rs771411621 Chr.8: 98003622-98003622 on Build GRCh38
rs778111623 Chr.8: 98003612-98003612 on Build GRCh38
rs551396726 Chr.8: 98003616-98003616 on Build GRCh38
rs539630518 Chr.8: 98003530-98003530 on Build GRCh38
rs1049225588 Chr.8: 98003590-98003590 on Build GRCh38
rs749850898 Chr.8: 98003615-98003615 on Build GRCh38
rs1036254039 Chr.8: 98003640-98003640 on Build GRCh38
rs561469051 Chr.8: 98003578-98003578 on Build GRCh38
rs776416495 Chr.8: 98003679-98003679 on Build GRCh38
rs34926402 Chr.8: 98003653-98003653 on Build GRCh38
rs57984035 Chr.8: 98003502-98003502 on Build GRCh38
rs764279419 Chr.8: 98003671-98003671 on Build GRCh38
rs369792295 Chr.8: 98003611-98003611 on Build GRCh38
rs978487154 Chr.8: 98003657-98003657 on Build GRCh38
rs112996173 Chr.8: 98003642-98003642 on Build GRCh38
rs10104918 Chr.8: 98003626-98003626 on Build GRCh38
rs553161019 Chr.8: 98003531-98003531 on Build GRCh38
rs761802656 Chr.8: 98003681-98003681 on Build GRCh38
rs541134060 Chr.8: 98003568-98003568 on Build GRCh38
rs954075154 Chr.8: 98003500-98003500 on Build GRCh38
rs760636713 Chr.8: 98003661-98003661 on Build GRCh38
rs779484792 Chr.8: 98003594-98003594 on Build GRCh38
rs369211555 Chr.8: 98003689-98003689 on Build GRCh38
rs566413234 Chr.8: 98003688-98003688 on Build GRCh38
rs777133896 Chr.8: 98003627-98003627 on Build GRCh38
rs1035705473 Chr.8: 98003600-98003600 on Build GRCh38
rs12680911 Chr.8: 98003513-98003513 on Build GRCh38
rs753680800 Chr.8: 98003600-98003611 on Build GRCh38
rs184239062 Chr.8: 98003628-98003628 on Build GRCh38

View Full Product Details