Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177386382-177386645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC34A1
Entrez Gene ID
6569
Gene Name
solute carrier family 34 member 1
Gene Aliases
FRTS2, HCINF2, NAPI-3, NPHLOP1, NPT2, NPTIIa, SLC11, SLC17A2
UniGene
Hs.936
Gene Chromosome Location
Chr.5: 177384431-177412021 on Build GRCh38
Transcripts
NM_001167579.1 NM_003052.4 XM_005265975.1 XM_017009773.1 XM_017009775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5935058 c.389G>A p.G130E Chr.5: 177386423-177386423 on Build GRCh38
215976 c.493A>C p.T165P Chr.5: 177386527-177386527 on Build GRCh38
420763 c.519G>A p.M173I Chr.5: 177386553-177386553 on Build GRCh38
5758550 c.471C>T p.T157T Chr.5: 177386505-177386505 on Build GRCh38
482586 c.449T>C p.L150P Chr.5: 177386483-177386483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3812036 Chr.5: 177386403-177386403 on Build GRCh38
rs755960096 Chr.5: 177386592-177386592 on Build GRCh38
rs121918611 Chr.5: 177386473-177386473 on Build GRCh38
rs757124444 Chr.5: 177386559-177386560 on Build GRCh38
rs189794265 Chr.5: 177386418-177386418 on Build GRCh38
rs147095485 Chr.5: 177386449-177386449 on Build GRCh38
rs876657375 Chr.5: 177386514-177386515 on Build GRCh38
rs754966307 Chr.5: 177386555-177386555 on Build GRCh38
rs200152945 Chr.5: 177386542-177386542 on Build GRCh38
rs200188041 Chr.5: 177386454-177386454 on Build GRCh38
rs775773480 Chr.5: 177386604-177386604 on Build GRCh38
rs781032664 Chr.5: 177386480-177386480 on Build GRCh38
rs767666753 Chr.5: 177386446-177386446 on Build GRCh38
rs768548093 Chr.5: 177386479-177386479 on Build GRCh38
rs777543926 Chr.5: 177386480-177386481 on Build GRCh38
rs769409705 Chr.5: 177386492-177386492 on Build GRCh38
rs138719745 Chr.5: 177386455-177386455 on Build GRCh38
rs747997092 Chr.5: 177386590-177386590 on Build GRCh38
rs777410389 Chr.5: 177386593-177386593 on Build GRCh38
rs772329350 Chr.5: 177386503-177386503 on Build GRCh38
rs937785918 Chr.5: 177386477-177386477 on Build GRCh38
rs761333934 Chr.5: 177386507-177386507 on Build GRCh38
rs781202369 Chr.5: 177386566-177386566 on Build GRCh38
rs772422439 Chr.5: 177386602-177386602 on Build GRCh38
rs368438452 Chr.5: 177386404-177386404 on Build GRCh38
rs771178295 Chr.5: 177386431-177386431 on Build GRCh38
rs570463028 Chr.5: 177386545-177386545 on Build GRCh38
rs778637456 Chr.5: 177386456-177386456 on Build GRCh38
rs757931846 Chr.5: 177386472-177386472 on Build GRCh38
rs746905134 Chr.5: 177386415-177386415 on Build GRCh38
rs576880299 Chr.5: 177386423-177386423 on Build GRCh38
rs975958205 Chr.5: 177386430-177386430 on Build GRCh38
rs552176812 Chr.5: 177386544-177386544 on Build GRCh38
rs758233945 Chr.5: 177386441-177386443 on Build GRCh38
rs373111976 Chr.5: 177386606-177386606 on Build GRCh38
rs550248810 Chr.5: 177386506-177386506 on Build GRCh38
rs369770760 Chr.5: 177386498-177386498 on Build GRCh38
rs764689472 Chr.5: 177386508-177386508 on Build GRCh38
rs759667506 Chr.5: 177386441-177386441 on Build GRCh38
rs548844573 Chr.5: 177386471-177386471 on Build GRCh38
rs775683749 Chr.5: 177386505-177386505 on Build GRCh38
rs376730264 Chr.5: 177386478-177386478 on Build GRCh38
rs765474466 Chr.5: 177386525-177386525 on Build GRCh38
rs955811738 Chr.5: 177386434-177386434 on Build GRCh38
rs762239647 Chr.5: 177386515-177386515 on Build GRCh38
rs774666109 Chr.5: 177386496-177386496 on Build GRCh38
rs148976897 Chr.5: 177386432-177386432 on Build GRCh38
rs1047665666 Chr.5: 177386584-177386584 on Build GRCh38
rs747102698 Chr.5: 177386608-177386608 on Build GRCh38
rs202211232 Chr.5: 177386483-177386483 on Build GRCh38
rs746134936 Chr.5: 177386598-177386598 on Build GRCh38
rs775408301 Chr.5: 177386407-177386407 on Build GRCh38
rs754244361 Chr.5: 177386513-177386513 on Build GRCh38
rs772535455 Chr.5: 177386482-177386496 on Build GRCh38

View Full Product Details