Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35968640-35968911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNBD2
Entrez Gene ID
140894
Gene Name
cyclic nucleotide binding domain containing 2
Gene Aliases
C20orf152, CNMPD1
UniGene
Hs.732697
Gene Chromosome Location
Chr.20: 35954561-36030700 on Build GRCh38
Transcripts
NM_001207076.2 NM_001304367.1 NM_080834.3 NR_130714.1 XM_005260295.2 XM_011528590.2 XM_011528593.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6467651 c.30G>A p.W10* Chr.20: 35968792-35968792 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572679811 Chr.20: 35968666-35968666 on Build GRCh38
rs936553663 Chr.20: 35968719-35968719 on Build GRCh38
rs373742482 Chr.20: 35968832-35968832 on Build GRCh38
rs994565302 Chr.20: 35968885-35968885 on Build GRCh38
rs1000124315 Chr.20: 35968760-35968760 on Build GRCh38
rs771871437 Chr.20: 35968839-35968839 on Build GRCh38
rs775383429 Chr.20: 35968847-35968847 on Build GRCh38
rs769607568 Chr.20: 35968693-35968693 on Build GRCh38
rs543330312 Chr.20: 35968670-35968670 on Build GRCh38
rs780350079 Chr.20: 35968775-35968775 on Build GRCh38
rs531349073 Chr.20: 35968776-35968776 on Build GRCh38
rs917615859 Chr.20: 35968685-35968685 on Build GRCh38
rs561390894 Chr.20: 35968754-35968754 on Build GRCh38
rs202125164 Chr.20: 35968834-35968834 on Build GRCh38
rs761874857 Chr.20: 35968715-35968715 on Build GRCh38
rs564686855 Chr.20: 35968858-35968858 on Build GRCh38
rs11476128 Chr.20: 35968833-35968833 on Build GRCh38
rs138019065 Chr.20: 35968786-35968786 on Build GRCh38
rs758632416 Chr.20: 35968735-35968735 on Build GRCh38
rs371997100 Chr.20: 35968711-35968711 on Build GRCh38
rs142034197 Chr.20: 35968792-35968792 on Build GRCh38
rs778916914 Chr.20: 35968712-35968712 on Build GRCh38
rs989125391 Chr.20: 35968807-35968807 on Build GRCh38
rs375289266 Chr.20: 35968713-35968713 on Build GRCh38
rs370279613 Chr.20: 35968796-35968796 on Build GRCh38
rs889587036 Chr.20: 35968766-35968766 on Build GRCh38
rs376975938 Chr.20: 35968784-35968784 on Build GRCh38
rs199581013 Chr.20: 35968770-35968770 on Build GRCh38

View Full Product Details