Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32076979-32077238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM39B
Entrez Gene ID
55116
Gene Name
transmembrane protein 39B
Gene Aliases
-
UniGene
Hs.25544
Gene Chromosome Location
Chr.1: 32072031-32102866 on Build GRCh38
Transcripts
NM_001319677.1 NM_001319678.1 NM_001319679.1 NM_018056.3 XM_005270988.2 XM_006710724.1 XM_006710725.3 XM_011541681.2 XM_011541683.1 XM_011541684.2 XM_017001595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6575644 c.61C>G p.Q21E Chr.1: 32077170-32077170 on Build GRCh38
4495665 c.458C>T p.S153F Chr.1: 32077186-32077186 on Build GRCh38
5827850 c.146G>A p.R49H Chr.1: 32077219-32077219 on Build GRCh38
3966524 c.161C>T p.T54M Chr.1: 32077234-32077234 on Build GRCh38
4495731 c.114C>T p.S38S Chr.1: 32077187-32077187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538164987 Chr.1: 32077156-32077156 on Build GRCh38
rs551458339 Chr.1: 32077055-32077055 on Build GRCh38
rs376026022 Chr.1: 32077008-32077008 on Build GRCh38
rs1057447172 Chr.1: 32077216-32077216 on Build GRCh38
rs779663959 Chr.1: 32077179-32077179 on Build GRCh38
rs187090348 Chr.1: 32077119-32077119 on Build GRCh38
rs764359527 Chr.1: 32077159-32077159 on Build GRCh38
rs748074521 Chr.1: 32077135-32077135 on Build GRCh38
rs903598247 Chr.1: 32077038-32077038 on Build GRCh38
rs910242675 Chr.1: 32077030-32077030 on Build GRCh38
rs772246191 Chr.1: 32077151-32077151 on Build GRCh38
rs371321787 Chr.1: 32077157-32077157 on Build GRCh38
rs769656874 Chr.1: 32077211-32077211 on Build GRCh38
rs985757895 Chr.1: 32077014-32077014 on Build GRCh38
rs1012050500 Chr.1: 32077193-32077193 on Build GRCh38
rs566544349 Chr.1: 32077108-32077108 on Build GRCh38
rs896421556 Chr.1: 32077014-32077015 on Build GRCh38
rs758005589 Chr.1: 32077178-32077178 on Build GRCh38
rs139522223 Chr.1: 32077172-32077172 on Build GRCh38
rs902989907 Chr.1: 32077196-32077196 on Build GRCh38
rs758056991 Chr.1: 32077117-32077117 on Build GRCh38
rs866526461 Chr.1: 32077089-32077089 on Build GRCh38
rs751143276 Chr.1: 32077188-32077188 on Build GRCh38
rs199702572 Chr.1: 32077121-32077121 on Build GRCh38
rs774359749 Chr.1: 32077141-32077141 on Build GRCh38
rs147916945 Chr.1: 32077175-32077175 on Build GRCh38
rs776531170 Chr.1: 32077123-32077123 on Build GRCh38
rs776150850 Chr.1: 32077173-32077173 on Build GRCh38
rs772011464 Chr.1: 32077219-32077219 on Build GRCh38
rs562694333 Chr.1: 32077111-32077111 on Build GRCh38
rs567595520 Chr.1: 32077142-32077142 on Build GRCh38
rs199599632 Chr.1: 32077218-32077218 on Build GRCh38
rs765843808 Chr.1: 32077174-32077174 on Build GRCh38
rs769785024 Chr.1: 32077140-32077140 on Build GRCh38
rs754727698 Chr.1: 32077194-32077194 on Build GRCh38
rs190656315 Chr.1: 32077195-32077195 on Build GRCh38
rs532846201 Chr.1: 32077054-32077054 on Build GRCh38
rs779602824 Chr.1: 32077118-32077118 on Build GRCh38
rs778551924 Chr.1: 32077114-32077114 on Build GRCh38
rs777723565 Chr.1: 32077214-32077214 on Build GRCh38
rs374449944 Chr.1: 32077182-32077182 on Build GRCh38
rs368020841 Chr.1: 32077220-32077220 on Build GRCh38
rs1000625410 Chr.1: 32077048-32077048 on Build GRCh38
rs747953802 Chr.1: 32077199-32077199 on Build GRCh38
rs745382940 Chr.1: 32077115-32077115 on Build GRCh38

View Full Product Details