Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27747952-27748225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622019 c.457C>T p.R153C Chr.8: 27748171-27748171 on Build GRCh38
5409337 c.468G>A p.R156R Chr.8: 27748160-27748160 on Build GRCh38
3768763 c.457C>G p.R153G Chr.8: 27748171-27748171 on Build GRCh38
5417202 c.450C>T p.C150C Chr.8: 27748178-27748178 on Build GRCh38
6622021 c.502A>G p.K168E Chr.8: 27748126-27748126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369943638 Chr.8: 27748147-27748147 on Build GRCh38
rs980999130 Chr.8: 27748198-27748198 on Build GRCh38
rs369502557 Chr.8: 27748070-27748070 on Build GRCh38
rs779289360 Chr.8: 27748178-27748178 on Build GRCh38
rs745998391 Chr.8: 27748046-27748046 on Build GRCh38
rs976565648 Chr.8: 27748017-27748017 on Build GRCh38
rs150006124 Chr.8: 27748205-27748205 on Build GRCh38
rs751077565 Chr.8: 27748121-27748121 on Build GRCh38
rs775584926 Chr.8: 27748061-27748061 on Build GRCh38
rs201322823 Chr.8: 27748096-27748096 on Build GRCh38
rs755803603 Chr.8: 27748156-27748156 on Build GRCh38
rs139289703 Chr.8: 27748078-27748078 on Build GRCh38
rs762964810 Chr.8: 27748093-27748093 on Build GRCh38
rs1047885663 Chr.8: 27748107-27748107 on Build GRCh38
rs148042574 Chr.8: 27748019-27748019 on Build GRCh38
rs773185109 Chr.8: 27748086-27748086 on Build GRCh38
rs1018149568 Chr.8: 27747998-27747998 on Build GRCh38
rs907473555 Chr.8: 27748033-27748033 on Build GRCh38
rs770569574 Chr.8: 27748146-27748146 on Build GRCh38
rs780171235 Chr.8: 27748114-27748114 on Build GRCh38
rs202197146 Chr.8: 27748170-27748170 on Build GRCh38
rs758974711 Chr.8: 27748130-27748130 on Build GRCh38
rs772267689 Chr.8: 27748055-27748055 on Build GRCh38
rs544933439 Chr.8: 27747992-27747992 on Build GRCh38
rs934783087 Chr.8: 27747987-27747987 on Build GRCh38
rs948947055 Chr.8: 27748024-27748024 on Build GRCh38
rs1044648829 Chr.8: 27748026-27748026 on Build GRCh38
rs778949131 Chr.8: 27748034-27748034 on Build GRCh38
rs1016075614 Chr.8: 27748191-27748191 on Build GRCh38
rs761616875 Chr.8: 27748069-27748069 on Build GRCh38
rs1005391107 Chr.8: 27747997-27747997 on Build GRCh38
rs776460710 Chr.8: 27748096-27748098 on Build GRCh38
rs766484443 Chr.8: 27748094-27748094 on Build GRCh38
rs539637083 Chr.8: 27748150-27748150 on Build GRCh38
rs757755754 Chr.8: 27748171-27748171 on Build GRCh38
rs144489715 Chr.8: 27748015-27748015 on Build GRCh38
rs759148084 Chr.8: 27748113-27748122 on Build GRCh38
rs200774099 Chr.8: 27748193-27748193 on Build GRCh38
rs758425766 Chr.8: 27748204-27748204 on Build GRCh38

View Full Product Details