Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7594538-7594809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXR2
Entrez Gene ID
9513
Gene Name
FMR1 autosomal homolog 2
Gene Aliases
FMR1L2, FXR2P
UniGene
Hs.52788
Gene Chromosome Location
Chr.17: 7591230-7614897 on Build GRCh38
Transcripts
NM_004860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853587 c.860G>T p.S287I Chr.17: 7594729-7594729 on Build GRCh38
707086 c.832-1G>C p.? Chr.17: 7594758-7594758 on Build GRCh38
6667967 c.856C>T p.R286* Chr.17: 7594733-7594733 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs181260928 Chr.17: 7594776-7594776 on Build GRCh38
rs368387062 Chr.17: 7594756-7594756 on Build GRCh38
rs531486234 Chr.17: 7594606-7594606 on Build GRCh38
rs761815605 Chr.17: 7594763-7594763 on Build GRCh38
rs756492540 Chr.17: 7594663-7594663 on Build GRCh38
rs781200094 Chr.17: 7594659-7594659 on Build GRCh38
rs1011830446 Chr.17: 7594751-7594751 on Build GRCh38
rs369134614 Chr.17: 7594777-7594777 on Build GRCh38
rs367700576 Chr.17: 7594645-7594645 on Build GRCh38
rs753115600 Chr.17: 7594662-7594662 on Build GRCh38
rs117166689 Chr.17: 7594570-7594570 on Build GRCh38
rs3744267 Chr.17: 7594683-7594683 on Build GRCh38
rs375991949 Chr.17: 7594752-7594752 on Build GRCh38
rs973754420 Chr.17: 7594593-7594593 on Build GRCh38
rs374715082 Chr.17: 7594627-7594627 on Build GRCh38
rs766610782 Chr.17: 7594633-7594633 on Build GRCh38
rs188720630 Chr.17: 7594617-7594617 on Build GRCh38
rs749833980 Chr.17: 7594686-7594686 on Build GRCh38
rs1041828920 Chr.17: 7594781-7594781 on Build GRCh38
rs768253300 Chr.17: 7594650-7594651 on Build GRCh38
rs755278128 Chr.17: 7594657-7594657 on Build GRCh38
rs778417682 Chr.17: 7594742-7594742 on Build GRCh38
rs771404358 Chr.17: 7594704-7594704 on Build GRCh38
rs201124410 Chr.17: 7594749-7594749 on Build GRCh38

View Full Product Details