Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62197807-62198080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTG2
Entrez Gene ID
26164
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5, ObgH1, dJ1005F21.2
UniGene
Hs.340636
Gene Chromosome Location
Chr.20: 62183025-62203568 on Build GRCh38
Transcripts
NM_015666.3 XM_005260393.1 XM_006723774.3 XM_011528776.1 XM_017027805.1 XM_017027806.1 XM_017027807.1 XM_017027808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7251585 c.353-8T>C p.? Chr.20: 62197844-62197844 on Build GRCh38
4756228 c.379T>A p.S127T Chr.20: 62197878-62197878 on Build GRCh38
379268 c.450C>T p.G150G Chr.20: 62197949-62197949 on Build GRCh38
1483816 c.377C>G p.S126W Chr.20: 62197876-62197876 on Build GRCh38
4689249 c.443G>A p.R148H Chr.20: 62197942-62197942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755632480 Chr.20: 62197992-62197992 on Build GRCh38
rs41302557 Chr.20: 62197830-62197830 on Build GRCh38
rs776505648 Chr.20: 62197962-62197962 on Build GRCh38
rs145129303 Chr.20: 62197953-62197953 on Build GRCh38
rs78123987 Chr.20: 62198036-62198036 on Build GRCh38
rs937910468 Chr.20: 62197915-62197915 on Build GRCh38
rs745660511 Chr.20: 62198008-62198008 on Build GRCh38
rs780062372 Chr.20: 62198015-62198015 on Build GRCh38
rs930348508 Chr.20: 62198047-62198047 on Build GRCh38
rs752057393 Chr.20: 62197923-62197923 on Build GRCh38
rs545500022 Chr.20: 62198042-62198042 on Build GRCh38
rs752103157 Chr.20: 62197990-62197990 on Build GRCh38
rs766770799 Chr.20: 62197914-62197914 on Build GRCh38
rs1055457324 Chr.20: 62198054-62198054 on Build GRCh38
rs117343642 Chr.20: 62197867-62197867 on Build GRCh38
rs564083574 Chr.20: 62198046-62198046 on Build GRCh38
rs972102668 Chr.20: 62198035-62198035 on Build GRCh38
rs966057749 Chr.20: 62198009-62198009 on Build GRCh38
rs553293444 Chr.20: 62197974-62197974 on Build GRCh38
rs779130835 Chr.20: 62197837-62197837 on Build GRCh38
rs750811305 Chr.20: 62197845-62197845 on Build GRCh38
rs745784951 Chr.20: 62197952-62197952 on Build GRCh38
rs769503072 Chr.20: 62197872-62197872 on Build GRCh38
rs148174053 Chr.20: 62197896-62197896 on Build GRCh38
rs201214722 Chr.20: 62197891-62197891 on Build GRCh38
rs777461480 Chr.20: 62197994-62197994 on Build GRCh38
rs780823733 Chr.20: 62197865-62197865 on Build GRCh38
rs376327555 Chr.20: 62197877-62197877 on Build GRCh38
rs758785320 Chr.20: 62197907-62197907 on Build GRCh38
rs142949953 Chr.20: 62197905-62197905 on Build GRCh38
rs754453784 Chr.20: 62197860-62197860 on Build GRCh38
rs961997142 Chr.20: 62198030-62198030 on Build GRCh38
rs373074268 Chr.20: 62197835-62197835 on Build GRCh38
rs780568989 Chr.20: 62197853-62197853 on Build GRCh38
rs755686028 Chr.20: 62197938-62197938 on Build GRCh38
rs903266451 Chr.20: 62197963-62197963 on Build GRCh38
rs561155322 Chr.20: 62197829-62197830 on Build GRCh38
rs145471301 Chr.20: 62197880-62197880 on Build GRCh38
rs764073039 Chr.20: 62197888-62197888 on Build GRCh38
rs911767472 Chr.20: 62198029-62198029 on Build GRCh38
rs780512559 Chr.20: 62197931-62197931 on Build GRCh38
rs757025132 Chr.20: 62198001-62198001 on Build GRCh38
rs950173255 Chr.20: 62197949-62197949 on Build GRCh38
rs143808782 Chr.20: 62197889-62197889 on Build GRCh38
rs778789327 Chr.20: 62198002-62198002 on Build GRCh38
rs758834549 Chr.20: 62197851-62197851 on Build GRCh38
rs370429275 Chr.20: 62197879-62197879 on Build GRCh38
rs200604036 Chr.20: 62197942-62197942 on Build GRCh38
rs773401413 Chr.20: 62197968-62197968 on Build GRCh38
rs202030848 Chr.20: 62197944-62197944 on Build GRCh38
rs751936187 Chr.20: 62197984-62197984 on Build GRCh38
rs777507826 Chr.20: 62197940-62197940 on Build GRCh38
rs761982766 Chr.20: 62197965-62197965 on Build GRCh38
rs201469338 Chr.20: 62197975-62197975 on Build GRCh38
rs757107174 Chr.20: 62197890-62197890 on Build GRCh38
rs987318050 Chr.20: 62198023-62198023 on Build GRCh38
rs563368145 Chr.20: 62197995-62197995 on Build GRCh38
rs748884084 Chr.20: 62197885-62197886 on Build GRCh38
rs112103829 Chr.20: 62197964-62197964 on Build GRCh38
rs781143736 Chr.20: 62197924-62197924 on Build GRCh38
rs6062140 Chr.20: 62197985-62197985 on Build GRCh38
rs138844562 Chr.20: 62197966-62197966 on Build GRCh38
rs764500330 Chr.20: 62197874-62197876 on Build GRCh38
rs747701172 Chr.20: 62197937-62197937 on Build GRCh38
rs774350773 Chr.20: 62197950-62197950 on Build GRCh38
rs746473715 Chr.20: 62197858-62197858 on Build GRCh38
rs915870996 Chr.20: 62197941-62197941 on Build GRCh38
rs772819909 Chr.20: 62197876-62197876 on Build GRCh38

View Full Product Details