Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20731772-20732029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1AKMT1
Entrez Gene ID
221143
Gene Name
eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1
Gene Aliases
ESP13, N6AMT2
UniGene
Hs.26674
Gene Chromosome Location
Chr.13: 20728931-20773958 on Build GRCh38
Transcripts
NM_001318939.1 NM_174928.2 NR_134934.1 XM_017020432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174000 c.467C>A p.T156N Chr.13: 20731882-20731882 on Build GRCh38
3467628 c.456A>C p.K152N Chr.13: 20731893-20731893 on Build GRCh38
3671249 c.438G>T p.S146S Chr.13: 20731911-20731911 on Build GRCh38
3417419 c.349T>G p.F117V Chr.13: 20732000-20732000 on Build GRCh38
3360043 c.406G>A p.D136N Chr.13: 20731943-20731943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761771447 Chr.13: 20731906-20731906 on Build GRCh38
rs772144111 Chr.13: 20731873-20731873 on Build GRCh38
rs749657822 Chr.13: 20731865-20731865 on Build GRCh38
rs753437846 Chr.13: 20731996-20731996 on Build GRCh38
rs765795109 Chr.13: 20731926-20731926 on Build GRCh38
rs749953569 Chr.13: 20731910-20731910 on Build GRCh38
rs755350464 Chr.13: 20731918-20731918 on Build GRCh38
rs774440467 Chr.13: 20731888-20731888 on Build GRCh38
rs765110309 Chr.13: 20731806-20731806 on Build GRCh38
rs779182481 Chr.13: 20731922-20731922 on Build GRCh38
rs780786236 Chr.13: 20731837-20731837 on Build GRCh38
rs375923475 Chr.13: 20731866-20731866 on Build GRCh38
rs750125960 Chr.13: 20731844-20731844 on Build GRCh38
rs576476935 Chr.13: 20731952-20731952 on Build GRCh38
rs148306539 Chr.13: 20731932-20731932 on Build GRCh38
rs146153743 Chr.13: 20731931-20731931 on Build GRCh38
rs760162605 Chr.13: 20731989-20731989 on Build GRCh38
rs542049732 Chr.13: 20731824-20731824 on Build GRCh38
rs141407092 Chr.13: 20731963-20731963 on Build GRCh38
rs202042737 Chr.13: 20731911-20731911 on Build GRCh38
rs369283175 Chr.13: 20731867-20731867 on Build GRCh38
rs771036966 Chr.13: 20731938-20731938 on Build GRCh38
rs888373340 Chr.13: 20731968-20731968 on Build GRCh38
rs61955729 Chr.13: 20731802-20731802 on Build GRCh38
rs773235638 Chr.13: 20731875-20731875 on Build GRCh38
rs201771529 Chr.13: 20731863-20731863 on Build GRCh38
rs768394262 Chr.13: 20731940-20731940 on Build GRCh38
rs142311304 Chr.13: 20731887-20731887 on Build GRCh38
rs200509905 Chr.13: 20731880-20731880 on Build GRCh38
rs7992250 Chr.13: 20731808-20731808 on Build GRCh38
rs766014793 Chr.13: 20731912-20731912 on Build GRCh38
rs762785430 Chr.13: 20731815-20731815 on Build GRCh38
rs764164398 Chr.13: 20731822-20731822 on Build GRCh38
rs34399030 Chr.13: 20731974-20731974 on Build GRCh38
rs756881913 Chr.13: 20731829-20731829 on Build GRCh38
rs779737549 Chr.13: 20731864-20731864 on Build GRCh38
rs1014073853 Chr.13: 20731831-20731831 on Build GRCh38
rs35948132 Chr.13: 20732001-20732001 on Build GRCh38
rs772127457 Chr.13: 20731797-20731797 on Build GRCh38
rs371451907 Chr.13: 20731838-20731838 on Build GRCh38
rs1034221566 Chr.13: 20731994-20731994 on Build GRCh38
rs747096067 Chr.13: 20731937-20731937 on Build GRCh38
rs765858954 Chr.13: 20731992-20731992 on Build GRCh38
rs768572062 Chr.13: 20731960-20731960 on Build GRCh38
rs905293155 Chr.13: 20731853-20731853 on Build GRCh38
rs978370525 Chr.13: 20731807-20731807 on Build GRCh38
rs746041478 Chr.13: 20731951-20731951 on Build GRCh38
rs776773528 Chr.13: 20731942-20731942 on Build GRCh38
rs753518016 Chr.13: 20731915-20731915 on Build GRCh38
rs773200202 Chr.13: 20731983-20731983 on Build GRCh38
rs767162001 Chr.13: 20731908-20731908 on Build GRCh38
rs773436390 Chr.13: 20731801-20731801 on Build GRCh38
rs202246353 Chr.13: 20731878-20731878 on Build GRCh38

View Full Product Details