Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149883354-149883619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6A
Entrez Gene ID
5145
Gene Name
phosphodiesterase 6A
Gene Aliases
CGPR-A, PDEA, RP43
UniGene
Hs.567314
Gene Chromosome Location
Chr.5: 149857956-149944793 on Build GRCh38
Transcripts
NM_000440.2 XM_011537650.2 XM_011537651.2 XM_011537652.1 XM_011537653.2 XM_011537654.2 XM_017009572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3852686 c.2119C>T p.R707W Chr.5: 149883445-149883445 on Build GRCh38
3919329 c.2056G>A p.D686N Chr.5: 149883508-149883508 on Build GRCh38
5597358 c.2097G>A p.Q699Q Chr.5: 149883467-149883467 on Build GRCh38
3919328 c.2125G>A p.E709K Chr.5: 149883439-149883439 on Build GRCh38
3612400 c.2130C>T p.I710I Chr.5: 149883434-149883434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148637474 Chr.5: 149883439-149883439 on Build GRCh38
rs868287120 Chr.5: 149883421-149883421 on Build GRCh38
rs758033844 Chr.5: 149883545-149883545 on Build GRCh38
rs769715536 Chr.5: 149883487-149883487 on Build GRCh38
rs374914136 Chr.5: 149883397-149883397 on Build GRCh38
rs561600163 Chr.5: 149883428-149883428 on Build GRCh38
rs773993473 Chr.5: 149883548-149883548 on Build GRCh38
rs766675633 Chr.5: 149883546-149883547 on Build GRCh38
rs759428024 Chr.5: 149883386-149883386 on Build GRCh38
rs763763317 Chr.5: 149883413-149883413 on Build GRCh38
rs895158467 Chr.5: 149883429-149883429 on Build GRCh38
rs945196543 Chr.5: 149883375-149883375 on Build GRCh38
rs748905273 Chr.5: 149883421-149883422 on Build GRCh38
rs774064748 Chr.5: 149883384-149883384 on Build GRCh38
rs767678078 Chr.5: 149883463-149883463 on Build GRCh38
rs121909835 Chr.5: 149883511-149883511 on Build GRCh38
rs148598583 Chr.5: 149883526-149883526 on Build GRCh38
rs147017310 Chr.5: 149883528-149883528 on Build GRCh38
rs753354342 Chr.5: 149883418-149883418 on Build GRCh38
rs775420166 Chr.5: 149883518-149883518 on Build GRCh38
rs112753117 Chr.5: 149883588-149883588 on Build GRCh38
rs773517408 Chr.5: 149883491-149883491 on Build GRCh38
rs748033023 Chr.5: 149883568-149883569 on Build GRCh38
rs1055520935 Chr.5: 149883426-149883427 on Build GRCh38
rs750090523 Chr.5: 149883536-149883536 on Build GRCh38
rs952946602 Chr.5: 149883500-149883500 on Build GRCh38
rs771897277 Chr.5: 149883585-149883586 on Build GRCh38
rs781169571 Chr.5: 149883469-149883469 on Build GRCh38
rs372372524 Chr.5: 149883396-149883396 on Build GRCh38
rs781735143 Chr.5: 149883584-149883584 on Build GRCh38
rs142092713 Chr.5: 149883445-149883445 on Build GRCh38
rs764962408 Chr.5: 149883433-149883433 on Build GRCh38
rs768391293 Chr.5: 149883431-149883431 on Build GRCh38
rs780341969 Chr.5: 149883444-149883444 on Build GRCh38
rs372808484 Chr.5: 149883414-149883414 on Build GRCh38
rs755475399 Chr.5: 149883458-149883458 on Build GRCh38
rs909565362 Chr.5: 149883468-149883468 on Build GRCh38
rs147636561 Chr.5: 149883531-149883531 on Build GRCh38
rs766797276 Chr.5: 149883546-149883546 on Build GRCh38
rs750000569 Chr.5: 149883434-149883434 on Build GRCh38
rs140267699 Chr.5: 149883527-149883527 on Build GRCh38
rs772049478 Chr.5: 149883512-149883512 on Build GRCh38
rs1037192141 Chr.5: 149883478-149883478 on Build GRCh38
rs779730800 Chr.5: 149883395-149883396 on Build GRCh38
rs533660014 Chr.5: 149883569-149883569 on Build GRCh38
rs369263399 Chr.5: 149883404-149883404 on Build GRCh38
rs376182220 Chr.5: 149883592-149883592 on Build GRCh38
rs970968217 Chr.5: 149883391-149883391 on Build GRCh38
rs751895061 Chr.5: 149883547-149883547 on Build GRCh38

View Full Product Details