Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 147938199-147938458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORC4
Entrez Gene ID
5000
Gene Name
origin recognition complex subunit 4
Gene Aliases
ORC4L, ORC4P
UniGene
Hs.558364
Gene Chromosome Location
Chr.2: 147930397-148021604 on Build GRCh38
Transcripts
NM_001190879.2 NM_001190881.2 NM_001190882.2 NM_002552.4 NM_181741.3 NM_181742.3 XM_011511255.2 XM_017004226.1 XM_017004227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1176598 c.1067T>C p.F356S Chr.2: 147938201-147938201 on Build GRCh38
1007195 c.982C>A p.L328I Chr.2: 147938370-147938370 on Build GRCh38
1007196 c.961C>A p.L321I Chr.2: 147938391-147938391 on Build GRCh38
1752044 c.1009G>A p.D337N Chr.2: 147938343-147938343 on Build GRCh38
3364297 c.1024G>C p.E342Q Chr.2: 147938328-147938328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374973087 Chr.2: 147938284-147938284 on Build GRCh38
rs755781534 Chr.2: 147938255-147938255 on Build GRCh38
rs762175361 Chr.2: 147938425-147938432 on Build GRCh38
rs775109530 Chr.2: 147938407-147938408 on Build GRCh38
rs777224956 Chr.2: 147938417-147938418 on Build GRCh38
rs767053958 Chr.2: 147938253-147938253 on Build GRCh38
rs770783840 Chr.2: 147938286-147938286 on Build GRCh38
rs369212077 Chr.2: 147938290-147938290 on Build GRCh38
rs745632291 Chr.2: 147938363-147938364 on Build GRCh38
rs769033885 Chr.2: 147938305-147938305 on Build GRCh38
rs1012559239 Chr.2: 147938261-147938261 on Build GRCh38
rs767982981 Chr.2: 147938414-147938416 on Build GRCh38
rs923300628 Chr.2: 147938399-147938399 on Build GRCh38
rs752337960 Chr.2: 147938254-147938254 on Build GRCh38
rs777362143 Chr.2: 147938263-147938263 on Build GRCh38
rs750941282 Chr.2: 147938397-147938397 on Build GRCh38
rs746381356 Chr.2: 147938293-147938293 on Build GRCh38
rs770839650 Chr.2: 147938346-147938346 on Build GRCh38
rs78758762 Chr.2: 147938389-147938389 on Build GRCh38
rs760245539 Chr.2: 147938408-147938408 on Build GRCh38
rs762798777 Chr.2: 147938228-147938228 on Build GRCh38
rs779453661 Chr.2: 147938281-147938281 on Build GRCh38
rs762857368 Chr.2: 147938335-147938335 on Build GRCh38
rs201217453 Chr.2: 147938229-147938229 on Build GRCh38
rs754204192 Chr.2: 147938272-147938272 on Build GRCh38
rs772083404 Chr.2: 147938296-147938296 on Build GRCh38
rs376702002 Chr.2: 147938303-147938303 on Build GRCh38
rs779304867 Chr.2: 147938395-147938395 on Build GRCh38
rs776929970 Chr.2: 147938327-147938327 on Build GRCh38
rs757684431 Chr.2: 147938390-147938390 on Build GRCh38
rs556939226 Chr.2: 147938408-147938408 on Build GRCh38
rs760109770 Chr.2: 147938368-147938368 on Build GRCh38
rs774183728 Chr.2: 147938333-147938335 on Build GRCh38
rs757557596 Chr.2: 147938278-147938278 on Build GRCh38
rs983460524 Chr.2: 147938423-147938423 on Build GRCh38
rs747012109 Chr.2: 147938301-147938301 on Build GRCh38
rs373470815 Chr.2: 147938417-147938417 on Build GRCh38
rs759491574 Chr.2: 147938365-147938365 on Build GRCh38
rs774224763 Chr.2: 147938358-147938358 on Build GRCh38
rs754507961 Chr.2: 147938242-147938242 on Build GRCh38
rs767381904 Chr.2: 147938377-147938377 on Build GRCh38
rs751459659 Chr.2: 147938241-147938241 on Build GRCh38
rs768801002 Chr.2: 147938418-147938418 on Build GRCh38
rs771780152 Chr.2: 147938368-147938371 on Build GRCh38
rs201791438 Chr.2: 147938298-147938298 on Build GRCh38
rs767305708 Chr.2: 147938366-147938366 on Build GRCh38
rs763748239 Chr.2: 147938373-147938373 on Build GRCh38

View Full Product Details