Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 93399304-93399529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT4
Entrez Gene ID
11163
Gene Name
nudix hydrolase 4
Gene Aliases
DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P
UniGene
Hs.506325
Gene Chromosome Location
Chr.12: 93377923-93403248 on Build GRCh38
Transcripts
NM_001301022.1 NM_001301023.1 NM_001301024.1 NM_019094.5 NM_199040.3 XM_011537794.2 XM_011537795.2 XM_011537796.2 NR_104005.1

Gene Symbol
NUDT4P2
Entrez Gene ID
170688
Gene Name
nudix hydrolase 4 pseudogene 2
Gene Aliases
-
UniGene
Hs.601274
Gene Chromosome Location
Chr.1: 146297773-146301610 on Build GRCh38
Transcripts
NM_001301022.1 NM_001301023.1 NM_001301024.1 NM_019094.5 NM_199040.3 XM_011537794.2 XM_011537795.2 XM_011537796.2 NR_104005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466502 c.477C>A p.S159S Chr.12: 93399310-93399310 on Build GRCh38
3466503 c.478C>A p.L160I Chr.12: 93399311-93399311 on Build GRCh38
3466504 c.482C>T p.P161L Chr.12: 93399315-93399315 on Build GRCh38
6731193 c.494C>T p.A165V Chr.12: 93399327-93399327 on Build GRCh38
1299969 c.486T>G p.D162E Chr.12: 93399319-93399319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139272816 Chr.12: 93399344-93399344 on Build GRCh38
rs375896561 Chr.12: 93399410-93399410 on Build GRCh38
rs765208039 Chr.12: 93399355-93399355 on Build GRCh38
rs779443667 Chr.12: 93399411-93399411 on Build GRCh38
rs760804361 Chr.12: 93399386-93399386 on Build GRCh38
rs34871538 Chr.12: 93399387-93399388 on Build GRCh38
rs536039988 Chr.12: 93399444-93399444 on Build GRCh38
rs751209568 Chr.12: 93399368-93399368 on Build GRCh38
rs762821513 Chr.12: 93399392-93399392 on Build GRCh38
rs749760554 Chr.12: 93399407-93399407 on Build GRCh38
rs776607930 Chr.12: 93399422-93399422 on Build GRCh38
rs747643894 Chr.12: 93399373-93399373 on Build GRCh38
rs748933858 Chr.12: 93399378-93399379 on Build GRCh38
rs777166707 Chr.12: 93399406-93399406 on Build GRCh38
rs142846389 Chr.12: 93399354-93399354 on Build GRCh38
rs771274514 Chr.12: 93399381-93399381 on Build GRCh38
rs200062744 Chr.12: 93399402-93399402 on Build GRCh38
rs375056789 Chr.12: 93399387-93399387 on Build GRCh38
rs768734502 Chr.12: 93399421-93399421 on Build GRCh38
rs755589515 Chr.12: 93399405-93399405 on Build GRCh38
rs768225095 Chr.12: 93399330-93399330 on Build GRCh38
rs567601229 Chr.12: 93399437-93399437 on Build GRCh38
rs770307321 Chr.12: 93399339-93399339 on Build GRCh38
rs767543283 Chr.12: 93399351-93399351 on Build GRCh38
rs758184271 Chr.12: 93399327-93399327 on Build GRCh38
rs556286521 Chr.12: 93399445-93399445 on Build GRCh38
rs773144969 Chr.12: 93399390-93399390 on Build GRCh38
rs759772901 Chr.12: 93399350-93399350 on Build GRCh38
rs777617004 Chr.12: 93399378-93399381 on Build GRCh38
rs769250241 Chr.12: 93399376-93399376 on Build GRCh38
rs779752048 Chr.12: 93399328-93399328 on Build GRCh38
rs774494362 Chr.12: 93399349-93399349 on Build GRCh38
rs774727094 Chr.12: 93399382-93399382 on Build GRCh38
rs771200195 Chr.12: 93399348-93399348 on Build GRCh38
rs772258463 Chr.12: 93399384-93399384 on Build GRCh38
rs778271680 Chr.12: 93399377-93399377 on Build GRCh38
rs767269898 Chr.12: 93399401-93399401 on Build GRCh38
rs373753344 Chr.12: 93399369-93399369 on Build GRCh38
rs780873833 Chr.12: 93399334-93399334 on Build GRCh38
rs200760978 Chr.12: 93399338-93399338 on Build GRCh38
rs758145881 Chr.12: 93399365-93399365 on Build GRCh38
rs772492422 Chr.12: 93399419-93399419 on Build GRCh38
rs569770913 Chr.12: 93399501-93399501 on Build GRCh38
rs766238083 Chr.12: 93399393-93399393 on Build GRCh38
rs746218271 Chr.12: 93399412-93399412 on Build GRCh38
rs750394465 Chr.12: 93399360-93399360 on Build GRCh38
rs754872327 Chr.12: 93399396-93399396 on Build GRCh38
rs773643984 Chr.12: 93399341-93399341 on Build GRCh38
rs749770180 Chr.12: 93399379-93399379 on Build GRCh38
rs775745707 Chr.12: 93399385-93399385 on Build GRCh38
rs747210238 Chr.12: 93399420-93399420 on Build GRCh38
rs746665041 Chr.12: 93399329-93399329 on Build GRCh38
rs879126643 Chr.12: 93399447-93399447 on Build GRCh38

View Full Product Details