Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111559336-111559609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF3IP2
Entrez Gene ID
10758
Gene Name
TRAF3 interacting protein 2
Gene Aliases
ACT1, C6orf2, C6orf4, C6orf5, C6orf6, CANDF8, CIKS, PSORS13
UniGene
Hs.561514
Gene Chromosome Location
Chr.6: 111555378-111606874 on Build GRCh38
Transcripts
NM_001164281.2 NM_001164283.2 NM_147200.2 NM_147686.3 XM_006715319.3 XM_011535386.1 NR_034108.1 NR_034109.1 NR_034110.1

Gene Symbol
TRAF3IP2-AS1
Entrez Gene ID
643749
Gene Name
TRAF3IP2 antisense RNA 1
Gene Aliases
C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2
UniGene
Hs.486228
Gene Chromosome Location
Chr.6: 111483472-111602295 on Build GRCh38
Transcripts
NM_001164281.2 NM_001164283.2 NM_147200.2 NM_147686.3 XM_006715319.3 XM_011535386.1 NR_034108.1 NR_034109.1 NR_034110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321205 c.1618C>A p.L540M Chr.6: 111559485-111559485 on Build GRCh38
6226111 c.1666C>G p.P556A Chr.6: 111559437-111559437 on Build GRCh38
6418027 c.1625G>T p.R542L Chr.6: 111559478-111559478 on Build GRCh38
39911 c.1626G>A p.R542R Chr.6: 111559477-111559477 on Build GRCh38
5640932 c.1583A>T p.H528L Chr.6: 111559520-111559520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770569685 Chr.6: 111559382-111559382 on Build GRCh38
rs745406713 Chr.6: 111559564-111559564 on Build GRCh38
rs748766419 Chr.6: 111559380-111559380 on Build GRCh38
rs777439425 Chr.6: 111559490-111559490 on Build GRCh38
rs766494097 Chr.6: 111559461-111559461 on Build GRCh38
rs202090066 Chr.6: 111559520-111559520 on Build GRCh38
rs1050358036 Chr.6: 111559573-111559573 on Build GRCh38
rs910982011 Chr.6: 111559361-111559361 on Build GRCh38
rs778653390 Chr.6: 111559386-111559386 on Build GRCh38
rs148316010 Chr.6: 111559479-111559479 on Build GRCh38
rs938108507 Chr.6: 111559362-111559362 on Build GRCh38
rs746647201 Chr.6: 111559497-111559498 on Build GRCh38
rs769958417 Chr.6: 111559429-111559429 on Build GRCh38
rs772026502 Chr.6: 111559571-111559571 on Build GRCh38
rs760085841 Chr.6: 111559478-111559478 on Build GRCh38
rs753055967 Chr.6: 111559513-111559513 on Build GRCh38
rs186097801 Chr.6: 111559489-111559489 on Build GRCh38
rs370109607 Chr.6: 111559401-111559401 on Build GRCh38
rs754257355 Chr.6: 111559402-111559403 on Build GRCh38
rs760514857 Chr.6: 111559426-111559426 on Build GRCh38
rs377585744 Chr.6: 111559547-111559547 on Build GRCh38
rs777590131 Chr.6: 111559375-111559375 on Build GRCh38
rs757024960 Chr.6: 111559556-111559556 on Build GRCh38
rs762959079 Chr.6: 111559449-111559449 on Build GRCh38
rs763055593 Chr.6: 111559400-111559400 on Build GRCh38
rs989603351 Chr.6: 111559408-111559408 on Build GRCh38
rs397518485 Chr.6: 111559523-111559523 on Build GRCh38
rs202020640 Chr.6: 111559557-111559557 on Build GRCh38
rs777500416 Chr.6: 111559534-111559534 on Build GRCh38
rs143248990 Chr.6: 111559411-111559411 on Build GRCh38
rs545921552 Chr.6: 111559367-111559367 on Build GRCh38
rs763984264 Chr.6: 111559495-111559495 on Build GRCh38
rs755328202 Chr.6: 111559409-111559410 on Build GRCh38
rs751648986 Chr.6: 111559468-111559468 on Build GRCh38
rs773458653 Chr.6: 111559443-111559443 on Build GRCh38
rs879018565 Chr.6: 111559471-111559471 on Build GRCh38

View Full Product Details