Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94664742-94664989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRP1
Entrez Gene ID
54845
Gene Name
epithelial splicing regulatory protein 1
Gene Aliases
RBM35A, RMB35A
UniGene
Hs.487471
Gene Chromosome Location
Chr.8: 94640999-94707466 on Build GRCh38
Transcripts
NM_001034915.2 NM_001122825.1 NM_001122826.1 NM_001122827.1 NM_017697.3 XM_005250991.3 XM_005250992.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4567639 c.799_800GG>AA p.G267K Chr.8: 94664970-94664971 on Build GRCh38
4567638 c.799_800GG>AA p.G267K Chr.8: 94664970-94664971 on Build GRCh38
6495630 c.718G>C p.D240H Chr.8: 94664770-94664770 on Build GRCh38
1102625 c.702G>A p.W234* Chr.8: 94664754-94664754 on Build GRCh38
6656433 c.756-1G>C p.? Chr.8: 94664926-94664926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758523117 Chr.8: 94664915-94664915 on Build GRCh38
rs753931997 Chr.8: 94664858-94664858 on Build GRCh38
rs1050406759 Chr.8: 94664881-94664881 on Build GRCh38
rs917019326 Chr.8: 94664778-94664778 on Build GRCh38
rs753964744 Chr.8: 94664940-94664940 on Build GRCh38
rs776321804 Chr.8: 94664886-94664886 on Build GRCh38
rs753884002 Chr.8: 94664886-94664887 on Build GRCh38
rs757268720 Chr.8: 94664945-94664945 on Build GRCh38
rs372594370 Chr.8: 94664882-94664882 on Build GRCh38
rs755864138 Chr.8: 94664883-94664884 on Build GRCh38
rs767790309 Chr.8: 94664908-94664908 on Build GRCh38
rs945818281 Chr.8: 94664809-94664809 on Build GRCh38
rs749322470 Chr.8: 94664842-94664842 on Build GRCh38
rs756560428 Chr.8: 94664833-94664833 on Build GRCh38
rs768676144 Chr.8: 94664884-94664884 on Build GRCh38
rs761386979 Chr.8: 94664903-94664903 on Build GRCh38
rs537588168 Chr.8: 94664920-94664920 on Build GRCh38
rs1426171 Chr.8: 94664904-94664904 on Build GRCh38
rs561435477 Chr.8: 94664880-94664880 on Build GRCh38
rs368010707 Chr.8: 94664905-94664905 on Build GRCh38
rs765366869 Chr.8: 94664822-94664822 on Build GRCh38
rs182950981 Chr.8: 94664915-94664915 on Build GRCh38
rs768139935 Chr.8: 94664793-94664793 on Build GRCh38
rs764309275 Chr.8: 94664835-94664835 on Build GRCh38
rs148374001 Chr.8: 94664852-94664852 on Build GRCh38
rs1009378512 Chr.8: 94664934-94664934 on Build GRCh38
rs752006196 Chr.8: 94664786-94664786 on Build GRCh38
rs745731638 Chr.8: 94664854-94664854 on Build GRCh38
rs753209922 Chr.8: 94664811-94664811 on Build GRCh38
rs200846428 Chr.8: 94664878-94664878 on Build GRCh38
rs780036078 Chr.8: 94664887-94664887 on Build GRCh38
rs75787732 Chr.8: 94664887-94664887 on Build GRCh38
rs550957467 Chr.8: 94664954-94664954 on Build GRCh38
rs750612777 Chr.8: 94664968-94664968 on Build GRCh38
rs750173226 Chr.8: 94664837-94664839 on Build GRCh38
rs376791493 Chr.8: 94664951-94664951 on Build GRCh38
rs568593667 Chr.8: 94664917-94664917 on Build GRCh38
rs755285759 Chr.8: 94664789-94664789 on Build GRCh38

View Full Product Details