Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131274702-131274975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
UniGene
Hs.22111
Gene Chromosome Location
Chr.X: 131273506-131289457 on Build GRCh38
Transcripts
NM_001170961.1 NM_001170962.1 NM_001555.4 XM_011531330.1 XM_011531333.1 XM_011531334.1

SNP Detail

SNP ID Chromosome Location
rs370446413 Chr.X: 131274786-131274786 on Build GRCh38
rs368422973 Chr.X: 131274912-131274912 on Build GRCh38
rs753208618 Chr.X: 131274745-131274745 on Build GRCh38
rs200127298 Chr.X: 131274811-131274811 on Build GRCh38
rs758376599 Chr.X: 131274926-131274926 on Build GRCh38
rs373571972 Chr.X: 131274789-131274789 on Build GRCh38
rs6529473 Chr.X: 131274771-131274771 on Build GRCh38
rs775724672 Chr.X: 131274873-131274873 on Build GRCh38
rs267606353 Chr.X: 131274851-131274851 on Build GRCh38
rs765697743 Chr.X: 131274734-131274734 on Build GRCh38
rs112522172 Chr.X: 131274843-131274843 on Build GRCh38
rs199540826 Chr.X: 131274910-131274910 on Build GRCh38
rs773045436 Chr.X: 131274792-131274792 on Build GRCh38
rs1003501926 Chr.X: 131274907-131274907 on Build GRCh38
rs758849300 Chr.X: 131274746-131274746 on Build GRCh38
rs376260455 Chr.X: 131274751-131274751 on Build GRCh38
rs373984167 Chr.X: 131274891-131274891 on Build GRCh38
rs956426611 Chr.X: 131274936-131274936 on Build GRCh38
rs146525641 Chr.X: 131274799-131274799 on Build GRCh38
rs376897895 Chr.X: 131274760-131274760 on Build GRCh38
rs745964986 Chr.X: 131274763-131274763 on Build GRCh38
rs201123183 Chr.X: 131274850-131274850 on Build GRCh38
rs988195384 Chr.X: 131274938-131274938 on Build GRCh38
rs781726830 Chr.X: 131274897-131274898 on Build GRCh38
rs371761367 Chr.X: 131274887-131274887 on Build GRCh38
rs776147631 Chr.X: 131274824-131274824 on Build GRCh38
rs746780456 Chr.X: 131274797-131274797 on Build GRCh38
rs750599205 Chr.X: 131274881-131274881 on Build GRCh38
rs764685434 Chr.X: 131274835-131274835 on Build GRCh38
rs763622284 Chr.X: 131274828-131274828 on Build GRCh38
rs398122921 Chr.X: 131274768-131274769 on Build GRCh38

View Full Product Details