Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12940598-12940847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALR
Entrez Gene ID
811
Gene Name
calreticulin
Gene Aliases
CRT, HEL-S-99n, RO, SSA, cC1qR
UniGene
Hs.515162
Gene Chromosome Location
Chr.19: 12938600-12944490 on Build GRCh38
Transcripts
NM_004343.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
991545 c.792A>G p.P264P Chr.19: 12940630-12940630 on Build GRCh38
2817018 c.909C>T p.P303P Chr.19: 12940836-12940836 on Build GRCh38
5343202 c.775G>A p.G259R Chr.19: 12940613-12940613 on Build GRCh38
6619142 c.769A>G p.M257V Chr.19: 12940607-12940607 on Build GRCh38
1390691 c.787delC p.P264fs*2 Chr.19: 12940625-12940625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770320546 Chr.19: 12940817-12940817 on Build GRCh38
rs374885707 Chr.19: 12940721-12940721 on Build GRCh38
rs184881678 Chr.19: 12940740-12940740 on Build GRCh38
rs956599530 Chr.19: 12940685-12940685 on Build GRCh38
rs769245573 Chr.19: 12940626-12940626 on Build GRCh38
rs766667846 Chr.19: 12940745-12940745 on Build GRCh38
rs11547562 Chr.19: 12940785-12940785 on Build GRCh38
rs367698874 Chr.19: 12940731-12940731 on Build GRCh38
rs148447453 Chr.19: 12940625-12940625 on Build GRCh38
rs113302836 Chr.19: 12940656-12940656 on Build GRCh38
rs373808786 Chr.19: 12940698-12940698 on Build GRCh38
rs778397758 Chr.19: 12940707-12940707 on Build GRCh38
rs878983219 Chr.19: 12940663-12940663 on Build GRCh38
rs879035965 Chr.19: 12940741-12940741 on Build GRCh38
rs752670678 Chr.19: 12940682-12940682 on Build GRCh38
rs1030686954 Chr.19: 12940659-12940659 on Build GRCh38
rs756041024 Chr.19: 12940690-12940690 on Build GRCh38
rs879111760 Chr.19: 12940741-12940742 on Build GRCh38
rs372688428 Chr.19: 12940636-12940636 on Build GRCh38
rs759720623 Chr.19: 12940759-12940759 on Build GRCh38
rs767611655 Chr.19: 12940677-12940677 on Build GRCh38
rs777096782 Chr.19: 12940648-12940648 on Build GRCh38
rs757976423 Chr.19: 12940808-12940808 on Build GRCh38
rs756980463 Chr.19: 12940780-12940780 on Build GRCh38
rs777186453 Chr.19: 12940729-12940729 on Build GRCh38
rs774414495 Chr.19: 12940747-12940747 on Build GRCh38
rs766352310 Chr.19: 12940624-12940625 on Build GRCh38
rs778115432 Chr.19: 12940818-12940818 on Build GRCh38
rs752760579 Chr.19: 12940765-12940765 on Build GRCh38
rs745429234 Chr.19: 12940709-12940709 on Build GRCh38
rs751695581 Chr.19: 12940674-12940674 on Build GRCh38
rs765579713 Chr.19: 12940657-12940657 on Build GRCh38
rs748984710 Chr.19: 12940693-12940693 on Build GRCh38
rs766466189 Chr.19: 12940671-12940671 on Build GRCh38
rs548980168 Chr.19: 12940733-12940733 on Build GRCh38
rs753706198 Chr.19: 12940768-12940768 on Build GRCh38
rs940720149 Chr.19: 12940790-12940790 on Build GRCh38
rs779550076 Chr.19: 12940810-12940810 on Build GRCh38
rs746475957 Chr.19: 12940812-12940812 on Build GRCh38
rs754985334 Chr.19: 12940675-12940675 on Build GRCh38
rs763253932 Chr.19: 12940666-12940666 on Build GRCh38
rs767701623 Chr.19: 12940760-12940760 on Build GRCh38
rs904154287 Chr.19: 12940738-12940738 on Build GRCh38
rs773495787 Chr.19: 12940661-12940661 on Build GRCh38
rs1022036281 Chr.19: 12940711-12940714 on Build GRCh38
rs760708514 Chr.19: 12940767-12940767 on Build GRCh38
rs367922867 Chr.19: 12940744-12940744 on Build GRCh38
rs771465997 Chr.19: 12940721-12940721 on Build GRCh38
rs778680432 Chr.19: 12940787-12940787 on Build GRCh38
rs879207308 Chr.19: 12940662-12940662 on Build GRCh38
rs907856910 Chr.19: 12940763-12940763 on Build GRCh38
rs915643856 Chr.19: 12940722-12940722 on Build GRCh38
rs74815302 Chr.19: 12940735-12940735 on Build GRCh38

View Full Product Details