Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7593228-7593501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXR2
Entrez Gene ID
9513
Gene Name
FMR1 autosomal homolog 2
Gene Aliases
FMR1L2, FXR2P
UniGene
Hs.52788
Gene Chromosome Location
Chr.17: 7591230-7614897 on Build GRCh38
Transcripts
NM_004860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043708 c.1294C>T p.R432C Chr.17: 7593439-7593439 on Build GRCh38
288660 c.1268G>A p.R423Q Chr.17: 7593465-7593465 on Build GRCh38
984807 c.1299C>T p.G433G Chr.17: 7593434-7593434 on Build GRCh38
7368963 c.1241G>A p.S414N Chr.17: 7593492-7593492 on Build GRCh38
6309686 c.1307G>A p.R436Q Chr.17: 7593426-7593426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772553286 Chr.17: 7593381-7593381 on Build GRCh38
rs902507122 Chr.17: 7593470-7593470 on Build GRCh38
rs376313217 Chr.17: 7593480-7593480 on Build GRCh38
rs545121685 Chr.17: 7593423-7593423 on Build GRCh38
rs867137331 Chr.17: 7593449-7593449 on Build GRCh38
rs967395105 Chr.17: 7593255-7593255 on Build GRCh38
rs950536080 Chr.17: 7593289-7593289 on Build GRCh38
rs1052322715 Chr.17: 7593354-7593354 on Build GRCh38
rs868233074 Chr.17: 7593414-7593414 on Build GRCh38
rs368455716 Chr.17: 7593416-7593416 on Build GRCh38
rs764814837 Chr.17: 7593427-7593427 on Build GRCh38
rs987992365 Chr.17: 7593249-7593249 on Build GRCh38
rs746972144 Chr.17: 7593437-7593438 on Build GRCh38
rs768030074 Chr.17: 7593439-7593440 on Build GRCh38
rs868253602 Chr.17: 7593439-7593439 on Build GRCh38
rs761315421 Chr.17: 7593390-7593390 on Build GRCh38
rs372345788 Chr.17: 7593447-7593447 on Build GRCh38
rs569146094 Chr.17: 7593292-7593292 on Build GRCh38
rs750150579 Chr.17: 7593438-7593438 on Build GRCh38
rs781040872 Chr.17: 7593454-7593454 on Build GRCh38
rs759661963 Chr.17: 7593372-7593372 on Build GRCh38
rs935222979 Chr.17: 7593342-7593342 on Build GRCh38
rs562549549 Chr.17: 7593251-7593251 on Build GRCh38
rs991419231 Chr.17: 7593309-7593309 on Build GRCh38
rs775967678 Chr.17: 7593386-7593386 on Build GRCh38
rs778853355 Chr.17: 7593477-7593477 on Build GRCh38
rs939207524 Chr.17: 7593413-7593413 on Build GRCh38
rs757413563 Chr.17: 7593473-7593473 on Build GRCh38
rs754935726 Chr.17: 7593448-7593448 on Build GRCh38
rs916528773 Chr.17: 7593432-7593432 on Build GRCh38
rs935309545 Chr.17: 7593471-7593471 on Build GRCh38
rs780797161 Chr.17: 7593440-7593451 on Build GRCh38
rs749341461 Chr.17: 7593468-7593468 on Build GRCh38
rs978583552 Chr.17: 7593263-7593265 on Build GRCh38
rs990586128 Chr.17: 7593422-7593422 on Build GRCh38
rs757985705 Chr.17: 7593442-7593442 on Build GRCh38
rs949754702 Chr.17: 7593343-7593343 on Build GRCh38
rs558270590 Chr.17: 7593279-7593279 on Build GRCh38
rs779926823 Chr.17: 7593426-7593431 on Build GRCh38
rs761324408 Chr.17: 7593426-7593426 on Build GRCh38
rs916934453 Chr.17: 7593329-7593329 on Build GRCh38
rs996873653 Chr.17: 7593451-7593451 on Build GRCh38
rs747749455 Chr.17: 7593479-7593481 on Build GRCh38

View Full Product Details