Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 115231184-115231437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM1
Entrez Gene ID
23705
Gene Name
cell adhesion molecule 1
Gene Aliases
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
UniGene
Hs.370510
Gene Chromosome Location
Chr.11: 115173625-115504523 on Build GRCh38
Transcripts
NM_001098517.1 NM_001301043.1 NM_001301044.1 NM_001301045.1 NM_014333.3 XM_005271494.2 XM_017017457.1 XM_017017458.1 XM_017017459.1 XM_017017460.1 XM_017017461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7429711 c.515A>C p.K172T Chr.11: 115231400-115231400 on Build GRCh38
6618835 c.508G>A p.A170T Chr.11: 115231407-115231407 on Build GRCh38
686270 c.531C>T p.I177I Chr.11: 115231384-115231384 on Build GRCh38
4760402 c.520G>A p.A174T Chr.11: 115231395-115231395 on Build GRCh38
6131324 c.495C>A p.N165K Chr.11: 115231420-115231420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929433705 Chr.11: 115231301-115231301 on Build GRCh38
rs548644814 Chr.11: 115231343-115231343 on Build GRCh38
rs373090749 Chr.11: 115231366-115231366 on Build GRCh38
rs533460943 Chr.11: 115231255-115231255 on Build GRCh38
rs780425374 Chr.11: 115231370-115231370 on Build GRCh38
rs528890135 Chr.11: 115231306-115231306 on Build GRCh38
rs150028427 Chr.11: 115231221-115231221 on Build GRCh38
rs749617712 Chr.11: 115231407-115231407 on Build GRCh38
rs764291402 Chr.11: 115231384-115231384 on Build GRCh38
rs770893654 Chr.11: 115231392-115231392 on Build GRCh38
rs753250226 Chr.11: 115231302-115231302 on Build GRCh38
rs184752303 Chr.11: 115231215-115231215 on Build GRCh38
rs748514117 Chr.11: 115231373-115231373 on Build GRCh38
rs559882843 Chr.11: 115231295-115231295 on Build GRCh38
rs1034830591 Chr.11: 115231284-115231284 on Build GRCh38
rs751706262 Chr.11: 115231391-115231391 on Build GRCh38
rs1027248468 Chr.11: 115231388-115231388 on Build GRCh38
rs376372384 Chr.11: 115231354-115231354 on Build GRCh38
rs189470146 Chr.11: 115231268-115231268 on Build GRCh38
rs547973804 Chr.11: 115231331-115231331 on Build GRCh38
rs1002989259 Chr.11: 115231269-115231269 on Build GRCh38
rs756584318 Chr.11: 115231386-115231386 on Build GRCh38
rs774683876 Chr.11: 115231303-115231303 on Build GRCh38
rs774599665 Chr.11: 115231410-115231410 on Build GRCh38
rs1037773872 Chr.11: 115231265-115231265 on Build GRCh38
rs757838824 Chr.11: 115231350-115231350 on Build GRCh38
rs770225289 Chr.11: 115231283-115231283 on Build GRCh38
rs776603083 Chr.11: 115231309-115231309 on Build GRCh38
rs758401125 Chr.11: 115231371-115231371 on Build GRCh38
rs751187404 Chr.11: 115231357-115231357 on Build GRCh38
rs761745748 Chr.11: 115231327-115231327 on Build GRCh38
rs765103367 Chr.11: 115231335-115231335 on Build GRCh38
rs568632394 Chr.11: 115231390-115231390 on Build GRCh38

View Full Product Details