Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 97888044-97888317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATN2
Entrez Gene ID
4147
Gene Name
matrilin 2
Gene Aliases
-
UniGene
Hs.189445
Gene Chromosome Location
Chr.8: 97869021-98036720 on Build GRCh38
Transcripts
NM_001317748.1 NM_002380.4 NM_030583.3 XM_005250920.1 XM_017013417.1 XM_017013418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6214188 c.48C>T p.I16I Chr.8: 97888148-97888148 on Build GRCh38
454993 c.64G>A p.E22K Chr.8: 97888164-97888164 on Build GRCh38
7376493 c.41G>C p.G14A Chr.8: 97888141-97888141 on Build GRCh38
3902308 c.115C>T p.R39W Chr.8: 97888215-97888215 on Build GRCh38
2792333 c.39C>T p.L13L Chr.8: 97888139-97888139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756404500 Chr.8: 97888285-97888285 on Build GRCh38
rs576660499 Chr.8: 97888269-97888269 on Build GRCh38
rs376715058 Chr.8: 97888139-97888139 on Build GRCh38
rs755486597 Chr.8: 97888271-97888271 on Build GRCh38
rs375784459 Chr.8: 97888220-97888220 on Build GRCh38
rs549943812 Chr.8: 97888154-97888154 on Build GRCh38
rs536601427 Chr.8: 97888226-97888226 on Build GRCh38
rs112395555 Chr.8: 97888250-97888250 on Build GRCh38
rs367644090 Chr.8: 97888096-97888096 on Build GRCh38
rs748507015 Chr.8: 97888277-97888277 on Build GRCh38
rs200952405 Chr.8: 97888203-97888203 on Build GRCh38
rs556327640 Chr.8: 97888258-97888258 on Build GRCh38
rs201879642 Chr.8: 97888177-97888177 on Build GRCh38
rs766263568 Chr.8: 97888163-97888163 on Build GRCh38
rs775483490 Chr.8: 97888232-97888232 on Build GRCh38
rs370121449 Chr.8: 97888256-97888256 on Build GRCh38
rs892663998 Chr.8: 97888206-97888206 on Build GRCh38
rs200041053 Chr.8: 97888182-97888182 on Build GRCh38
rs924954196 Chr.8: 97888278-97888278 on Build GRCh38
rs567767391 Chr.8: 97888225-97888225 on Build GRCh38
rs778619387 Chr.8: 97888180-97888180 on Build GRCh38
rs751475828 Chr.8: 97888165-97888165 on Build GRCh38
rs534833968 Chr.8: 97888166-97888166 on Build GRCh38
rs373581202 Chr.8: 97888276-97888276 on Build GRCh38
rs764838730 Chr.8: 97888260-97888260 on Build GRCh38
rs747554325 Chr.8: 97888117-97888117 on Build GRCh38
rs373506520 Chr.8: 97888172-97888172 on Build GRCh38
rs755790601 Chr.8: 97888079-97888079 on Build GRCh38
rs186093758 Chr.8: 97888148-97888148 on Build GRCh38
rs980171732 Chr.8: 97888216-97888216 on Build GRCh38
rs372776833 Chr.8: 97888211-97888211 on Build GRCh38
rs946341581 Chr.8: 97888262-97888262 on Build GRCh38
rs777273761 Chr.8: 97888080-97888080 on Build GRCh38
rs775304395 Chr.8: 97888231-97888231 on Build GRCh38
rs1034491467 Chr.8: 97888219-97888219 on Build GRCh38
rs752438138 Chr.8: 97888169-97888169 on Build GRCh38
rs763985941 Chr.8: 97888243-97888243 on Build GRCh38
rs35804177 Chr.8: 97888141-97888141 on Build GRCh38
rs753344547 Chr.8: 97888176-97888176 on Build GRCh38
rs936423497 Chr.8: 97888085-97888085 on Build GRCh38
rs778259029 Chr.8: 97888290-97888290 on Build GRCh38
rs748477428 Chr.8: 97888186-97888186 on Build GRCh38
rs769115950 Chr.8: 97888118-97888118 on Build GRCh38
rs897104851 Chr.8: 97888155-97888155 on Build GRCh38
rs755612604 Chr.8: 97888183-97888183 on Build GRCh38
rs773283216 Chr.8: 97888152-97888152 on Build GRCh38
rs11559203 Chr.8: 97888100-97888100 on Build GRCh38
rs780741252 Chr.8: 97888116-97888116 on Build GRCh38
rs1009309714 Chr.8: 97888217-97888217 on Build GRCh38

View Full Product Details