Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149886204-149886439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6A
Entrez Gene ID
5145
Gene Name
phosphodiesterase 6A
Gene Aliases
CGPR-A, PDEA, RP43
UniGene
Hs.567314
Gene Chromosome Location
Chr.5: 149857956-149944793 on Build GRCh38
Transcripts
NM_000440.2 XM_011537650.2 XM_011537651.2 XM_011537652.1 XM_011537653.2 XM_011537654.2 XM_017009572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5812975 c.1826T>A p.L609H Chr.5: 149886277-149886277 on Build GRCh38
3211744 c.1762G>C p.E588Q Chr.5: 149886341-149886341 on Build GRCh38
149987 c.1791C>T p.F597F Chr.5: 149886312-149886312 on Build GRCh38
215968 c.1730C>T p.T577M Chr.5: 149886373-149886373 on Build GRCh38
3852687 c.1731G>A p.T577T Chr.5: 149886372-149886372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753002443 Chr.5: 149886351-149886356 on Build GRCh38
rs121918576 Chr.5: 149886354-149886354 on Build GRCh38
rs761600125 Chr.5: 149886303-149886303 on Build GRCh38
rs982392860 Chr.5: 149886230-149886230 on Build GRCh38
rs191628052 Chr.5: 149886347-149886347 on Build GRCh38
rs773601046 Chr.5: 149886279-149886279 on Build GRCh38
rs368690470 Chr.5: 149886367-149886367 on Build GRCh38
rs754543374 Chr.5: 149886404-149886404 on Build GRCh38
rs763461797 Chr.5: 149886287-149886287 on Build GRCh38
rs752872810 Chr.5: 149886301-149886301 on Build GRCh38
rs770977429 Chr.5: 149886288-149886288 on Build GRCh38
rs765007196 Chr.5: 149886373-149886373 on Build GRCh38
rs765024330 Chr.5: 149886305-149886305 on Build GRCh38
rs758350163 Chr.5: 149886319-149886319 on Build GRCh38
rs921058026 Chr.5: 149886262-149886262 on Build GRCh38
rs139011078 Chr.5: 149886348-149886348 on Build GRCh38
rs767744053 Chr.5: 149886300-149886300 on Build GRCh38
rs142934552 Chr.5: 149886267-149886267 on Build GRCh38
rs562967163 Chr.5: 149886363-149886363 on Build GRCh38
rs371975833 Chr.5: 149886392-149886392 on Build GRCh38
rs760874208 Chr.5: 149886372-149886372 on Build GRCh38
rs370425142 Chr.5: 149886231-149886231 on Build GRCh38
rs775702453 Chr.5: 149886371-149886371 on Build GRCh38
rs755789997 Chr.5: 149886414-149886414 on Build GRCh38
rs573153907 Chr.5: 149886289-149886289 on Build GRCh38
rs770246880 Chr.5: 149886277-149886277 on Build GRCh38
rs374867097 Chr.5: 149886237-149886237 on Build GRCh38
rs778045905 Chr.5: 149886418-149886418 on Build GRCh38
rs148785740 Chr.5: 149886359-149886359 on Build GRCh38
rs61733360 Chr.5: 149886312-149886312 on Build GRCh38
rs779471780 Chr.5: 149886326-149886326 on Build GRCh38
rs768394732 Chr.5: 149886251-149886251 on Build GRCh38
rs75283260 Chr.5: 149886240-149886240 on Build GRCh38
rs779601968 Chr.5: 149886245-149886245 on Build GRCh38
rs781724083 Chr.5: 149886254-149886254 on Build GRCh38
rs746726551 Chr.5: 149886246-149886246 on Build GRCh38
rs555052906 Chr.5: 149886378-149886378 on Build GRCh38
rs773030606 Chr.5: 149886383-149886383 on Build GRCh38
rs150511819 Chr.5: 149886360-149886360 on Build GRCh38
rs759801201 Chr.5: 149886296-149886296 on Build GRCh38
rs77760924 Chr.5: 149886358-149886358 on Build GRCh38
rs367796771 Chr.5: 149886389-149886389 on Build GRCh38
rs751503649 Chr.5: 149886393-149886393 on Build GRCh38
rs199509222 Chr.5: 149886234-149886234 on Build GRCh38
rs908179722 Chr.5: 149886415-149886415 on Build GRCh38
rs751094700 Chr.5: 149886331-149886331 on Build GRCh38
rs752458299 Chr.5: 149886412-149886412 on Build GRCh38
rs547792004 Chr.5: 149886349-149886349 on Build GRCh38
rs569313193 Chr.5: 149886243-149886243 on Build GRCh38
rs770071916 Chr.5: 149886355-149886355 on Build GRCh38

View Full Product Details