Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 86256818-86257063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL1
Entrez Gene ID
123624
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
UniGene
Hs.31795
Gene Chromosome Location
Chr.15: 86079635-87031476 on Build GRCh38
Transcripts
XM_011521226.2 XM_011521227.2 XM_011521228.2 XM_011521230.2 XM_017021918.1 XM_017021919.1 XM_017021920.1 XM_017021921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906558 c.683C>T p.P228L Chr.15: 86256938-86256938 on Build GRCh38
5456515 c.694G>A p.A232T Chr.15: 86256949-86256949 on Build GRCh38
5456516 c.694G>A p.A232T Chr.15: 86256949-86256949 on Build GRCh38
6522345 c.718G>A p.V240I Chr.15: 86256973-86256973 on Build GRCh38
6143260 c.725G>C p.G242A Chr.15: 86256980-86256980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779647718 Chr.15: 86257004-86257004 on Build GRCh38
rs1390866 Chr.15: 86256985-86256985 on Build GRCh38
rs199638717 Chr.15: 86256944-86256944 on Build GRCh38
rs772624518 Chr.15: 86257014-86257014 on Build GRCh38
rs750144535 Chr.15: 86256989-86256989 on Build GRCh38
rs368107286 Chr.15: 86256862-86256862 on Build GRCh38
rs375624311 Chr.15: 86256882-86256882 on Build GRCh38
rs200765292 Chr.15: 86256934-86256934 on Build GRCh38
rs372417529 Chr.15: 86256906-86256906 on Build GRCh38
rs138185152 Chr.15: 86257020-86257020 on Build GRCh38
rs369807683 Chr.15: 86256926-86256926 on Build GRCh38
rs762908428 Chr.15: 86256975-86256975 on Build GRCh38
rs906438402 Chr.15: 86256963-86256963 on Build GRCh38
rs1353578 Chr.15: 86256978-86256978 on Build GRCh38
rs756831984 Chr.15: 86256983-86256983 on Build GRCh38
rs750483799 Chr.15: 86256897-86256897 on Build GRCh38
rs755714357 Chr.15: 86256937-86256937 on Build GRCh38
rs748326793 Chr.15: 86256957-86256957 on Build GRCh38
rs369685746 Chr.15: 86256924-86256924 on Build GRCh38
rs752739323 Chr.15: 86256920-86256920 on Build GRCh38
rs760659339 Chr.15: 86256970-86256970 on Build GRCh38
rs183456981 Chr.15: 86256885-86256885 on Build GRCh38
rs372045551 Chr.15: 86256872-86256872 on Build GRCh38
rs141984932 Chr.15: 86256883-86256883 on Build GRCh38
rs190666485 Chr.15: 86256981-86256981 on Build GRCh38
rs200544184 Chr.15: 86257027-86257027 on Build GRCh38
rs935027822 Chr.15: 86256913-86256913 on Build GRCh38
rs749844541 Chr.15: 86256947-86256947 on Build GRCh38
rs767660369 Chr.15: 86256889-86256889 on Build GRCh38
rs758282474 Chr.15: 86256857-86256857 on Build GRCh38
rs755551642 Chr.15: 86257001-86257001 on Build GRCh38
rs567679729 Chr.15: 86256927-86256927 on Build GRCh38
rs1948571 Chr.15: 86256907-86256907 on Build GRCh38
rs755559068 Chr.15: 86257011-86257011 on Build GRCh38
rs746334255 Chr.15: 86256876-86256876 on Build GRCh38
rs770396517 Chr.15: 86256863-86256863 on Build GRCh38
rs762669952 Chr.15: 86256950-86256950 on Build GRCh38
rs199544767 Chr.15: 86256971-86256971 on Build GRCh38
rs765083113 Chr.15: 86256911-86256911 on Build GRCh38
rs185896474 Chr.15: 86256923-86256923 on Build GRCh38
rs150851131 Chr.15: 86256960-86256960 on Build GRCh38
rs762919204 Chr.15: 86256892-86256894 on Build GRCh38
rs775257741 Chr.15: 86256972-86256972 on Build GRCh38
rs540734125 Chr.15: 86256851-86256851 on Build GRCh38
rs4496106 Chr.15: 86256903-86256903 on Build GRCh38
rs770212726 Chr.15: 86256879-86256879 on Build GRCh38
rs779355087 Chr.15: 86257012-86257012 on Build GRCh38
rs936377293 Chr.15: 86256890-86256890 on Build GRCh38
rs1390867 Chr.15: 86256852-86256852 on Build GRCh38
rs764020090 Chr.15: 86257032-86257032 on Build GRCh38
rs749713329 Chr.15: 86257008-86257008 on Build GRCh38
rs780619899 Chr.15: 86256932-86256932 on Build GRCh38
rs946297996 Chr.15: 86256880-86256880 on Build GRCh38
rs539881376 Chr.15: 86256868-86256868 on Build GRCh38
rs1024622762 Chr.15: 86256961-86256961 on Build GRCh38
rs768460386 Chr.15: 86256966-86256966 on Build GRCh38
rs754374344 Chr.15: 86257031-86257031 on Build GRCh38
rs774882154 Chr.15: 86256954-86256954 on Build GRCh38
rs958933255 Chr.15: 86257024-86257024 on Build GRCh38
rs973861559 Chr.15: 86257009-86257009 on Build GRCh38
rs539107789 Chr.15: 86257018-86257018 on Build GRCh38

View Full Product Details