Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 109931854-109932109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALM2
Entrez Gene ID
114299
Gene Name
paralemmin 2
Gene Aliases
AKAP2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 109640788-109951476 on Build GRCh38
Transcripts
NM_001037293.2 NM_053016.5 NM_007203.4 NM_147150.2

Gene Symbol
PALM2-AKAP2
Entrez Gene ID
445815
Gene Name
PALM2-AKAP2 readthrough
Gene Aliases
AKAP2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 109780297-110172512 on Build GRCh38
Transcripts
NM_001037293.2 NM_053016.5 NM_007203.4 NM_147150.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3847302 c.465A>C p.S155S Chr.9: 109931997-109931997 on Build GRCh38
109007 c.413C>T p.S138F Chr.9: 109931945-109931945 on Build GRCh38
6349134 c.477C>T p.D159D Chr.9: 109932009-109932009 on Build GRCh38
273071 c.443G>A p.C148Y Chr.9: 109931975-109931975 on Build GRCh38
5784323 c.496G>A p.G166R Chr.9: 109932028-109932028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186559507 Chr.9: 109932051-109932051 on Build GRCh38
rs748723983 Chr.9: 109931998-109931998 on Build GRCh38
rs771281592 Chr.9: 109932010-109932010 on Build GRCh38
rs766745626 Chr.9: 109931980-109931980 on Build GRCh38
rs372422935 Chr.9: 109932024-109932024 on Build GRCh38
rs748547261 Chr.9: 109931916-109931916 on Build GRCh38
rs537782915 Chr.9: 109932027-109932027 on Build GRCh38
rs781754956 Chr.9: 109931909-109931909 on Build GRCh38
rs778211592 Chr.9: 109932055-109932055 on Build GRCh38
rs1035892062 Chr.9: 109932043-109932043 on Build GRCh38
rs751837630 Chr.9: 109932041-109932041 on Build GRCh38
rs377498789 Chr.9: 109932022-109932022 on Build GRCh38
rs547916509 Chr.9: 109931923-109931923 on Build GRCh38
rs369606105 Chr.9: 109932061-109932061 on Build GRCh38
rs770201463 Chr.9: 109931917-109931917 on Build GRCh38
rs747131016 Chr.9: 109931897-109931897 on Build GRCh38
rs755214612 Chr.9: 109932042-109932042 on Build GRCh38
rs779729555 Chr.9: 109932073-109932073 on Build GRCh38
rs373349447 Chr.9: 109931994-109931994 on Build GRCh38
rs267602081 Chr.9: 109931971-109931971 on Build GRCh38
rs758653185 Chr.9: 109931881-109931881 on Build GRCh38
rs763979984 Chr.9: 109931959-109931959 on Build GRCh38
rs969266131 Chr.9: 109931910-109931910 on Build GRCh38
rs779180478 Chr.9: 109932066-109932066 on Build GRCh38
rs1020464534 Chr.9: 109931904-109931904 on Build GRCh38
rs554236832 Chr.9: 109932012-109932012 on Build GRCh38
rs370773077 Chr.9: 109932023-109932023 on Build GRCh38
rs61489557 Chr.9: 109932074-109932074 on Build GRCh38
rs901598702 Chr.9: 109931922-109931922 on Build GRCh38
rs369150990 Chr.9: 109931981-109931981 on Build GRCh38
rs150965931 Chr.9: 109931945-109931945 on Build GRCh38
rs750661992 Chr.9: 109931876-109931876 on Build GRCh38
rs944202107 Chr.9: 109931885-109931885 on Build GRCh38
rs763982997 Chr.9: 109932025-109932025 on Build GRCh38
rs745550402 Chr.9: 109932067-109932067 on Build GRCh38
rs774206740 Chr.9: 109931936-109931936 on Build GRCh38
rs775508946 Chr.9: 109931958-109931958 on Build GRCh38
rs753523699 Chr.9: 109931960-109931960 on Build GRCh38
rs773412761 Chr.9: 109931920-109931920 on Build GRCh38
rs759114610 Chr.9: 109931948-109931948 on Build GRCh38
rs752874662 Chr.9: 109931983-109931984 on Build GRCh38
rs996296757 Chr.9: 109931950-109931950 on Build GRCh38
rs781068844 Chr.9: 109931991-109931991 on Build GRCh38
rs755075393 Chr.9: 109931903-109931903 on Build GRCh38
rs201066314 Chr.9: 109932052-109932052 on Build GRCh38
rs771613841 Chr.9: 109932020-109932020 on Build GRCh38
rs920078663 Chr.9: 109932030-109932030 on Build GRCh38
rs757092195 Chr.9: 109931968-109931968 on Build GRCh38
rs575343856 Chr.9: 109931937-109931937 on Build GRCh38
rs767003696 Chr.9: 109931955-109931955 on Build GRCh38
rs765108654 Chr.9: 109932029-109932029 on Build GRCh38
rs139241824 Chr.9: 109931982-109931982 on Build GRCh38
rs771574411 Chr.9: 109932070-109932070 on Build GRCh38
rs745463626 Chr.9: 109932017-109932017 on Build GRCh38
rs778320722 Chr.9: 109932003-109932003 on Build GRCh38
rs749693840 Chr.9: 109932009-109932009 on Build GRCh38
rs374800756 Chr.9: 109931891-109931891 on Build GRCh38
rs566753922 Chr.9: 109932028-109932028 on Build GRCh38
rs991980629 Chr.9: 109932071-109932071 on Build GRCh38
rs542620758 Chr.9: 109931940-109931940 on Build GRCh38

View Full Product Details