Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54399539-54399812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA5
Entrez Gene ID
3678
Gene Name
integrin subunit alpha 5
Gene Aliases
CD49e, FNRA, VLA-5, VLA5A
UniGene
Hs.505654
Gene Chromosome Location
Chr.12: 54395261-54419266 on Build GRCh38
Transcripts
NM_002205.4 NR_120486.1 NR_120487.1

Gene Symbol
LOC102724050
Entrez Gene ID
102724050
Gene Name
uncharacterized LOC102724050
Gene Aliases
-
UniGene
Hs.670941
Gene Chromosome Location
Chr.12: 54353691-54467030 on Build GRCh38
Transcripts
NM_002205.4 NR_120486.1 NR_120487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
941131 c.2757G>A p.L919L Chr.12: 54399729-54399729 on Build GRCh38
5660792 c.2748T>C p.C916C Chr.12: 54399738-54399738 on Build GRCh38
348704 c.2829G>T p.K943N Chr.12: 54399657-54399657 on Build GRCh38
1255009 c.2759G>A p.R920H Chr.12: 54399727-54399727 on Build GRCh38
941130 c.2810A>G p.H937R Chr.12: 54399676-54399676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765745888 Chr.12: 54399782-54399782 on Build GRCh38
rs764112002 Chr.12: 54399741-54399741 on Build GRCh38
rs370583307 Chr.12: 54399621-54399621 on Build GRCh38
rs1056913750 Chr.12: 54399770-54399770 on Build GRCh38
rs922797052 Chr.12: 54399657-54399657 on Build GRCh38
rs1051843978 Chr.12: 54399711-54399711 on Build GRCh38
rs777252762 Chr.12: 54399636-54399636 on Build GRCh38
rs1023525712 Chr.12: 54399608-54399608 on Build GRCh38
rs200141012 Chr.12: 54399765-54399765 on Build GRCh38
rs776795701 Chr.12: 54399605-54399605 on Build GRCh38
rs779406495 Chr.12: 54399704-54399704 on Build GRCh38
rs762843772 Chr.12: 54399740-54399740 on Build GRCh38
rs755823731 Chr.12: 54399774-54399774 on Build GRCh38
rs767076112 Chr.12: 54399762-54399762 on Build GRCh38
rs756669983 Chr.12: 54399650-54399650 on Build GRCh38
rs759333262 Chr.12: 54399613-54399613 on Build GRCh38
rs758815022 Chr.12: 54399628-54399628 on Build GRCh38
rs753127929 Chr.12: 54399792-54399792 on Build GRCh38
rs748731524 Chr.12: 54399712-54399712 on Build GRCh38
rs201031426 Chr.12: 54399716-54399716 on Build GRCh38
rs772610449 Chr.12: 54399713-54399713 on Build GRCh38
rs142652896 Chr.12: 54399751-54399751 on Build GRCh38
rs878972350 Chr.12: 54399668-54399668 on Build GRCh38
rs750027552 Chr.12: 54399682-54399682 on Build GRCh38
rs764956878 Chr.12: 54399617-54399617 on Build GRCh38
rs759744172 Chr.12: 54399728-54399728 on Build GRCh38
rs756706920 Chr.12: 54399750-54399750 on Build GRCh38
rs375343882 Chr.12: 54399693-54399693 on Build GRCh38
rs960507525 Chr.12: 54399588-54399588 on Build GRCh38
rs780926237 Chr.12: 54399678-54399678 on Build GRCh38
rs138614417 Chr.12: 54399736-54399736 on Build GRCh38
rs776855334 Chr.12: 54399722-54399722 on Build GRCh38
rs371960269 Chr.12: 54399597-54399597 on Build GRCh38
rs142009562 Chr.12: 54399714-54399714 on Build GRCh38
rs771208809 Chr.12: 54399604-54399604 on Build GRCh38
rs752692825 Chr.12: 54399635-54399635 on Build GRCh38
rs781738005 Chr.12: 54399699-54399701 on Build GRCh38
rs201382501 Chr.12: 54399601-54399601 on Build GRCh38
rs373766369 Chr.12: 54399629-54399629 on Build GRCh38
rs899436783 Chr.12: 54399727-54399727 on Build GRCh38
rs116004551 Chr.12: 54399717-54399717 on Build GRCh38
rs751055025 Chr.12: 54399649-54399649 on Build GRCh38

View Full Product Details