Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36891460-36891715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF345
Entrez Gene ID
25850
Gene Name
zinc finger protein 345
Gene Aliases
HZF10
UniGene
Hs.362324
Gene Chromosome Location
Chr.19: 36850350-36893218 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

Gene Symbol
ZNF829
Entrez Gene ID
374899
Gene Name
zinc finger protein 829
Gene Aliases
-
UniGene
Hs.720312
Gene Chromosome Location
Chr.19: 36888124-36916387 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078875 c.1216A>C p.K406Q Chr.19: 36891575-36891575 on Build GRCh38
1480975 c.1115G>A p.R372K Chr.19: 36891676-36891676 on Build GRCh38
7426496 c.1126G>T p.D376Y Chr.19: 36891665-36891665 on Build GRCh38
6323690 c.1274G>A p.R425H Chr.19: 36891517-36891517 on Build GRCh38
7098570 c.1255A>C p.S419R Chr.19: 36891536-36891536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200748250 Chr.19: 36891602-36891602 on Build GRCh38
rs774705661 Chr.19: 36891514-36891514 on Build GRCh38
rs759428106 Chr.19: 36891646-36891646 on Build GRCh38
rs750930987 Chr.19: 36891503-36891503 on Build GRCh38
rs773600149 Chr.19: 36891517-36891517 on Build GRCh38
rs757970073 Chr.19: 36891541-36891541 on Build GRCh38
rs370190247 Chr.19: 36891616-36891616 on Build GRCh38
rs760930868 Chr.19: 36891520-36891520 on Build GRCh38
rs750851502 Chr.19: 36891636-36891644 on Build GRCh38
rs756169350 Chr.19: 36891501-36891519 on Build GRCh38
rs756833731 Chr.19: 36891580-36891580 on Build GRCh38
rs776706408 Chr.19: 36891638-36891638 on Build GRCh38
rs751222241 Chr.19: 36891561-36891561 on Build GRCh38
rs780815980 Chr.19: 36891581-36891581 on Build GRCh38
rs766291147 Chr.19: 36891529-36891529 on Build GRCh38
rs377686659 Chr.19: 36891653-36891653 on Build GRCh38
rs756758300 Chr.19: 36891667-36891667 on Build GRCh38
rs372663992 Chr.19: 36891601-36891601 on Build GRCh38
rs868863246 Chr.19: 36891642-36891642 on Build GRCh38
rs369348025 Chr.19: 36891592-36891592 on Build GRCh38
rs192798904 Chr.19: 36891494-36891494 on Build GRCh38
rs751130418 Chr.19: 36891655-36891655 on Build GRCh38
rs756736847 Chr.19: 36891518-36891518 on Build GRCh38
rs1048738550 Chr.19: 36891691-36891691 on Build GRCh38
rs764913013 Chr.19: 36891648-36891648 on Build GRCh38
rs767241898 Chr.19: 36891688-36891688 on Build GRCh38
rs966547273 Chr.19: 36891578-36891578 on Build GRCh38
rs771348288 Chr.19: 36891631-36891631 on Build GRCh38
rs990493512 Chr.19: 36891630-36891630 on Build GRCh38
rs866477046 Chr.19: 36891664-36891664 on Build GRCh38
rs748307111 Chr.19: 36891515-36891515 on Build GRCh38
rs752558777 Chr.19: 36891539-36891539 on Build GRCh38
rs779514046 Chr.19: 36891598-36891598 on Build GRCh38
rs749717750 Chr.19: 36891584-36891584 on Build GRCh38
rs79277718 Chr.19: 36891516-36891516 on Build GRCh38
rs767854488 Chr.19: 36891621-36891622 on Build GRCh38
rs776421835 Chr.19: 36891532-36891532 on Build GRCh38
rs763558897 Chr.19: 36891557-36891557 on Build GRCh38
rs578174327 Chr.19: 36891654-36891654 on Build GRCh38
rs374436128 Chr.19: 36891651-36891651 on Build GRCh38
rs376922609 Chr.19: 36891603-36891603 on Build GRCh38
rs556562482 Chr.19: 36891533-36891533 on Build GRCh38
rs762238694 Chr.19: 36891561-36891562 on Build GRCh38

View Full Product Details