Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151596356-151596627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC170
Entrez Gene ID
80129
Gene Name
coiled-coil domain containing 170
Gene Aliases
C6orf97, bA282P11.1
UniGene
Hs.660044
Gene Chromosome Location
Chr.6: 151494040-151621193 on Build GRCh38
Transcripts
NM_025059.3 XM_011536147.2 XM_011536148.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1074612 c.1542_1544delGGA p.E517delE Chr.6: 151596409-151596411 on Build GRCh38
740586 c.1701T>G p.N567K Chr.6: 151596568-151596568 on Build GRCh38
1074611 c.1519C>A p.L507I Chr.6: 151596386-151596386 on Build GRCh38
3019434 c.1693G>A p.D565N Chr.6: 151596560-151596560 on Build GRCh38
740587 c.1690G>A p.A564T Chr.6: 151596557-151596557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202238373 Chr.6: 151596519-151596519 on Build GRCh38
rs373104415 Chr.6: 151596511-151596511 on Build GRCh38
rs754298052 Chr.6: 151596577-151596577 on Build GRCh38
rs770432018 Chr.6: 151596533-151596533 on Build GRCh38
rs764088024 Chr.6: 151596488-151596488 on Build GRCh38
rs930429602 Chr.6: 151596449-151596449 on Build GRCh38
rs758475601 Chr.6: 151596405-151596405 on Build GRCh38
rs745463589 Chr.6: 151596539-151596539 on Build GRCh38
rs779873946 Chr.6: 151596559-151596559 on Build GRCh38
rs916926627 Chr.6: 151596585-151596585 on Build GRCh38
rs781444753 Chr.6: 151596427-151596427 on Build GRCh38
rs770534076 Chr.6: 151596458-151596458 on Build GRCh38
rs564830302 Chr.6: 151596413-151596413 on Build GRCh38
rs371982631 Chr.6: 151596404-151596404 on Build GRCh38
rs751644202 Chr.6: 151596415-151596415 on Build GRCh38
rs780510284 Chr.6: 151596520-151596520 on Build GRCh38
rs375012599 Chr.6: 151596567-151596567 on Build GRCh38
rs3757321 Chr.6: 151596460-151596460 on Build GRCh38
rs202125938 Chr.6: 151596552-151596552 on Build GRCh38
rs768524694 Chr.6: 151596557-151596557 on Build GRCh38
rs201569308 Chr.6: 151596562-151596562 on Build GRCh38
rs527492425 Chr.6: 151596395-151596395 on Build GRCh38
rs376587476 Chr.6: 151596428-151596428 on Build GRCh38
rs369246635 Chr.6: 151596432-151596432 on Build GRCh38
rs748907440 Chr.6: 151596576-151596576 on Build GRCh38
rs754244719 Chr.6: 151596522-151596522 on Build GRCh38
rs767640050 Chr.6: 151596561-151596561 on Build GRCh38
rs370430081 Chr.6: 151596524-151596524 on Build GRCh38
rs765178434 Chr.6: 151596396-151596396 on Build GRCh38
rs753178676 Chr.6: 151596599-151596599 on Build GRCh38
rs762934786 Chr.6: 151596471-151596471 on Build GRCh38
rs756729097 Chr.6: 151596602-151596602 on Build GRCh38
rs1005416670 Chr.6: 151596545-151596545 on Build GRCh38
rs746519473 Chr.6: 151596526-151596526 on Build GRCh38
rs756499565 Chr.6: 151596429-151596429 on Build GRCh38
rs563094004 Chr.6: 151596538-151596538 on Build GRCh38
rs954244273 Chr.6: 151596542-151596542 on Build GRCh38
rs753228753 Chr.6: 151596560-151596560 on Build GRCh38
rs183044234 Chr.6: 151596389-151596389 on Build GRCh38
rs769421068 Chr.6: 151596544-151596544 on Build GRCh38
rs749747525 Chr.6: 151596433-151596433 on Build GRCh38
rs772924463 Chr.6: 151596385-151596385 on Build GRCh38
rs369414988 Chr.6: 151596390-151596390 on Build GRCh38
rs200108871 Chr.6: 151596462-151596462 on Build GRCh38
rs531816992 Chr.6: 151596554-151596554 on Build GRCh38
rs371629858 Chr.6: 151596503-151596503 on Build GRCh38
rs1041091744 Chr.6: 151596465-151596465 on Build GRCh38
rs267600860 Chr.6: 151596410-151596410 on Build GRCh38
rs559878206 Chr.6: 151596394-151596394 on Build GRCh38
rs200776453 Chr.6: 151596434-151596434 on Build GRCh38
rs549058340 Chr.6: 151596400-151596400 on Build GRCh38
rs779315882 Chr.6: 151596578-151596578 on Build GRCh38
rs375623063 Chr.6: 151596457-151596457 on Build GRCh38
rs777209570 Chr.6: 151596438-151596438 on Build GRCh38
rs201487502 Chr.6: 151596459-151596459 on Build GRCh38
rs34430497 Chr.6: 151596525-151596525 on Build GRCh38
rs377387471 Chr.6: 151596512-151596512 on Build GRCh38
rs79790416 Chr.6: 151596571-151596571 on Build GRCh38
rs757486543 Chr.6: 151596426-151596426 on Build GRCh38
rs201625561 Chr.6: 151596461-151596461 on Build GRCh38

View Full Product Details