Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55719171-55719432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAGB
Entrez Gene ID
10325
Gene Name
Ras related GTP binding B
Gene Aliases
RAGB, bA465E19.1
UniGene
Hs.50282
Gene Chromosome Location
Chr.X: 55717677-55760209 on Build GRCh38
Transcripts
NM_006064.4 NM_016656.3 XM_005261973.4 XM_017029217.1 XM_017029218.1

SNP Detail

SNP ID Chromosome Location
rs377086149 Chr.X: 55719343-55719343 on Build GRCh38
rs779146249 Chr.X: 55719336-55719336 on Build GRCh38
rs766581962 Chr.X: 55719297-55719297 on Build GRCh38
rs889470568 Chr.X: 55719223-55719223 on Build GRCh38
rs755309297 Chr.X: 55719323-55719323 on Build GRCh38
rs778468257 Chr.X: 55719365-55719365 on Build GRCh38
rs754025741 Chr.X: 55719314-55719314 on Build GRCh38
rs745403613 Chr.X: 55719205-55719205 on Build GRCh38
rs769213337 Chr.X: 55719368-55719368 on Build GRCh38
rs775712318 Chr.X: 55719224-55719225 on Build GRCh38
rs752633873 Chr.X: 55719366-55719366 on Build GRCh38
rs369735980 Chr.X: 55719370-55719370 on Build GRCh38
rs146392102 Chr.X: 55719321-55719321 on Build GRCh38
rs368664349 Chr.X: 55719292-55719292 on Build GRCh38
rs747609850 Chr.X: 55719374-55719374 on Build GRCh38
rs767576738 Chr.X: 55719247-55719247 on Build GRCh38
rs1049972224 Chr.X: 55719221-55719221 on Build GRCh38
rs188485143 Chr.X: 55719235-55719235 on Build GRCh38
rs867944189 Chr.X: 55719316-55719316 on Build GRCh38
rs866465682 Chr.X: 55719397-55719397 on Build GRCh38
rs112651852 Chr.X: 55719360-55719360 on Build GRCh38
rs768422167 Chr.X: 55719390-55719390 on Build GRCh38
rs758954773 Chr.X: 55719355-55719355 on Build GRCh38

View Full Product Details