Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81922778-81923041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAFG
Entrez Gene ID
4097
Gene Name
MAF bZIP transcription factor G
Gene Aliases
hMAF
UniGene
Hs.252229
Gene Chromosome Location
Chr.17: 81918269-81927711 on Build GRCh38
Transcripts
NM_002359.3 NM_032711.3 XM_011523578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233771 c.315C>T p.D105D Chr.17: 81922779-81922779 on Build GRCh38
6705888 c.292G>A p.A98T Chr.17: 81922802-81922802 on Build GRCh38
2805734 c.206G>A p.R69H Chr.17: 81922888-81922888 on Build GRCh38
6362026 c.65C>T p.T22I Chr.17: 81923029-81923029 on Build GRCh38
2805733 c.207C>T p.R69R Chr.17: 81922887-81922887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987455359 Chr.17: 81922878-81922878 on Build GRCh38
rs781513645 Chr.17: 81922946-81922946 on Build GRCh38
rs374433638 Chr.17: 81922920-81922920 on Build GRCh38
rs766990632 Chr.17: 81922872-81922872 on Build GRCh38
rs1054499 Chr.17: 81922890-81922890 on Build GRCh38
rs376037603 Chr.17: 81923019-81923019 on Build GRCh38
rs757533469 Chr.17: 81922935-81922935 on Build GRCh38
rs139997763 Chr.17: 81922902-81922902 on Build GRCh38
rs1002776452 Chr.17: 81922978-81922978 on Build GRCh38
rs1053237930 Chr.17: 81922995-81922995 on Build GRCh38
rs377657966 Chr.17: 81922907-81922907 on Build GRCh38
rs374190880 Chr.17: 81922893-81922893 on Build GRCh38
rs112776865 Chr.17: 81922845-81922845 on Build GRCh38
rs770074111 Chr.17: 81922842-81922842 on Build GRCh38
rs770125662 Chr.17: 81922964-81922964 on Build GRCh38
rs769216706 Chr.17: 81923009-81923009 on Build GRCh38
rs747225135 Chr.17: 81922809-81922809 on Build GRCh38
rs575859230 Chr.17: 81922965-81922965 on Build GRCh38
rs149842973 Chr.17: 81922908-81922908 on Build GRCh38
rs766058581 Chr.17: 81922887-81922887 on Build GRCh38
rs903082314 Chr.17: 81922956-81922956 on Build GRCh38
rs139968514 Chr.17: 81922812-81922812 on Build GRCh38
rs772766552 Chr.17: 81922873-81922873 on Build GRCh38
rs144455047 Chr.17: 81923004-81923004 on Build GRCh38
rs746105372 Chr.17: 81922947-81922947 on Build GRCh38
rs368364514 Chr.17: 81922977-81922977 on Build GRCh38
rs148289755 Chr.17: 81922802-81922802 on Build GRCh38
rs760314644 Chr.17: 81922879-81922879 on Build GRCh38
rs772676495 Chr.17: 81923013-81923013 on Build GRCh38
rs368117522 Chr.17: 81922851-81922851 on Build GRCh38
rs371063454 Chr.17: 81922941-81922941 on Build GRCh38
rs144043335 Chr.17: 81922830-81922830 on Build GRCh38
rs751732962 Chr.17: 81922932-81922932 on Build GRCh38
rs777107870 Chr.17: 81922814-81922814 on Build GRCh38
rs143133030 Chr.17: 81922803-81922803 on Build GRCh38
rs776022053 Chr.17: 81922843-81922843 on Build GRCh38

View Full Product Details