Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33528851-33529124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18B
Entrez Gene ID
441459
Gene Name
ankyrin repeat domain 18B
Gene Aliases
bA255A11.3, bA255A11.5
UniGene
Hs.493710
Gene Chromosome Location
Chr.9: 33524403-33578755 on Build GRCh38
Transcripts
NM_001244752.1 XM_011517881.2 XM_011517883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122205 c.383A>G p.N128S Chr.9: 33529061-33529061 on Build GRCh38
6289231 c.433G>A p.E145K Chr.9: 33529111-33529111 on Build GRCh38
3699302 c.389A>C p.K130T Chr.9: 33529067-33529067 on Build GRCh38
6367437 c.325G>A p.V109I Chr.9: 33529003-33529003 on Build GRCh38
6331061 c.385A>C p.I129L Chr.9: 33529063-33529063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2890510 Chr.9: 33529033-33529033 on Build GRCh38
rs779564791 Chr.9: 33529025-33529025 on Build GRCh38
rs751342735 Chr.9: 33528987-33528987 on Build GRCh38
rs142689832 Chr.9: 33528888-33528888 on Build GRCh38
rs771110484 Chr.9: 33529037-33529037 on Build GRCh38
rs776137139 Chr.9: 33529030-33529032 on Build GRCh38
rs774179943 Chr.9: 33529073-33529073 on Build GRCh38
rs767105533 Chr.9: 33529076-33529076 on Build GRCh38
rs2380775 Chr.9: 33529009-33529009 on Build GRCh38
rs759018089 Chr.9: 33529081-33529081 on Build GRCh38
rs2890511 Chr.9: 33529045-33529045 on Build GRCh38
rs975276791 Chr.9: 33529023-33529023 on Build GRCh38
rs753389982 Chr.9: 33529079-33529079 on Build GRCh38
rs764679485 Chr.9: 33529090-33529090 on Build GRCh38
rs767174813 Chr.9: 33529004-33529004 on Build GRCh38
rs775420630 Chr.9: 33529056-33529056 on Build GRCh38
rs772851925 Chr.9: 33529077-33529077 on Build GRCh38
rs759394828 Chr.9: 33529045-33529049 on Build GRCh38
rs758956644 Chr.9: 33529028-33529028 on Build GRCh38
rs952370729 Chr.9: 33529055-33529055 on Build GRCh38
rs751894761 Chr.9: 33529092-33529092 on Build GRCh38
rs745968858 Chr.9: 33529046-33529046 on Build GRCh38
rs769847543 Chr.9: 33529050-33529050 on Build GRCh38
rs967274336 Chr.9: 33528904-33528904 on Build GRCh38
rs775365186 Chr.9: 33528956-33528956 on Build GRCh38
rs759636580 Chr.9: 33528876-33528876 on Build GRCh38
rs749281512 Chr.9: 33529062-33529062 on Build GRCh38
rs766040903 Chr.9: 33529078-33529078 on Build GRCh38
rs753194150 Chr.9: 33529027-33529027 on Build GRCh38
rs574293426 Chr.9: 33529101-33529101 on Build GRCh38
rs2380776 Chr.9: 33529042-33529042 on Build GRCh38
rs761619001 Chr.9: 33529074-33529074 on Build GRCh38
rs761412790 Chr.9: 33528996-33528996 on Build GRCh38
rs757604151 Chr.9: 33529094-33529094 on Build GRCh38
rs371977538 Chr.9: 33529063-33529063 on Build GRCh38
rs4008879 Chr.9: 33529032-33529032 on Build GRCh38
rs762742300 Chr.9: 33528966-33528966 on Build GRCh38
rs1019409848 Chr.9: 33528997-33528997 on Build GRCh38
rs778345291 Chr.9: 33529030-33529030 on Build GRCh38
rs755608154 Chr.9: 33529021-33529021 on Build GRCh38
rs763819806 Chr.9: 33528984-33528984 on Build GRCh38

View Full Product Details