Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 124747043-124747276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR37
Entrez Gene ID
2861
Gene Name
G protein-coupled receptor 37
Gene Aliases
EDNRBL, PAELR, hET(B)R-LP
UniGene
Hs.406094
Gene Chromosome Location
Chr.7: 124745601-124766025 on Build GRCh38
Transcripts
NM_005302.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5037832 c.1324A>T p.R442* Chr.7: 124747043-124747043 on Build GRCh38
1253558 c.1166G>T p.G389V Chr.7: 124747201-124747201 on Build GRCh38
1253557 c.1303G>T p.A435S Chr.7: 124747064-124747064 on Build GRCh38
4523443 c.1215G>A p.K405K Chr.7: 124747152-124747152 on Build GRCh38
5884473 c.1233T>A p.S411R Chr.7: 124747134-124747134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779855100 Chr.7: 124747149-124747149 on Build GRCh38
rs768728044 Chr.7: 124747176-124747176 on Build GRCh38
rs755831636 Chr.7: 124747146-124747146 on Build GRCh38
rs777144844 Chr.7: 124747229-124747229 on Build GRCh38
rs763636472 Chr.7: 124747123-124747123 on Build GRCh38
rs767092858 Chr.7: 124747248-124747248 on Build GRCh38
rs149358350 Chr.7: 124747088-124747088 on Build GRCh38
rs897565558 Chr.7: 124747063-124747063 on Build GRCh38
rs780225867 Chr.7: 124747076-124747076 on Build GRCh38
rs531412697 Chr.7: 124747129-124747129 on Build GRCh38
rs778123567 Chr.7: 124747194-124747194 on Build GRCh38
rs369674783 Chr.7: 124747165-124747165 on Build GRCh38
rs749552573 Chr.7: 124747083-124747083 on Build GRCh38
rs775256152 Chr.7: 124747122-124747122 on Build GRCh38
rs761204777 Chr.7: 124747247-124747247 on Build GRCh38
rs768681144 Chr.7: 124747085-124747085 on Build GRCh38
rs373099722 Chr.7: 124747171-124747171 on Build GRCh38
rs747595694 Chr.7: 124747220-124747220 on Build GRCh38
rs888707405 Chr.7: 124747114-124747114 on Build GRCh38
rs771473340 Chr.7: 124747121-124747121 on Build GRCh38
rs750422451 Chr.7: 124747135-124747135 on Build GRCh38
rs774107557 Chr.7: 124747240-124747240 on Build GRCh38
rs767340618 Chr.7: 124747130-124747130 on Build GRCh38
rs201188374 Chr.7: 124747096-124747096 on Build GRCh38
rs771385378 Chr.7: 124747226-124747226 on Build GRCh38
rs1011743228 Chr.7: 124747072-124747072 on Build GRCh38
rs755207415 Chr.7: 124747158-124747158 on Build GRCh38
rs755994683 Chr.7: 124747065-124747065 on Build GRCh38
rs768417202 Chr.7: 124747239-124747239 on Build GRCh38
rs148194574 Chr.7: 124747086-124747086 on Build GRCh38
rs754010081 Chr.7: 124747156-124747156 on Build GRCh38
rs1048515888 Chr.7: 124747113-124747113 on Build GRCh38
rs776871185 Chr.7: 124747153-124747154 on Build GRCh38
rs144128090 Chr.7: 124747106-124747106 on Build GRCh38
rs554445623 Chr.7: 124747232-124747232 on Build GRCh38
rs141258559 Chr.7: 124747168-124747168 on Build GRCh38
rs760938124 Chr.7: 124747119-124747119 on Build GRCh38

View Full Product Details