Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 77191176-77191413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC14L1
Entrez Gene ID
6397
Gene Name
SEC14 like lipid binding 1
Gene Aliases
PRELID4A, SEC14L
UniGene
Hs.464184
Gene Chromosome Location
Chr.17: 77088643-77217101 on Build GRCh38
Transcripts
NM_001039573.2 NM_001143998.1 NM_001143999.1 NM_001144001.1 NM_001204408.1 NM_001204410.1 NM_003003.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1386288 c.257T>C p.L86P Chr.17: 77191224-77191224 on Build GRCh38
6773972 c.266G>A p.R89Q Chr.17: 77191233-77191233 on Build GRCh38
6773971 c.266G>A p.R89Q Chr.17: 77191233-77191233 on Build GRCh38
6081957 c.276T>C p.T92T Chr.17: 77191243-77191243 on Build GRCh38
6081956 c.276T>C p.T92T Chr.17: 77191243-77191243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052176962 Chr.17: 77191366-77191366 on Build GRCh38
rs755382423 Chr.17: 77191270-77191270 on Build GRCh38
rs79265302 Chr.17: 77191355-77191355 on Build GRCh38
rs764634974 Chr.17: 77191345-77191345 on Build GRCh38
rs960317593 Chr.17: 77191223-77191223 on Build GRCh38
rs768660926 Chr.17: 77191329-77191329 on Build GRCh38
rs756653156 Chr.17: 77191344-77191344 on Build GRCh38
rs753191916 Chr.17: 77191280-77191280 on Build GRCh38
rs73378315 Chr.17: 77191321-77191321 on Build GRCh38
rs757317007 Chr.17: 77191308-77191308 on Build GRCh38
rs200035066 Chr.17: 77191289-77191289 on Build GRCh38
rs746888985 Chr.17: 77191326-77191326 on Build GRCh38
rs73998646 Chr.17: 77191331-77191331 on Build GRCh38
rs1047050269 Chr.17: 77191353-77191353 on Build GRCh38
rs578095610 Chr.17: 77191224-77191224 on Build GRCh38
rs201830390 Chr.17: 77191334-77191334 on Build GRCh38
rs1049422 Chr.17: 77191257-77191257 on Build GRCh38
rs764712443 Chr.17: 77191332-77191332 on Build GRCh38
rs777669180 Chr.17: 77191293-77191293 on Build GRCh38
rs757343697 Chr.17: 77191354-77191354 on Build GRCh38
rs763526107 Chr.17: 77191233-77191233 on Build GRCh38
rs1049423 Chr.17: 77191259-77191259 on Build GRCh38
rs750093696 Chr.17: 77191260-77191260 on Build GRCh38
rs746144718 Chr.17: 77191320-77191320 on Build GRCh38
rs774591837 Chr.17: 77191328-77191328 on Build GRCh38
rs929788882 Chr.17: 77191340-77191340 on Build GRCh38
rs760669863 Chr.17: 77191230-77191230 on Build GRCh38
rs749420346 Chr.17: 77191305-77191305 on Build GRCh38
rs768061912 Chr.17: 77191278-77191278 on Build GRCh38
rs775267222 Chr.17: 77191324-77191324 on Build GRCh38
rs531791468 Chr.17: 77191343-77191343 on Build GRCh38
rs376407245 Chr.17: 77191246-77191246 on Build GRCh38
rs879201590 Chr.17: 77191244-77191244 on Build GRCh38
rs201215904 Chr.17: 77191330-77191330 on Build GRCh38
rs754348377 Chr.17: 77191348-77191348 on Build GRCh38
rs200241315 Chr.17: 77191333-77191333 on Build GRCh38
rs575025415 Chr.17: 77191232-77191232 on Build GRCh38
rs764972268 Chr.17: 77191255-77191255 on Build GRCh38
rs768881794 Chr.17: 77191312-77191312 on Build GRCh38

View Full Product Details