Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100374273-100374498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PILRA
Entrez Gene ID
29992
Gene Name
paired immunoglobin like type 2 receptor alpha
Gene Aliases
FDF03
UniGene
Hs.444407
Gene Chromosome Location
Chr.7: 100373445-100400099 on Build GRCh38
Transcripts
NM_013439.2 NM_178272.1 NM_178273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4414550 c.435C>T p.T145T Chr.7: 100374414-100374414 on Build GRCh38
4488823 c.338C>T p.S113F Chr.7: 100374317-100374317 on Build GRCh38
5606911 c.348G>A p.Q116Q Chr.7: 100374327-100374327 on Build GRCh38
1094056 c.410A>G p.Q137R Chr.7: 100374389-100374389 on Build GRCh38
3643070 c.423C>T p.S141S Chr.7: 100374402-100374402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762150606 Chr.7: 100374361-100374361 on Build GRCh38
rs779293942 Chr.7: 100374406-100374406 on Build GRCh38
rs545578307 Chr.7: 100374421-100374421 on Build GRCh38
rs751467803 Chr.7: 100374315-100374315 on Build GRCh38
rs578108899 Chr.7: 100374458-100374458 on Build GRCh38
rs373188096 Chr.7: 100374432-100374432 on Build GRCh38
rs749991894 Chr.7: 100374309-100374309 on Build GRCh38
rs555728760 Chr.7: 100374300-100374300 on Build GRCh38
rs760958715 Chr.7: 100374293-100374293 on Build GRCh38
rs138900405 Chr.7: 100374295-100374295 on Build GRCh38
rs746051766 Chr.7: 100374408-100374408 on Build GRCh38
rs1025566838 Chr.7: 100374319-100374319 on Build GRCh38
rs761489628 Chr.7: 100374301-100374301 on Build GRCh38
rs754761561 Chr.7: 100374318-100374318 on Build GRCh38
rs752609650 Chr.7: 100374397-100374397 on Build GRCh38
rs777628186 Chr.7: 100374380-100374381 on Build GRCh38
rs150831633 Chr.7: 100374393-100374393 on Build GRCh38
rs769116330 Chr.7: 100374360-100374360 on Build GRCh38
rs556203759 Chr.7: 100374377-100374377 on Build GRCh38
rs747938446 Chr.7: 100374329-100374329 on Build GRCh38
rs865966743 Chr.7: 100374302-100374302 on Build GRCh38
rs141936238 Chr.7: 100374355-100374355 on Build GRCh38
rs200088454 Chr.7: 100374412-100374412 on Build GRCh38
rs764848285 Chr.7: 100374306-100374306 on Build GRCh38
rs747561669 Chr.7: 100374466-100374466 on Build GRCh38
rs755014519 Chr.7: 100374378-100374378 on Build GRCh38
rs116478382 Chr.7: 100374374-100374374 on Build GRCh38
rs765873300 Chr.7: 100374375-100374375 on Build GRCh38
rs779858617 Chr.7: 100374313-100374313 on Build GRCh38
rs553057992 Chr.7: 100374356-100374356 on Build GRCh38
rs567501556 Chr.7: 100374372-100374372 on Build GRCh38
rs770085510 Chr.7: 100374470-100374470 on Build GRCh38
rs376921518 Chr.7: 100374469-100374469 on Build GRCh38
rs146248193 Chr.7: 100374473-100374473 on Build GRCh38
rs200150930 Chr.7: 100374457-100374457 on Build GRCh38
rs745941741 Chr.7: 100374345-100374345 on Build GRCh38
rs747034333 Chr.7: 100374423-100374423 on Build GRCh38
rs139606576 Chr.7: 100374405-100374405 on Build GRCh38
rs764934471 Chr.7: 100374371-100374371 on Build GRCh38
rs781172979 Chr.7: 100374326-100374326 on Build GRCh38
rs779275754 Chr.7: 100374343-100374343 on Build GRCh38

View Full Product Details