Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36098260-36098521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD3
Entrez Gene ID
143458
Gene Name
low density lipoprotein receptor class A domain containing 3
Gene Aliases
LRAD3
UniGene
Hs.745134
Gene Chromosome Location
Chr.11: 35943981-36232136 on Build GRCh38
Transcripts
NM_001304263.1 NM_001304264.1 NM_174902.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687285 c.346G>T p.A116S Chr.11: 36098353-36098353 on Build GRCh38
394391 c.371T>A p.L124H Chr.11: 36098378-36098378 on Build GRCh38
4032595 c.366C>T p.N122N Chr.11: 36098373-36098373 on Build GRCh38
1675944 c.418G>A p.D140N Chr.11: 36098425-36098425 on Build GRCh38
7374894 c.436A>T p.S146C Chr.11: 36098443-36098443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563065338 Chr.11: 36098353-36098353 on Build GRCh38
rs781129718 Chr.11: 36098469-36098469 on Build GRCh38
rs1034287751 Chr.11: 36098296-36098296 on Build GRCh38
rs147309219 Chr.11: 36098389-36098389 on Build GRCh38
rs888037198 Chr.11: 36098283-36098283 on Build GRCh38
rs762189885 Chr.11: 36098435-36098435 on Build GRCh38
rs779093434 Chr.11: 36098391-36098391 on Build GRCh38
rs775910416 Chr.11: 36098407-36098407 on Build GRCh38
rs775865096 Chr.11: 36098348-36098348 on Build GRCh38
rs781042761 Chr.11: 36098374-36098374 on Build GRCh38
rs200985232 Chr.11: 36098404-36098404 on Build GRCh38
rs543035243 Chr.11: 36098352-36098352 on Build GRCh38
rs551961774 Chr.11: 36098431-36098431 on Build GRCh38
rs751357485 Chr.11: 36098367-36098367 on Build GRCh38
rs758015390 Chr.11: 36098357-36098357 on Build GRCh38
rs775963709 Chr.11: 36098319-36098319 on Build GRCh38
rs939847695 Chr.11: 36098495-36098495 on Build GRCh38
rs7122283 Chr.11: 36098473-36098473 on Build GRCh38
rs143972981 Chr.11: 36098373-36098373 on Build GRCh38
rs373533104 Chr.11: 36098493-36098493 on Build GRCh38
rs992772256 Chr.11: 36098422-36098422 on Build GRCh38
rs766017562 Chr.11: 36098470-36098470 on Build GRCh38
rs754880327 Chr.11: 36098442-36098442 on Build GRCh38
rs528265275 Chr.11: 36098497-36098497 on Build GRCh38
rs372681106 Chr.11: 36098292-36098292 on Build GRCh38
rs1048442312 Chr.11: 36098461-36098461 on Build GRCh38
rs370620850 Chr.11: 36098427-36098427 on Build GRCh38
rs770391322 Chr.11: 36098297-36098337 on Build GRCh38
rs560929485 Chr.11: 36098498-36098498 on Build GRCh38
rs762510042 Chr.11: 36098460-36098460 on Build GRCh38
rs574403233 Chr.11: 36098328-36098328 on Build GRCh38
rs376946994 Chr.11: 36098300-36098300 on Build GRCh38
rs374622108 Chr.11: 36098350-36098350 on Build GRCh38
rs770326887 Chr.11: 36098440-36098440 on Build GRCh38
rs751104676 Chr.11: 36098490-36098490 on Build GRCh38
rs144877934 Chr.11: 36098452-36098452 on Build GRCh38
rs141018089 Chr.11: 36098406-36098406 on Build GRCh38
rs966329991 Chr.11: 36098388-36098388 on Build GRCh38
rs756142163 Chr.11: 36098390-36098390 on Build GRCh38
rs370552901 Chr.11: 36098310-36098310 on Build GRCh38
rs754922813 Chr.11: 36098281-36098281 on Build GRCh38
rs747262995 Chr.11: 36098414-36098414 on Build GRCh38
rs749967264 Chr.11: 36098356-36098356 on Build GRCh38
rs774585643 Chr.11: 36098301-36098301 on Build GRCh38
rs764661438 Chr.11: 36098329-36098329 on Build GRCh38

View Full Product Details