Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 14218759-14218994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MKL2
Entrez Gene ID
57496
Gene Name
MKL1/myocardin like 2
Gene Aliases
MRTF-B, MRTFB, NPD001
UniGene
Hs.49143
Gene Chromosome Location
Chr.16: 14071076-14266779 on Build GRCh38
Transcripts
NM_001308142.1 NM_014048.4 XM_005255452.3 XM_005255453.4 XM_005255455.3 XM_006720908.3 XM_006720909.3 XM_006720912.3 XM_006720913.2 XM_006720914.2 XM_011522568.2 XM_011522569.2 XM_017023501.1 XM_017023502.1 XM_017023503.1 XM_017023504.1 XM_017023505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608913 c.530A>G p.D177G Chr.16: 14218835-14218835 on Build GRCh38
5752985 c.611delA p.E204fs*21 Chr.16: 14218916-14218916 on Build GRCh38
6031881 c.503C>T p.P168L Chr.16: 14218841-14218841 on Build GRCh38
6713106 c.550G>A p.D184N Chr.16: 14218855-14218855 on Build GRCh38
5951960 c.552G>T p.L184F Chr.16: 14218890-14218890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763599709 Chr.16: 14218836-14218836 on Build GRCh38
rs759911105 Chr.16: 14218933-14218933 on Build GRCh38
rs773767840 Chr.16: 14218914-14218914 on Build GRCh38
rs766627475 Chr.16: 14218789-14218793 on Build GRCh38
rs1051953224 Chr.16: 14218847-14218847 on Build GRCh38
rs772100405 Chr.16: 14218917-14218925 on Build GRCh38
rs775526616 Chr.16: 14218936-14218936 on Build GRCh38
rs138917129 Chr.16: 14218930-14218930 on Build GRCh38
rs147706639 Chr.16: 14218932-14218932 on Build GRCh38
rs780048102 Chr.16: 14218885-14218885 on Build GRCh38
rs758337180 Chr.16: 14218884-14218884 on Build GRCh38
rs760946083 Chr.16: 14218939-14218939 on Build GRCh38
rs1040957196 Chr.16: 14218962-14218962 on Build GRCh38
rs919695122 Chr.16: 14218903-14218903 on Build GRCh38
rs747839637 Chr.16: 14218907-14218907 on Build GRCh38
rs372008307 Chr.16: 14218897-14218897 on Build GRCh38
rs367882813 Chr.16: 14218854-14218854 on Build GRCh38
rs116509814 Chr.16: 14218964-14218964 on Build GRCh38
rs759668731 Chr.16: 14218846-14218846 on Build GRCh38
rs766043027 Chr.16: 14218968-14218968 on Build GRCh38
rs752938315 Chr.16: 14218855-14218855 on Build GRCh38
rs550904085 Chr.16: 14218851-14218851 on Build GRCh38
rs959603063 Chr.16: 14218826-14218826 on Build GRCh38
rs775630661 Chr.16: 14218922-14218923 on Build GRCh38
rs374057367 Chr.16: 14218840-14218840 on Build GRCh38
rs751549938 Chr.16: 14218971-14218971 on Build GRCh38
rs774359364 Chr.16: 14218838-14218838 on Build GRCh38
rs767667006 Chr.16: 14218935-14218935 on Build GRCh38
rs749663624 Chr.16: 14218923-14218923 on Build GRCh38
rs993593904 Chr.16: 14218965-14218965 on Build GRCh38
rs750206940 Chr.16: 14218875-14218875 on Build GRCh38
rs371242494 Chr.16: 14218837-14218837 on Build GRCh38
rs1021048612 Chr.16: 14218787-14218787 on Build GRCh38
rs985141721 Chr.16: 14218925-14218925 on Build GRCh38
rs374507083 Chr.16: 14218784-14218784 on Build GRCh38
rs780812900 Chr.16: 14218902-14218902 on Build GRCh38
rs764296867 Chr.16: 14218958-14218958 on Build GRCh38
rs769545298 Chr.16: 14218911-14218911 on Build GRCh38
rs143161598 Chr.16: 14218896-14218896 on Build GRCh38
rs747758219 Chr.16: 14218817-14218817 on Build GRCh38
rs933681631 Chr.16: 14218833-14218833 on Build GRCh38
rs746526481 Chr.16: 14218796-14218796 on Build GRCh38
rs367798067 Chr.16: 14218805-14218805 on Build GRCh38
rs756298765 Chr.16: 14218858-14218858 on Build GRCh38
rs115834470 Chr.16: 14218822-14218822 on Build GRCh38
rs773677919 Chr.16: 14218827-14218827 on Build GRCh38
rs776362770 Chr.16: 14218810-14218810 on Build GRCh38
rs745749315 Chr.16: 14218804-14218805 on Build GRCh38
rs753994004 Chr.16: 14218959-14218959 on Build GRCh38

View Full Product Details