Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45878430-45878693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM3
Entrez Gene ID
140831
Gene Name
zinc finger SWIM-type containing 3
Gene Aliases
C20orf164, PPP1R174
UniGene
Hs.292135
Gene Chromosome Location
Chr.20: 45857581-45879131 on Build GRCh38
Transcripts
NM_080752.3 NR_037628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681718 c.1919G>C p.R640P Chr.20: 45878495-45878495 on Build GRCh38
5718112 c.2065C>T p.P689S Chr.20: 45878623-45878623 on Build GRCh38
1412094 c.2010C>T p.D670D Chr.20: 45878568-45878568 on Build GRCh38
3293914 c.2033G>T p.W678L Chr.20: 45878591-45878591 on Build GRCh38
5797232 c.2066A>G p.H689R Chr.20: 45878642-45878642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148099263 Chr.20: 45878556-45878556 on Build GRCh38
rs775442140 Chr.20: 45878532-45878532 on Build GRCh38
rs961557879 Chr.20: 45878508-45878508 on Build GRCh38
rs751159706 Chr.20: 45878495-45878495 on Build GRCh38
rs747605406 Chr.20: 45878511-45878511 on Build GRCh38
rs141779185 Chr.20: 45878575-45878575 on Build GRCh38
rs753812522 Chr.20: 45878608-45878608 on Build GRCh38
rs757046850 Chr.20: 45878553-45878555 on Build GRCh38
rs754235021 Chr.20: 45878490-45878490 on Build GRCh38
rs757139461 Chr.20: 45878614-45878614 on Build GRCh38
rs369388471 Chr.20: 45878617-45878617 on Build GRCh38
rs768713497 Chr.20: 45878656-45878656 on Build GRCh38
rs1038020173 Chr.20: 45878509-45878509 on Build GRCh38
rs774688845 Chr.20: 45878526-45878526 on Build GRCh38
rs116532261 Chr.20: 45878505-45878505 on Build GRCh38
rs145639872 Chr.20: 45878531-45878531 on Build GRCh38
rs771307649 Chr.20: 45878456-45878456 on Build GRCh38
rs372827829 Chr.20: 45878635-45878635 on Build GRCh38
rs761172793 Chr.20: 45878479-45878479 on Build GRCh38
rs770609478 Chr.20: 45878520-45878520 on Build GRCh38
rs141913268 Chr.20: 45878491-45878491 on Build GRCh38
rs750733599 Chr.20: 45878569-45878569 on Build GRCh38
rs766108911 Chr.20: 45878494-45878494 on Build GRCh38
rs529895998 Chr.20: 45878517-45878517 on Build GRCh38
rs781451177 Chr.20: 45878625-45878625 on Build GRCh38
rs911326872 Chr.20: 45878545-45878545 on Build GRCh38
rs375272202 Chr.20: 45878559-45878559 on Build GRCh38
rs759710196 Chr.20: 45878458-45878458 on Build GRCh38
rs777811141 Chr.20: 45878516-45878516 on Build GRCh38
rs777906961 Chr.20: 45878601-45878601 on Build GRCh38
rs17446511 Chr.20: 45878668-45878668 on Build GRCh38
rs749029309 Chr.20: 45878662-45878662 on Build GRCh38
rs750376917 Chr.20: 45878608-45878609 on Build GRCh38
rs201805977 Chr.20: 45878622-45878622 on Build GRCh38
rs747145684 Chr.20: 45878655-45878655 on Build GRCh38
rs139396693 Chr.20: 45878473-45878473 on Build GRCh38
rs756141994 Chr.20: 45878512-45878512 on Build GRCh38
rs749042009 Chr.20: 45878454-45878454 on Build GRCh38
rs755652199 Chr.20: 45878597-45878597 on Build GRCh38
rs936114296 Chr.20: 45878561-45878561 on Build GRCh38
rs1054538733 Chr.20: 45878564-45878564 on Build GRCh38
rs772203694 Chr.20: 45878640-45878640 on Build GRCh38
rs537547455 Chr.20: 45878568-45878568 on Build GRCh38
rs780221869 Chr.20: 45878654-45878654 on Build GRCh38
rs201321759 Chr.20: 45878576-45878576 on Build GRCh38
rs777104903 Chr.20: 45878548-45878548 on Build GRCh38
rs368149535 Chr.20: 45878469-45878469 on Build GRCh38
rs967978052 Chr.20: 45878624-45878624 on Build GRCh38
rs548540451 Chr.20: 45878541-45878541 on Build GRCh38
rs764523680 Chr.20: 45878487-45878487 on Build GRCh38
rs774404420 Chr.20: 45878457-45878457 on Build GRCh38
rs745616813 Chr.20: 45878629-45878629 on Build GRCh38
rs780574081 Chr.20: 45878507-45878507 on Build GRCh38
rs754335703 Chr.20: 45878618-45878618 on Build GRCh38
rs764627937 Chr.20: 45878542-45878542 on Build GRCh38
rs752286337 Chr.20: 45878581-45878581 on Build GRCh38
rs767424682 Chr.20: 45878600-45878602 on Build GRCh38

View Full Product Details