Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20157078-20157329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBX1
Entrez Gene ID
120237
Gene Name
developing brain homeobox 1
Gene Aliases
-
UniGene
Hs.558604
Gene Chromosome Location
Chr.11: 20156214-20160324 on Build GRCh38
Transcripts
NM_001029865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7424054 c.505T>C p.S169P Chr.11: 20157204-20157204 on Build GRCh38
1297921 c.528G>A p.G176G Chr.11: 20157181-20157181 on Build GRCh38
4645044 c.525C>G p.R175R Chr.11: 20157184-20157184 on Build GRCh38
1203034 c.625C>T p.P209S Chr.11: 20157084-20157084 on Build GRCh38
4550295 c.497G>A p.G166E Chr.11: 20157212-20157212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896824992 Chr.11: 20157106-20157106 on Build GRCh38
rs896883380 Chr.11: 20157300-20157300 on Build GRCh38
rs748158152 Chr.11: 20157142-20157142 on Build GRCh38
rs780517304 Chr.11: 20157113-20157113 on Build GRCh38
rs957514977 Chr.11: 20157258-20157258 on Build GRCh38
rs563896524 Chr.11: 20157281-20157281 on Build GRCh38
rs780459348 Chr.11: 20157239-20157239 on Build GRCh38
rs781386086 Chr.11: 20157266-20157266 on Build GRCh38
rs759565509 Chr.11: 20157167-20157167 on Build GRCh38
rs763073718 Chr.11: 20157169-20157169 on Build GRCh38
rs898271793 Chr.11: 20157148-20157148 on Build GRCh38
rs578042640 Chr.11: 20157249-20157249 on Build GRCh38
rs144313400 Chr.11: 20157143-20157143 on Build GRCh38
rs768885730 Chr.11: 20157138-20157138 on Build GRCh38
rs774257702 Chr.11: 20157155-20157155 on Build GRCh38
rs760406115 Chr.11: 20157174-20157174 on Build GRCh38
rs200639137 Chr.11: 20157198-20157198 on Build GRCh38
rs372163354 Chr.11: 20157278-20157278 on Build GRCh38
rs769926331 Chr.11: 20157147-20157147 on Build GRCh38
rs1007096830 Chr.11: 20157156-20157156 on Build GRCh38
rs201030070 Chr.11: 20157118-20157118 on Build GRCh38
rs905054161 Chr.11: 20157211-20157211 on Build GRCh38
rs564645477 Chr.11: 20157214-20157214 on Build GRCh38
rs758984330 Chr.11: 20157235-20157235 on Build GRCh38
rs776861724 Chr.11: 20157139-20157139 on Build GRCh38
rs749353648 Chr.11: 20157289-20157289 on Build GRCh38
rs1027671120 Chr.11: 20157120-20157120 on Build GRCh38
rs750873262 Chr.11: 20157233-20157233 on Build GRCh38
rs914531810 Chr.11: 20157263-20157263 on Build GRCh38
rs831464 Chr.11: 20157171-20157171 on Build GRCh38
rs1037215206 Chr.11: 20157284-20157284 on Build GRCh38
rs773315991 Chr.11: 20157282-20157282 on Build GRCh38
rs143300593 Chr.11: 20157181-20157181 on Build GRCh38
rs756824999 Chr.11: 20157208-20157208 on Build GRCh38

View Full Product Details