Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64202993-64203264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G1
Entrez Gene ID
53944
Gene Name
casein kinase 1 gamma 1
Gene Aliases
CK1gamma1
UniGene
Hs.646508
Gene Chromosome Location
Chr.15: 64165517-64356243 on Build GRCh38
Transcripts
NM_022048.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7349487 c.1068T>G p.H356Q Chr.15: 64203121-64203121 on Build GRCh38
700725 c.1052G>C p.R351P Chr.15: 64203137-64203137 on Build GRCh38
963919 c.1049C>A p.T350N Chr.15: 64203140-64203140 on Build GRCh38
1374026 c.1100G>A p.R367Q Chr.15: 64203089-64203089 on Build GRCh38
6077533 c.1048A>G p.T350A Chr.15: 64203141-64203141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778614961 Chr.15: 64203216-64203216 on Build GRCh38
rs1030155813 Chr.15: 64203021-64203021 on Build GRCh38
rs371686682 Chr.15: 64203233-64203233 on Build GRCh38
rs1032693955 Chr.15: 64203094-64203094 on Build GRCh38
rs560750523 Chr.15: 64203086-64203086 on Build GRCh38
rs772862682 Chr.15: 64203033-64203033 on Build GRCh38
rs949147177 Chr.15: 64203181-64203181 on Build GRCh38
rs748889847 Chr.15: 64203035-64203035 on Build GRCh38
rs922737799 Chr.15: 64203015-64203015 on Build GRCh38
rs369164703 Chr.15: 64203175-64203175 on Build GRCh38
rs372653620 Chr.15: 64203144-64203144 on Build GRCh38
rs769102862 Chr.15: 64203057-64203057 on Build GRCh38
rs528335045 Chr.15: 64203166-64203166 on Build GRCh38
rs777194377 Chr.15: 64203068-64203068 on Build GRCh38
rs772458389 Chr.15: 64203042-64203042 on Build GRCh38
rs867657659 Chr.15: 64203183-64203183 on Build GRCh38
rs376835504 Chr.15: 64203165-64203165 on Build GRCh38
rs777225635 Chr.15: 64203192-64203192 on Build GRCh38
rs752620688 Chr.15: 64203180-64203180 on Build GRCh38
rs375517581 Chr.15: 64203032-64203032 on Build GRCh38
rs761079289 Chr.15: 64203055-64203055 on Build GRCh38
rs367693773 Chr.15: 64203199-64203199 on Build GRCh38
rs747402454 Chr.15: 64203227-64203227 on Build GRCh38
rs780995284 Chr.15: 64203167-64203167 on Build GRCh38
rs936576896 Chr.15: 64203093-64203093 on Build GRCh38
rs373960684 Chr.15: 64203237-64203237 on Build GRCh38
rs879242288 Chr.15: 64203029-64203029 on Build GRCh38
rs1050315057 Chr.15: 64203046-64203046 on Build GRCh38
rs759321674 Chr.15: 64203205-64203205 on Build GRCh38
rs910377376 Chr.15: 64203077-64203077 on Build GRCh38
rs775874536 Chr.15: 64203051-64203051 on Build GRCh38
rs755642308 Chr.15: 64203188-64203188 on Build GRCh38
rs957700531 Chr.15: 64203113-64203113 on Build GRCh38
rs765353731 Chr.15: 64203114-64203114 on Build GRCh38
rs761807608 Chr.15: 64203103-64203103 on Build GRCh38
rs750702027 Chr.15: 64203120-64203120 on Build GRCh38
rs766682369 Chr.15: 64203155-64203155 on Build GRCh38

View Full Product Details