Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77114120-77114385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEAK1
Entrez Gene ID
79834
Gene Name
pseudopodium enriched atypical kinase 1
Gene Aliases
SGK269
UniGene
Hs.731402
Gene Chromosome Location
Chr.15: 77100829-77420104 on Build GRCh38
Transcripts
NM_024776.3 XM_005254670.2 XM_005254671.2 XM_005254673.2 XM_005254674.2 XM_005254675.4 XM_011522033.2 XM_011522034.2 XM_011522035.2 XM_011522036.2 XM_011522037.2 XM_011522038.2 XM_011522039.2 XM_011522040.1 XM_011522042.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3988143 c.5050C>T p.L1684L Chr.15: 77114347-77114347 on Build GRCh38
6743799 c.5225T>G p.I1742S Chr.15: 77114172-77114172 on Build GRCh38
5614284 c.5119G>T p.A1707S Chr.15: 77114278-77114278 on Build GRCh38
1940079 c.5227C>T p.L1743L Chr.15: 77114170-77114170 on Build GRCh38
3988142 c.5050C>T p.L1684L Chr.15: 77114347-77114347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766447394 Chr.15: 77114150-77114150 on Build GRCh38
rs764379960 Chr.15: 77114327-77114327 on Build GRCh38
rs187996767 Chr.15: 77114348-77114348 on Build GRCh38
rs201627499 Chr.15: 77114359-77114359 on Build GRCh38
rs180909441 Chr.15: 77114360-77114360 on Build GRCh38
rs747206025 Chr.15: 77114266-77114266 on Build GRCh38
rs748032799 Chr.15: 77114220-77114220 on Build GRCh38
rs753445197 Chr.15: 77114352-77114352 on Build GRCh38
rs1027509828 Chr.15: 77114172-77114172 on Build GRCh38
rs777985165 Chr.15: 77114263-77114263 on Build GRCh38
rs772179237 Chr.15: 77114224-77114224 on Build GRCh38
rs61732437 Chr.15: 77114165-77114165 on Build GRCh38
rs371351051 Chr.15: 77114329-77114329 on Build GRCh38
rs779734780 Chr.15: 77114193-77114193 on Build GRCh38
rs751969726 Chr.15: 77114290-77114290 on Build GRCh38
rs765543180 Chr.15: 77114160-77114160 on Build GRCh38
rs767270694 Chr.15: 77114177-77114177 on Build GRCh38
rs1008443089 Chr.15: 77114287-77114287 on Build GRCh38
rs755008125 Chr.15: 77114204-77114204 on Build GRCh38
rs377408644 Chr.15: 77114154-77114154 on Build GRCh38
rs374551274 Chr.15: 77114185-77114185 on Build GRCh38
rs34004337 Chr.15: 77114302-77114302 on Build GRCh38
rs374121790 Chr.15: 77114354-77114354 on Build GRCh38
rs755593639 Chr.15: 77114189-77114189 on Build GRCh38
rs796487315 Chr.15: 77114213-77114213 on Build GRCh38
rs758375500 Chr.15: 77114277-77114277 on Build GRCh38
rs763314529 Chr.15: 77114309-77114309 on Build GRCh38
rs750044340 Chr.15: 77114330-77114330 on Build GRCh38
rs776720120 Chr.15: 77114289-77114289 on Build GRCh38
rs760436165 Chr.15: 77114344-77114344 on Build GRCh38
rs780185098 Chr.15: 77114278-77114278 on Build GRCh38
rs770946174 Chr.15: 77114283-77114283 on Build GRCh38
rs760055004 Chr.15: 77114156-77114156 on Build GRCh38
rs770347223 Chr.15: 77114301-77114301 on Build GRCh38
rs761361923 Chr.15: 77114166-77114166 on Build GRCh38
rs778827709 Chr.15: 77114211-77114211 on Build GRCh38
rs753788084 Chr.15: 77114203-77114203 on Build GRCh38

View Full Product Details