Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30784354-30784579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF3
Entrez Gene ID
51379
Gene Name
cytokine receptor like factor 3
Gene Aliases
CREME-9, CREME9, CRLM9, CYTOR4, FRWS, p48.2
UniGene
Hs.649372
Gene Chromosome Location
Chr.17: 30782684-30824760 on Build GRCh38
Transcripts
NM_015986.3 NR_073118.1 XM_011524879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363679 c.1154A>G p.E385G Chr.17: 30784362-30784362 on Build GRCh38
5129505 c.1158C>T p.A386A Chr.17: 30784358-30784358 on Build GRCh38
1128768 c.1162A>C p.T388P Chr.17: 30784354-30784354 on Build GRCh38
4965360 c.1159G>A p.V387M Chr.17: 30784357-30784357 on Build GRCh38
6501449 c.1084G>C p.V362L Chr.17: 30784432-30784432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913631928 Chr.17: 30784394-30784394 on Build GRCh38
rs554016959 Chr.17: 30784453-30784457 on Build GRCh38
rs780082061 Chr.17: 30784456-30784456 on Build GRCh38
rs113727899 Chr.17: 30784506-30784506 on Build GRCh38
rs981800206 Chr.17: 30784482-30784482 on Build GRCh38
rs747366932 Chr.17: 30784417-30784421 on Build GRCh38
rs767225232 Chr.17: 30784428-30784428 on Build GRCh38
rs907355009 Chr.17: 30784507-30784507 on Build GRCh38
rs766976106 Chr.17: 30784387-30784387 on Build GRCh38
rs769101368 Chr.17: 30784449-30784453 on Build GRCh38
rs111711307 Chr.17: 30784399-30784399 on Build GRCh38
rs887824952 Chr.17: 30784542-30784542 on Build GRCh38
rs927689268 Chr.17: 30784491-30784491 on Build GRCh38
rs76343572 Chr.17: 30784535-30784535 on Build GRCh38
rs750015318 Chr.17: 30784439-30784439 on Build GRCh38
rs772338217 Chr.17: 30784479-30784479 on Build GRCh38
rs777967213 Chr.17: 30784495-30784495 on Build GRCh38
rs758358535 Chr.17: 30784410-30784410 on Build GRCh38
rs755578376 Chr.17: 30784445-30784445 on Build GRCh38
rs778681113 Chr.17: 30784472-30784472 on Build GRCh38
rs987945475 Chr.17: 30784382-30784382 on Build GRCh38
rs773292810 Chr.17: 30784374-30784374 on Build GRCh38
rs929747993 Chr.17: 30784493-30784493 on Build GRCh38
rs542565383 Chr.17: 30784514-30784514 on Build GRCh38
rs759996220 Chr.17: 30784400-30784400 on Build GRCh38
rs754901398 Chr.17: 30784464-30784464 on Build GRCh38
rs189363338 Chr.17: 30784556-30784556 on Build GRCh38
rs779375392 Chr.17: 30784459-30784459 on Build GRCh38
rs1006628670 Chr.17: 30784547-30784547 on Build GRCh38
rs753071686 Chr.17: 30784407-30784407 on Build GRCh38
rs762349741 Chr.17: 30784480-30784481 on Build GRCh38
rs747875828 Chr.17: 30784477-30784477 on Build GRCh38
rs756975854 Chr.17: 30784411-30784411 on Build GRCh38
rs1023275277 Chr.17: 30784430-30784430 on Build GRCh38
rs375115688 Chr.17: 30784396-30784396 on Build GRCh38
rs760729327 Chr.17: 30784378-30784378 on Build GRCh38
rs937624972 Chr.17: 30784515-30784515 on Build GRCh38

View Full Product Details