Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2801302-2801533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP4
Entrez Gene ID
2288
Gene Name
FK506 binding protein 4
Gene Aliases
FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52
UniGene
Hs.524183
Gene Chromosome Location
Chr.12: 2794942-2805423 on Build GRCh38
Transcripts
NM_002014.3 XM_011520929.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2204677 c.1228C>T p.L410F Chr.12: 2801312-2801312 on Build GRCh38
4041284 c.1230C>T p.L410L Chr.12: 2801314-2801314 on Build GRCh38
3968165 c.1266G>A p.E422E Chr.12: 2801350-2801350 on Build GRCh38
3954625 c.1254G>A p.L418L Chr.12: 2801338-2801338 on Build GRCh38
5663246 c.1260G>A p.E420E Chr.12: 2801344-2801344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372618932 Chr.12: 2801393-2801393 on Build GRCh38
rs959413061 Chr.12: 2801361-2801361 on Build GRCh38
rs369736046 Chr.12: 2801414-2801414 on Build GRCh38
rs1033322201 Chr.12: 2801350-2801350 on Build GRCh38
rs878943006 Chr.12: 2801374-2801374 on Build GRCh38
rs776738288 Chr.12: 2801399-2801399 on Build GRCh38
rs370229453 Chr.12: 2801377-2801377 on Build GRCh38
rs755891690 Chr.12: 2801371-2801371 on Build GRCh38
rs770067112 Chr.12: 2801408-2801408 on Build GRCh38
rs891183298 Chr.12: 2801461-2801461 on Build GRCh38
rs780443420 Chr.12: 2801389-2801389 on Build GRCh38
rs780892168 Chr.12: 2801364-2801364 on Build GRCh38
rs962406714 Chr.12: 2801452-2801452 on Build GRCh38
rs374303706 Chr.12: 2801383-2801383 on Build GRCh38
rs1054889765 Chr.12: 2801404-2801404 on Build GRCh38
rs932770627 Chr.12: 2801428-2801428 on Build GRCh38
rs762007075 Chr.12: 2801403-2801403 on Build GRCh38
rs879015932 Chr.12: 2801353-2801353 on Build GRCh38
rs898429519 Chr.12: 2801512-2801512 on Build GRCh38
rs995160594 Chr.12: 2801453-2801453 on Build GRCh38
rs746227190 Chr.12: 2801380-2801380 on Build GRCh38
rs191161145 Chr.12: 2801475-2801475 on Build GRCh38
rs149312843 Chr.12: 2801451-2801451 on Build GRCh38
rs1015414342 Chr.12: 2801485-2801485 on Build GRCh38
rs375946541 Chr.12: 2801342-2801342 on Build GRCh38
rs756555189 Chr.12: 2801356-2801357 on Build GRCh38
rs752376658 Chr.12: 2801366-2801366 on Build GRCh38
rs532018282 Chr.12: 2801493-2801493 on Build GRCh38
rs369416682 Chr.12: 2801391-2801391 on Build GRCh38
rs752845341 Chr.12: 2801342-2801344 on Build GRCh38
rs778229245 Chr.12: 2801376-2801383 on Build GRCh38
rs768689059 Chr.12: 2801392-2801392 on Build GRCh38

View Full Product Details