Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 9016249-9016522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERI1
Entrez Gene ID
90459
Gene Name
exoribonuclease 1
Gene Aliases
3'HEXO, HEXO, THEX1
UniGene
Hs.20000
Gene Chromosome Location
Chr.8: 8892225-9100262 on Build GRCh38
Transcripts
NM_153332.3 XM_005272401.2 XM_011543848.2 XM_011543849.1 XM_011543850.2 XM_011543851.1 XM_017013949.1 XM_017013950.1 XM_017013951.1 XM_017013952.1 XM_017013953.1 XM_017013954.1 XM_017013955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
751529 c.499-1G>T p.? Chr.8: 9016321-9016321 on Build GRCh38
751528 c.555C>G p.F185L Chr.8: 9016378-9016378 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760404463 Chr.8: 9016283-9016283 on Build GRCh38
rs754180504 Chr.8: 9016364-9016364 on Build GRCh38
rs545978361 Chr.8: 9016417-9016417 on Build GRCh38
rs374060224 Chr.8: 9016386-9016386 on Build GRCh38
rs764465639 Chr.8: 9016327-9016327 on Build GRCh38
rs768074086 Chr.8: 9016295-9016295 on Build GRCh38
rs776241254 Chr.8: 9016292-9016296 on Build GRCh38
rs200548227 Chr.8: 9016336-9016336 on Build GRCh38
rs555519734 Chr.8: 9016324-9016324 on Build GRCh38
rs768247644 Chr.8: 9016437-9016437 on Build GRCh38
rs569724361 Chr.8: 9016305-9016305 on Build GRCh38
rs556419888 Chr.8: 9016458-9016458 on Build GRCh38
rs766220583 Chr.8: 9016284-9016284 on Build GRCh38
rs1008655871 Chr.8: 9016469-9016469 on Build GRCh38
rs766868059 Chr.8: 9016450-9016450 on Build GRCh38
rs776540943 Chr.8: 9016325-9016325 on Build GRCh38
rs545301062 Chr.8: 9016429-9016429 on Build GRCh38
rs868309686 Chr.8: 9016373-9016373 on Build GRCh38
rs752205788 Chr.8: 9016291-9016291 on Build GRCh38
rs901398993 Chr.8: 9016480-9016480 on Build GRCh38
rs965594069 Chr.8: 9016486-9016487 on Build GRCh38
rs759275280 Chr.8: 9016326-9016326 on Build GRCh38
rs773164114 Chr.8: 9016277-9016277 on Build GRCh38
rs139484248 Chr.8: 9016378-9016378 on Build GRCh38
rs756530368 Chr.8: 9016301-9016301 on Build GRCh38
rs546173439 Chr.8: 9016430-9016430 on Build GRCh38
rs113245883 Chr.8: 9016415-9016415 on Build GRCh38
rs149839132 Chr.8: 9016422-9016422 on Build GRCh38
rs761428491 Chr.8: 9016441-9016441 on Build GRCh38
rs751542382 Chr.8: 9016403-9016403 on Build GRCh38
rs773120911 Chr.8: 9016314-9016314 on Build GRCh38
rs764633395 Chr.8: 9016290-9016290 on Build GRCh38
rs188096743 Chr.8: 9016307-9016307 on Build GRCh38
rs769450266 Chr.8: 9016328-9016328 on Build GRCh38
rs1041098106 Chr.8: 9016467-9016467 on Build GRCh38
rs971848322 Chr.8: 9016425-9016425 on Build GRCh38
rs750958948 Chr.8: 9016341-9016341 on Build GRCh38
rs766628427 Chr.8: 9016362-9016362 on Build GRCh38
rs761301348 Chr.8: 9016345-9016345 on Build GRCh38
rs771947781 Chr.8: 9016275-9016275 on Build GRCh38
rs942660992 Chr.8: 9016460-9016460 on Build GRCh38
rs948204617 Chr.8: 9016366-9016366 on Build GRCh38
rs752920657 Chr.8: 9016370-9016370 on Build GRCh38
rs759102125 Chr.8: 9016295-9016304 on Build GRCh38
rs150505422 Chr.8: 9016329-9016329 on Build GRCh38
rs748218783 Chr.8: 9016309-9016309 on Build GRCh38
rs576466474 Chr.8: 9016459-9016459 on Build GRCh38
rs779124008 Chr.8: 9016369-9016369 on Build GRCh38
rs750976737 Chr.8: 9016298-9016298 on Build GRCh38
rs764795012 Chr.8: 9016420-9016422 on Build GRCh38
rs183273578 Chr.8: 9016288-9016288 on Build GRCh38
rs746927963 Chr.8: 9016396-9016396 on Build GRCh38
rs780628752 Chr.8: 9016302-9016302 on Build GRCh38
rs772143919 Chr.8: 9016310-9016310 on Build GRCh38
rs774051573 Chr.8: 9016439-9016439 on Build GRCh38
rs762490722 Chr.8: 9016330-9016330 on Build GRCh38
rs920443563 Chr.8: 9016432-9016432 on Build GRCh38
rs757881533 Chr.8: 9016292-9016292 on Build GRCh38

View Full Product Details