Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135485263-135485536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R26
Entrez Gene ID
9858
Gene Name
protein phosphatase 1 regulatory subunit 26
Gene Aliases
KIAA0649
UniGene
Hs.533260
Gene Chromosome Location
Chr.9: 135477650-135488894 on Build GRCh38
Transcripts
NM_014811.3 XM_005263411.2 XM_017015355.1 XM_017015356.1 XM_017015357.1 XM_017015358.1 XM_017015359.1 XM_017015360.1 XM_017015361.1 XM_017015362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1643778 c.777C>T p.S259S Chr.9: 135485287-135485287 on Build GRCh38
3905499 c.977G>A p.R326H Chr.9: 135485487-135485487 on Build GRCh38
3655523 c.829C>T p.R277W Chr.9: 135485339-135485339 on Build GRCh38
3655522 c.797C>T p.S266F Chr.9: 135485307-135485307 on Build GRCh38
5583963 c.967A>T p.T323S Chr.9: 135485477-135485477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761538566 Chr.9: 135485487-135485487 on Build GRCh38
rs201945292 Chr.9: 135485504-135485504 on Build GRCh38
rs61743074 Chr.9: 135485455-135485455 on Build GRCh38
rs868766086 Chr.9: 135485472-135485472 on Build GRCh38
rs758067676 Chr.9: 135485341-135485341 on Build GRCh38
rs866649125 Chr.9: 135485345-135485352 on Build GRCh38
rs760394859 Chr.9: 135485502-135485502 on Build GRCh38
rs201645627 Chr.9: 135485340-135485340 on Build GRCh38
rs563753237 Chr.9: 135485391-135485391 on Build GRCh38
rs780658652 Chr.9: 135485369-135485369 on Build GRCh38
rs534583468 Chr.9: 135485446-135485446 on Build GRCh38
rs767658645 Chr.9: 135485321-135485321 on Build GRCh38
rs77234665 Chr.9: 135485356-135485356 on Build GRCh38
rs530929090 Chr.9: 135485397-135485397 on Build GRCh38
rs765150533 Chr.9: 135485337-135485337 on Build GRCh38
rs774881863 Chr.9: 135485382-135485382 on Build GRCh38
rs762856511 Chr.9: 135485466-135485466 on Build GRCh38
rs549878517 Chr.9: 135485408-135485408 on Build GRCh38
rs780369383 Chr.9: 135485376-135485376 on Build GRCh38
rs774650953 Chr.9: 135485315-135485315 on Build GRCh38
rs758771893 Chr.9: 135485517-135485517 on Build GRCh38
rs750103409 Chr.9: 135485411-135485411 on Build GRCh38
rs750002293 Chr.9: 135485501-135485501 on Build GRCh38
rs35490599 Chr.9: 135485357-135485357 on Build GRCh38
rs751406058 Chr.9: 135485401-135485401 on Build GRCh38
rs755532693 Chr.9: 135485506-135485506 on Build GRCh38
rs150290969 Chr.9: 135485486-135485486 on Build GRCh38
rs773333544 Chr.9: 135485388-135485388 on Build GRCh38
rs772414352 Chr.9: 135485440-135485440 on Build GRCh38
rs548732620 Chr.9: 135485398-135485398 on Build GRCh38
rs145135469 Chr.9: 135485429-135485429 on Build GRCh38
rs750583506 Chr.9: 135485332-135485332 on Build GRCh38
rs747262895 Chr.9: 135485453-135485453 on Build GRCh38
rs370270608 Chr.9: 135485433-135485433 on Build GRCh38
rs759491479 Chr.9: 135485335-135485335 on Build GRCh38
rs538582352 Chr.9: 135485491-135485491 on Build GRCh38
rs369529963 Chr.9: 135485320-135485320 on Build GRCh38
rs772467513 Chr.9: 135485385-135485385 on Build GRCh38
rs146594963 Chr.9: 135485407-135485407 on Build GRCh38
rs779626069 Chr.9: 135485510-135485510 on Build GRCh38
rs142983153 Chr.9: 135485379-135485379 on Build GRCh38
rs764059072 Chr.9: 135485473-135485473 on Build GRCh38
rs116980218 Chr.9: 135485434-135485434 on Build GRCh38
rs775221025 Chr.9: 135485396-135485396 on Build GRCh38
rs1056126900 Chr.9: 135485326-135485326 on Build GRCh38
rs147273345 Chr.9: 135485288-135485288 on Build GRCh38
rs765781609 Chr.9: 135485503-135485503 on Build GRCh38
rs71483260 Chr.9: 135485372-135485372 on Build GRCh38
rs953393104 Chr.9: 135485383-135485383 on Build GRCh38
rs753273862 Chr.9: 135485516-135485516 on Build GRCh38
rs141353268 Chr.9: 135485426-135485426 on Build GRCh38
rs760721442 Chr.9: 135485390-135485390 on Build GRCh38
rs149000148 Chr.9: 135485378-135485378 on Build GRCh38
rs756631202 Chr.9: 135485373-135485373 on Build GRCh38
rs755511009 Chr.9: 135485415-135485415 on Build GRCh38
rs776797025 Chr.9: 135485461-135485461 on Build GRCh38
rs756924041 Chr.9: 135485366-135485366 on Build GRCh38

View Full Product Details