Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27452821-27453050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF184
Entrez Gene ID
7738
Gene Name
zinc finger protein 184
Gene Aliases
kr-ZNF3
UniGene
Hs.158174
Gene Chromosome Location
Chr.6: 27450742-27473118 on Build GRCh38
Transcripts
NM_001318891.1 NM_001318892.1 NM_001318893.1 NM_007149.2 XM_011514861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5357783 c.707C>T p.A236V Chr.6: 27452852-27452852 on Build GRCh38
6173347 c.550G>T p.G184W Chr.6: 27453009-27453009 on Build GRCh38
1442753 c.657G>T p.E219D Chr.6: 27452902-27452902 on Build GRCh38
7081324 c.668A>C p.K223T Chr.6: 27452891-27452891 on Build GRCh38
1621320 c.734C>T p.T245I Chr.6: 27452825-27452825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762401256 Chr.6: 27453026-27453026 on Build GRCh38
rs749440050 Chr.6: 27452983-27452983 on Build GRCh38
rs745593991 Chr.6: 27452974-27452974 on Build GRCh38
rs763062679 Chr.6: 27452872-27452872 on Build GRCh38
rs757890489 Chr.6: 27452932-27452932 on Build GRCh38
rs769862272 Chr.6: 27452857-27452857 on Build GRCh38
rs372124595 Chr.6: 27452918-27452918 on Build GRCh38
rs781064950 Chr.6: 27452958-27452958 on Build GRCh38
rs951401941 Chr.6: 27453012-27453012 on Build GRCh38
rs767851912 Chr.6: 27453027-27453027 on Build GRCh38
rs774528426 Chr.6: 27453015-27453015 on Build GRCh38
rs775655902 Chr.6: 27452859-27452859 on Build GRCh38
rs770062270 Chr.6: 27452975-27452975 on Build GRCh38
rs530759439 Chr.6: 27453008-27453008 on Build GRCh38
rs779035257 Chr.6: 27452904-27452904 on Build GRCh38
rs192196098 Chr.6: 27452956-27452956 on Build GRCh38
rs752102829 Chr.6: 27452884-27452884 on Build GRCh38
rs182253281 Chr.6: 27453002-27453002 on Build GRCh38
rs750701391 Chr.6: 27452901-27452901 on Build GRCh38
rs1004198374 Chr.6: 27453028-27453028 on Build GRCh38
rs775853516 Chr.6: 27452979-27452979 on Build GRCh38
rs376073172 Chr.6: 27452945-27452945 on Build GRCh38
rs759834697 Chr.6: 27452848-27452848 on Build GRCh38
rs764767432 Chr.6: 27452875-27452875 on Build GRCh38
rs368178770 Chr.6: 27452916-27452916 on Build GRCh38
rs762304704 Chr.6: 27452887-27452887 on Build GRCh38
rs139647756 Chr.6: 27452995-27452995 on Build GRCh38
rs770736899 Chr.6: 27452843-27452843 on Build GRCh38
rs866083572 Chr.6: 27452879-27452879 on Build GRCh38
rs45553937 Chr.6: 27452919-27452919 on Build GRCh38
rs143410636 Chr.6: 27452844-27452844 on Build GRCh38
rs746917706 Chr.6: 27452950-27452950 on Build GRCh38
rs757769656 Chr.6: 27452927-27452927 on Build GRCh38
rs374618294 Chr.6: 27452895-27452895 on Build GRCh38

View Full Product Details