Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 95274667-95274924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROM2
Entrez Gene ID
150696
Gene Name
prominin 2
Gene Aliases
PROML2
UniGene
Hs.437376
Gene Chromosome Location
Chr.2: 95274448-95291320 on Build GRCh38
Transcripts
NM_001165977.2 NM_001165978.2 NM_001321070.1 NM_144707.3 XM_011510672.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6756440 c.202C>T p.R68C Chr.2: 95274787-95274787 on Build GRCh38
5758496 c.203G>A p.R68H Chr.2: 95274788-95274788 on Build GRCh38
3933718 c.215C>T p.S72L Chr.2: 95274800-95274800 on Build GRCh38
460980 c.215C>G p.S72W Chr.2: 95274800-95274800 on Build GRCh38
2824150 c.148C>T p.L50L Chr.2: 95274733-95274733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774757628 Chr.2: 95274773-95274773 on Build GRCh38
rs748613828 Chr.2: 95274833-95274833 on Build GRCh38
rs765302602 Chr.2: 95274877-95274877 on Build GRCh38
rs746041078 Chr.2: 95274750-95274750 on Build GRCh38
rs200376966 Chr.2: 95274846-95274846 on Build GRCh38
rs752533853 Chr.2: 95274743-95274743 on Build GRCh38
rs746677602 Chr.2: 95274855-95274855 on Build GRCh38
rs765249855 Chr.2: 95274788-95274788 on Build GRCh38
rs567457297 Chr.2: 95274817-95274817 on Build GRCh38
rs373422154 Chr.2: 95274848-95274848 on Build GRCh38
rs200313492 Chr.2: 95274879-95274879 on Build GRCh38
rs769148897 Chr.2: 95274767-95274767 on Build GRCh38
rs776099688 Chr.2: 95274690-95274690 on Build GRCh38
rs780249714 Chr.2: 95274802-95274802 on Build GRCh38
rs774635026 Chr.2: 95274710-95274710 on Build GRCh38
rs538946414 Chr.2: 95274895-95274895 on Build GRCh38
rs775936462 Chr.2: 95274785-95274785 on Build GRCh38
rs757607037 Chr.2: 95274748-95274748 on Build GRCh38
rs530271538 Chr.2: 95274716-95274716 on Build GRCh38
rs146674837 Chr.2: 95274749-95274749 on Build GRCh38
rs755529989 Chr.2: 95274809-95274809 on Build GRCh38
rs552197251 Chr.2: 95274810-95274810 on Build GRCh38
rs760200490 Chr.2: 95274728-95274728 on Build GRCh38
rs765125055 Chr.2: 95274740-95274740 on Build GRCh38
rs375545886 Chr.2: 95274800-95274800 on Build GRCh38
rs765762536 Chr.2: 95274752-95274752 on Build GRCh38
rs764263969 Chr.2: 95274871-95274871 on Build GRCh38
rs752763995 Chr.2: 95274784-95274784 on Build GRCh38
rs759091355 Chr.2: 95274873-95274873 on Build GRCh38
rs370988812 Chr.2: 95274787-95274787 on Build GRCh38
rs35733728 Chr.2: 95274903-95274903 on Build GRCh38
rs770249622 Chr.2: 95274689-95274689 on Build GRCh38
rs769024446 Chr.2: 95274698-95274698 on Build GRCh38
rs879491950 Chr.2: 95274718-95274718 on Build GRCh38
rs201105654 Chr.2: 95274801-95274801 on Build GRCh38
rs749809675 Chr.2: 95274762-95274762 on Build GRCh38
rs35203035 Chr.2: 95274879-95274880 on Build GRCh38
rs746729119 Chr.2: 95274689-95274690 on Build GRCh38
rs773388506 Chr.2: 95274847-95274847 on Build GRCh38
rs113477824 Chr.2: 95274880-95274880 on Build GRCh38
rs550658106 Chr.2: 95274805-95274805 on Build GRCh38
rs753833989 Chr.2: 95274736-95274736 on Build GRCh38
rs138718796 Chr.2: 95274727-95274727 on Build GRCh38
rs748491200 Chr.2: 95274776-95274776 on Build GRCh38
rs754085916 Chr.2: 95274875-95274875 on Build GRCh38
rs199870049 Chr.2: 95274857-95274857 on Build GRCh38
rs754812539 Chr.2: 95274738-95274738 on Build GRCh38
rs150054018 Chr.2: 95274753-95274753 on Build GRCh38
rs546884538 Chr.2: 95274719-95274719 on Build GRCh38
rs377370048 Chr.2: 95274778-95274778 on Build GRCh38
rs140148451 Chr.2: 95274783-95274783 on Build GRCh38
rs758232721 Chr.2: 95274791-95274791 on Build GRCh38
rs571240281 Chr.2: 95274790-95274790 on Build GRCh38
rs763937191 Chr.2: 95274793-95274793 on Build GRCh38

View Full Product Details