Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69055053-69055547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPCN2
Entrez Gene ID
219931
Gene Name
two pore segment channel 2
Gene Aliases
SHEP10, TPC2
UniGene
Hs.131851
Gene Chromosome Location
Chr.11: 69048876-69090604 on Build GRCh38
Transcripts
NM_139075.3 XM_005273824.3 XM_005273826.3 XM_005273830.3 XM_005273831.3 XM_005273832.3 XM_011544802.2 XM_011544805.1 XM_011544806.1 XM_011544807.2 XM_011544808.2 XM_017017328.1 XM_017017329.1 XM_017017330.1 XM_017017331.1 XM_017017332.1 XM_017017333.1 XM_017017334.1 XM_017017335.1 XM_017017336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3670882 c.339C>A p.Y113* Chr.11: 69055262-69055262 on Build GRCh38
5787097 c.258G>C p.L86F Chr.11: 69055181-69055181 on Build GRCh38
7159708 c.305C>T p.P102L Chr.11: 69055228-69055228 on Build GRCh38
6841591 c.358C>T p.P120S Chr.11: 69055281-69055281 on Build GRCh38
931348 c.342C>T p.R114R Chr.11: 69055265-69055265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535139656 Chr.11: 69055241-69055241 on Build GRCh38
rs778919810 Chr.11: 69055199-69055199 on Build GRCh38
rs112634076 Chr.11: 69055475-69055475 on Build GRCh38
rs759510555 Chr.11: 69055132-69055132 on Build GRCh38
rs199743860 Chr.11: 69055264-69055264 on Build GRCh38
rs980027366 Chr.11: 69055109-69055109 on Build GRCh38
rs922060933 Chr.11: 69055427-69055427 on Build GRCh38
rs150684835 Chr.11: 69055419-69055419 on Build GRCh38
rs116215960 Chr.11: 69055086-69055086 on Build GRCh38
rs372815048 Chr.11: 69055249-69055249 on Build GRCh38
rs772440038 Chr.11: 69055338-69055338 on Build GRCh38
rs1009360573 Chr.11: 69055284-69055284 on Build GRCh38
rs370156980 Chr.11: 69055265-69055265 on Build GRCh38
rs759083583 Chr.11: 69055347-69055347 on Build GRCh38
rs771380104 Chr.11: 69055398-69055398 on Build GRCh38
rs752419441 Chr.11: 69055360-69055360 on Build GRCh38
rs1031875212 Chr.11: 69055266-69055266 on Build GRCh38
rs150386880 Chr.11: 69055184-69055184 on Build GRCh38
rs539160820 Chr.11: 69055489-69055489 on Build GRCh38
rs1009955270 Chr.11: 69055084-69055084 on Build GRCh38
rs1038408263 Chr.11: 69055216-69055216 on Build GRCh38
rs374040368 Chr.11: 69055377-69055377 on Build GRCh38
rs774592065 Chr.11: 69055323-69055323 on Build GRCh38
rs115204200 Chr.11: 69055190-69055190 on Build GRCh38
rs773165448 Chr.11: 69055138-69055138 on Build GRCh38
rs757823111 Chr.11: 69055164-69055164 on Build GRCh38
rs376409865 Chr.11: 69055166-69055166 on Build GRCh38
rs200172503 Chr.11: 69055333-69055333 on Build GRCh38
rs768750333 Chr.11: 69055383-69055383 on Build GRCh38
rs12287555 Chr.11: 69055293-69055293 on Build GRCh38
rs199768477 Chr.11: 69055253-69055253 on Build GRCh38
rs1049775536 Chr.11: 69055406-69055406 on Build GRCh38
rs944904769 Chr.11: 69055255-69055255 on Build GRCh38
rs142221271 Chr.11: 69055226-69055226 on Build GRCh38
rs12288978 Chr.11: 69055513-69055513 on Build GRCh38
rs961201910 Chr.11: 69055492-69055492 on Build GRCh38
rs1010006089 Chr.11: 69055337-69055337 on Build GRCh38
rs559693513 Chr.11: 69055291-69055291 on Build GRCh38
rs939193188 Chr.11: 69055135-69055135 on Build GRCh38
rs201853135 Chr.11: 69055282-69055282 on Build GRCh38
rs781156910 Chr.11: 69055378-69055378 on Build GRCh38
rs891415329 Chr.11: 69055334-69055334 on Build GRCh38
rs748374933 Chr.11: 69055385-69055385 on Build GRCh38
rs919737168 Chr.11: 69055516-69055516 on Build GRCh38
rs778686638 Chr.11: 69055275-69055275 on Build GRCh38
rs746306487 Chr.11: 69055221-69055221 on Build GRCh38
rs868476650 Chr.11: 69055285-69055285 on Build GRCh38
rs1011718986 Chr.11: 69055444-69055444 on Build GRCh38
rs538157837 Chr.11: 69055114-69055114 on Build GRCh38
rs143487572 Chr.11: 69055346-69055346 on Build GRCh38
rs1043878439 Chr.11: 69055447-69055447 on Build GRCh38
rs898528215 Chr.11: 69055224-69055224 on Build GRCh38
rs750953918 Chr.11: 69055259-69055259 on Build GRCh38
rs760070294 Chr.11: 69055401-69055401 on Build GRCh38
rs142764445 Chr.11: 69055298-69055298 on Build GRCh38
rs991927040 Chr.11: 69055172-69055172 on Build GRCh38
rs944307977 Chr.11: 69055409-69055409 on Build GRCh38
rs980433532 Chr.11: 69055357-69055357 on Build GRCh38
rs145775659 Chr.11: 69055263-69055263 on Build GRCh38
rs746923249 Chr.11: 69055240-69055240 on Build GRCh38
rs190040116 Chr.11: 69055250-69055250 on Build GRCh38
rs764799947 Chr.11: 69055359-69055359 on Build GRCh38
rs373181469 Chr.11: 69055163-69055163 on Build GRCh38
rs541105565 Chr.11: 69055290-69055290 on Build GRCh38
rs749977548 Chr.11: 69055182-69055182 on Build GRCh38
rs552445433 Chr.11: 69055081-69055081 on Build GRCh38
rs778397171 Chr.11: 69055476-69055476 on Build GRCh38
rs201107466 Chr.11: 69055387-69055387 on Build GRCh38
rs751361901 Chr.11: 69055372-69055372 on Build GRCh38
rs756697476 Chr.11: 69055399-69055399 on Build GRCh38
rs753451447 Chr.11: 69055312-69055312 on Build GRCh38
rs773250590 Chr.11: 69055227-69055227 on Build GRCh38
rs758062897 Chr.11: 69055361-69055361 on Build GRCh38
rs1015424889 Chr.11: 69055490-69055490 on Build GRCh38
rs761077360 Chr.11: 69055397-69055397 on Build GRCh38
rs369434788 Chr.11: 69055189-69055189 on Build GRCh38
rs776907811 Chr.11: 69055131-69055131 on Build GRCh38
rs114020605 Chr.11: 69055501-69055501 on Build GRCh38
rs768909741 Chr.11: 69055302-69055302 on Build GRCh38
rs200548419 Chr.11: 69055289-69055289 on Build GRCh38
rs772129471 Chr.11: 69055127-69055127 on Build GRCh38
rs761153685 Chr.11: 69055345-69055345 on Build GRCh38
rs185329045 Chr.11: 69055077-69055077 on Build GRCh38
rs1470892 Chr.11: 69055271-69055271 on Build GRCh38
rs960354485 Chr.11: 69055486-69055486 on Build GRCh38
rs752580675 Chr.11: 69055280-69055280 on Build GRCh38
rs767830813 Chr.11: 69055258-69055258 on Build GRCh38
rs779361967 Chr.11: 69055093-69055093 on Build GRCh38
rs903577441 Chr.11: 69055462-69055462 on Build GRCh38
rs765535152 Chr.11: 69055137-69055137 on Build GRCh38
rs763635872 Chr.11: 69055271-69055271 on Build GRCh38
rs971194862 Chr.11: 69055104-69055104 on Build GRCh38
rs745755451 Chr.11: 69055382-69055382 on Build GRCh38
rs553492385 Chr.11: 69055423-69055423 on Build GRCh38
rs962927046 Chr.11: 69055162-69055162 on Build GRCh38
rs763184008 Chr.11: 69055139-69055139 on Build GRCh38
rs772693267 Chr.11: 69055407-69055407 on Build GRCh38
rs764375724 Chr.11: 69055147-69055147 on Build GRCh38
rs762138291 Chr.11: 69055257-69055257 on Build GRCh38
rs562272031 Chr.11: 69055283-69055283 on Build GRCh38
rs775580353 Chr.11: 69055246-69055246 on Build GRCh38
rs72917389 Chr.11: 69055363-69055363 on Build GRCh38
rs960633111 Chr.11: 69055366-69055366 on Build GRCh38
rs1022414938 Chr.11: 69055076-69055076 on Build GRCh38

View Full Product Details