Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78067909-78068454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFC2
Entrez Gene ID
4718
Gene Name
NADH:ubiquinone oxidoreductase subunit C2
Gene Aliases
B14.5b, CI-B14.5b, HLC-1, NADHDH2
UniGene
Hs.407860
Gene Chromosome Location
Chr.11: 78068347-78080219 on Build GRCh38
Transcripts
NM_001204054.1 NM_001204055.1 NM_004549.5 NM_001203260.1 NM_001203261.1 NM_001203262.1 NM_003251.3

Gene Symbol
NDUFC2-KCTD14
Entrez Gene ID
100532726
Gene Name
NDUFC2-KCTD14 readthrough
Gene Aliases
-
UniGene
Hs.407860
Gene Chromosome Location
Chr.11: 78015715-78080219 on Build GRCh38
Transcripts
NM_001204054.1 NM_001204055.1 NM_004549.5 NM_001203260.1 NM_001203261.1 NM_001203262.1 NM_003251.3

Gene Symbol
THRSP
Entrez Gene ID
7069
Gene Name
thyroid hormone responsive
Gene Aliases
LPGP1, Lpgp, S14, SPOT14, THRP
UniGene
Hs.591969
Gene Chromosome Location
Chr.11: 78063861-78068357 on Build GRCh38
Transcripts
NM_001204054.1 NM_001204055.1 NM_004549.5 NM_001203260.1 NM_001203261.1 NM_001203262.1 NM_003251.3

SNP Detail

SNP ID Chromosome Location
rs911883566 Chr.11: 78068271-78068271 on Build GRCh38
rs79927345 Chr.11: 78068055-78068055 on Build GRCh38
rs772989486 Chr.11: 78068078-78068079 on Build GRCh38
rs969866000 Chr.11: 78068262-78068265 on Build GRCh38
rs637887 Chr.11: 78068242-78068242 on Build GRCh38
rs1013927474 Chr.11: 78068037-78068037 on Build GRCh38
rs529040717 Chr.11: 78068158-78068158 on Build GRCh38
rs936293235 Chr.11: 78067993-78067993 on Build GRCh38
rs561758442 Chr.11: 78067951-78067951 on Build GRCh38
rs750477289 Chr.11: 78068186-78068186 on Build GRCh38
rs966186895 Chr.11: 78068428-78068428 on Build GRCh38
rs944850410 Chr.11: 78068051-78068051 on Build GRCh38
rs913983468 Chr.11: 78068211-78068211 on Build GRCh38
rs565283112 Chr.11: 78068132-78068132 on Build GRCh38
rs930446292 Chr.11: 78068340-78068340 on Build GRCh38
rs3832724 Chr.11: 78068004-78068007 on Build GRCh38
rs899013413 Chr.11: 78067962-78067962 on Build GRCh38
rs989001812 Chr.11: 78068036-78068036 on Build GRCh38
rs899110773 Chr.11: 78068314-78068314 on Build GRCh38
rs1038826488 Chr.11: 78067956-78067956 on Build GRCh38
rs925599929 Chr.11: 78067964-78067964 on Build GRCh38
rs945497219 Chr.11: 78068236-78068236 on Build GRCh38
rs943276279 Chr.11: 78068272-78068272 on Build GRCh38
rs550877899 Chr.11: 78068414-78068414 on Build GRCh38
rs913454936 Chr.11: 78068416-78068416 on Build GRCh38
rs754102468 Chr.11: 78067940-78067940 on Build GRCh38
rs199883650 Chr.11: 78068003-78068006 on Build GRCh38
rs1047635228 Chr.11: 78068360-78068360 on Build GRCh38
rs757351739 Chr.11: 78067969-78067969 on Build GRCh38
rs896998097 Chr.11: 78068204-78068204 on Build GRCh38
rs545163750 Chr.11: 78068097-78068097 on Build GRCh38
rs970972276 Chr.11: 78068069-78068069 on Build GRCh38
rs550274200 Chr.11: 78068182-78068182 on Build GRCh38
rs932057244 Chr.11: 78068100-78068100 on Build GRCh38
rs1039377049 Chr.11: 78068277-78068277 on Build GRCh38
rs989630178 Chr.11: 78068208-78068208 on Build GRCh38
rs143136237 Chr.11: 78068183-78068183 on Build GRCh38
rs532861109 Chr.11: 78068374-78068374 on Build GRCh38
rs886213938 Chr.11: 78068027-78068027 on Build GRCh38
rs1026056861 Chr.11: 78068020-78068020 on Build GRCh38
rs987454866 Chr.11: 78068250-78068250 on Build GRCh38
rs1014023495 Chr.11: 78068389-78068389 on Build GRCh38
rs887763081 Chr.11: 78068157-78068158 on Build GRCh38
rs666124 Chr.11: 78068348-78068348 on Build GRCh38
rs980244328 Chr.11: 78068270-78068270 on Build GRCh38
rs560067432 Chr.11: 78068042-78068042 on Build GRCh38
rs1049095656 Chr.11: 78068114-78068114 on Build GRCh38
rs1024027866 Chr.11: 78067950-78067950 on Build GRCh38
rs550990549 Chr.11: 78068394-78068394 on Build GRCh38
rs1040446775 Chr.11: 78068062-78068062 on Build GRCh38

View Full Product Details