Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 90152250-90152768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAALAD2
Entrez Gene ID
10003
Gene Name
N-acetylated alpha-linked acidic dipeptidase 2
Gene Aliases
GCPIII, GPCIII, NAADALASE2, NAALADASE2
UniGene
Hs.112482
Gene Chromosome Location
Chr.11: 90131694-90193577 on Build GRCh38
Transcripts
NM_001300930.1 NM_005467.3 XM_017017043.1 XM_017017044.1 XM_017017045.1 XM_017017046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4705376 c.774C>G p.L258L Chr.11: 90152462-90152462 on Build GRCh38
395472 c.692C>A p.P231Q Chr.11: 90152380-90152380 on Build GRCh38
3953935 c.778C>A p.P260T Chr.11: 90152466-90152466 on Build GRCh38
5932812 c.784T>G p.Y262D Chr.11: 90152472-90152472 on Build GRCh38
5373779 c.645C>T p.I215I Chr.11: 90152333-90152333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760541192 Chr.11: 90152397-90152397 on Build GRCh38
rs563183977 Chr.11: 90152311-90152311 on Build GRCh38
rs766624593 Chr.11: 90152704-90152704 on Build GRCh38
rs1019555946 Chr.11: 90152527-90152527 on Build GRCh38
rs948892404 Chr.11: 90152708-90152708 on Build GRCh38
rs775929591 Chr.11: 90152327-90152327 on Build GRCh38
rs769024856 Chr.11: 90152524-90152524 on Build GRCh38
rs529891486 Chr.11: 90152550-90152550 on Build GRCh38
rs772056710 Chr.11: 90152529-90152529 on Build GRCh38
rs369582299 Chr.11: 90152685-90152685 on Build GRCh38
rs769048259 Chr.11: 90152359-90152359 on Build GRCh38
rs754748604 Chr.11: 90152446-90152446 on Build GRCh38
rs531379941 Chr.11: 90152333-90152333 on Build GRCh38
rs933870498 Chr.11: 90152408-90152408 on Build GRCh38
rs1030600817 Chr.11: 90152572-90152572 on Build GRCh38
rs7937708 Chr.11: 90152635-90152635 on Build GRCh38
rs375511564 Chr.11: 90152528-90152528 on Build GRCh38
rs757856804 Chr.11: 90152482-90152482 on Build GRCh38
rs984937477 Chr.11: 90152636-90152636 on Build GRCh38
rs767585417 Chr.11: 90152392-90152392 on Build GRCh38
rs921107186 Chr.11: 90152567-90152569 on Build GRCh38
rs965586095 Chr.11: 90152539-90152539 on Build GRCh38
rs760778385 Chr.11: 90152349-90152349 on Build GRCh38
rs896108326 Chr.11: 90152681-90152682 on Build GRCh38
rs897084814 Chr.11: 90152407-90152407 on Build GRCh38
rs940689109 Chr.11: 90152386-90152386 on Build GRCh38
rs764841434 Chr.11: 90152462-90152462 on Build GRCh38
rs554221382 Chr.11: 90152721-90152721 on Build GRCh38
rs372472392 Chr.11: 90152526-90152526 on Build GRCh38
rs1006681866 Chr.11: 90152497-90152497 on Build GRCh38
rs934356976 Chr.11: 90152651-90152651 on Build GRCh38
rs375623967 Chr.11: 90152292-90152292 on Build GRCh38
rs755926818 Chr.11: 90152284-90152284 on Build GRCh38
rs569619135 Chr.11: 90152618-90152618 on Build GRCh38
rs766302193 Chr.11: 90152410-90152410 on Build GRCh38
rs16916533 Chr.11: 90152632-90152632 on Build GRCh38
rs549854382 Chr.11: 90152602-90152602 on Build GRCh38
rs773038229 Chr.11: 90152396-90152396 on Build GRCh38
rs776061921 Chr.11: 90152353-90152353 on Build GRCh38
rs975309843 Chr.11: 90152560-90152560 on Build GRCh38
rs545034883 Chr.11: 90152339-90152339 on Build GRCh38
rs752340814 Chr.11: 90152465-90152465 on Build GRCh38
rs950711606 Chr.11: 90152443-90152443 on Build GRCh38
rs780683722 Chr.11: 90152510-90152510 on Build GRCh38
rs1026278278 Chr.11: 90152430-90152430 on Build GRCh38
rs778617123 Chr.11: 90152298-90152298 on Build GRCh38
rs766156333 Chr.11: 90152477-90152482 on Build GRCh38
rs540889664 Chr.11: 90152425-90152425 on Build GRCh38
rs547448777 Chr.11: 90152489-90152489 on Build GRCh38
rs140831197 Chr.11: 90152357-90152357 on Build GRCh38
rs770283714 Chr.11: 90152346-90152346 on Build GRCh38
rs899281073 Chr.11: 90152320-90152320 on Build GRCh38
rs761830046 Chr.11: 90152387-90152387 on Build GRCh38
rs372480635 Chr.11: 90152289-90152289 on Build GRCh38
rs756740954 Chr.11: 90152507-90152507 on Build GRCh38
rs889603251 Chr.11: 90152456-90152456 on Build GRCh38
rs73543373 Chr.11: 90152299-90152299 on Build GRCh38
rs369002435 Chr.11: 90152297-90152297 on Build GRCh38
rs909296514 Chr.11: 90152679-90152679 on Build GRCh38
rs972075283 Chr.11: 90152693-90152693 on Build GRCh38
rs773397097 Chr.11: 90152533-90152533 on Build GRCh38
rs766105688 Chr.11: 90152287-90152287 on Build GRCh38
rs916730180 Chr.11: 90152283-90152283 on Build GRCh38
rs567384649 Chr.11: 90152512-90152512 on Build GRCh38
rs201051787 Chr.11: 90152369-90152369 on Build GRCh38
rs746511527 Chr.11: 90152496-90152496 on Build GRCh38
rs753496379 Chr.11: 90152295-90152295 on Build GRCh38

View Full Product Details