Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123977289-123977785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10S1
Entrez Gene ID
219873
Gene Name
olfactory receptor family 10 subfamily S member 1
Gene Aliases
OR11-279
UniGene
Hs.553637
Gene Chromosome Location
Chr.11: 123976696-123977691 on Build GRCh38
Transcripts
NM_001004474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703902 c.73T>G p.F25V Chr.11: 123977619-123977619 on Build GRCh38
3868680 c.30G>A p.M10I Chr.11: 123977662-123977662 on Build GRCh38
3868678 c.168C>T p.L56L Chr.11: 123977524-123977524 on Build GRCh38
4478693 c.225C>T p.F75F Chr.11: 123977467-123977467 on Build GRCh38
7330373 c.202C>A p.L68I Chr.11: 123977490-123977490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755973894 Chr.11: 123977582-123977582 on Build GRCh38
rs145346852 Chr.11: 123977544-123977544 on Build GRCh38
rs966820200 Chr.11: 123977373-123977373 on Build GRCh38
rs373541476 Chr.11: 123977374-123977374 on Build GRCh38
rs148762471 Chr.11: 123977455-123977455 on Build GRCh38
rs902593699 Chr.11: 123977639-123977639 on Build GRCh38
rs775734563 Chr.11: 123977355-123977355 on Build GRCh38
rs775185165 Chr.11: 123977684-123977684 on Build GRCh38
rs574200105 Chr.11: 123977670-123977673 on Build GRCh38
rs1011385354 Chr.11: 123977680-123977680 on Build GRCh38
rs745935163 Chr.11: 123977500-123977500 on Build GRCh38
rs780210753 Chr.11: 123977627-123977627 on Build GRCh38
rs942245839 Chr.11: 123977559-123977559 on Build GRCh38
rs17759447 Chr.11: 123977357-123977357 on Build GRCh38
rs938859564 Chr.11: 123977746-123977746 on Build GRCh38
rs752584599 Chr.11: 123977466-123977466 on Build GRCh38
rs758375947 Chr.11: 123977618-123977618 on Build GRCh38
rs765955247 Chr.11: 123977542-123977542 on Build GRCh38
rs575104063 Chr.11: 123977676-123977676 on Build GRCh38
rs147657646 Chr.11: 123977671-123977671 on Build GRCh38
rs970009407 Chr.11: 123977707-123977707 on Build GRCh38
rs746047640 Chr.11: 123977410-123977410 on Build GRCh38
rs765942640 Chr.11: 123977452-123977452 on Build GRCh38
rs762599678 Chr.11: 123977451-123977451 on Build GRCh38
rs779839829 Chr.11: 123977383-123977383 on Build GRCh38
rs764877400 Chr.11: 123977360-123977360 on Build GRCh38
rs748125574 Chr.11: 123977392-123977392 on Build GRCh38
rs777244702 Chr.11: 123977425-123977425 on Build GRCh38
rs372624131 Chr.11: 123977386-123977386 on Build GRCh38
rs139752751 Chr.11: 123977364-123977364 on Build GRCh38
rs928341913 Chr.11: 123977491-123977491 on Build GRCh38
rs770960376 Chr.11: 123977398-123977398 on Build GRCh38
rs536775538 Chr.11: 123977490-123977490 on Build GRCh38
rs748421047 Chr.11: 123977730-123977730 on Build GRCh38
rs1006954352 Chr.11: 123977616-123977616 on Build GRCh38
rs773836292 Chr.11: 123977532-123977532 on Build GRCh38
rs747148840 Chr.11: 123977633-123977633 on Build GRCh38
rs762347379 Chr.11: 123977538-123977538 on Build GRCh38
rs759711106 Chr.11: 123977334-123977334 on Build GRCh38
rs771048444 Chr.11: 123977321-123977321 on Build GRCh38
rs773208928 Chr.11: 123977315-123977315 on Build GRCh38
rs905993345 Chr.11: 123977446-123977446 on Build GRCh38
rs778989901 Chr.11: 123977408-123977408 on Build GRCh38
rs755261842 Chr.11: 123977640-123977640 on Build GRCh38
rs763986055 Chr.11: 123977709-123977709 on Build GRCh38
rs557897828 Chr.11: 123977556-123977556 on Build GRCh38
rs957267145 Chr.11: 123977422-123977422 on Build GRCh38
rs762279536 Chr.11: 123977450-123977450 on Build GRCh38
rs181255416 Chr.11: 123977726-123977726 on Build GRCh38
rs781085900 Chr.11: 123977387-123977387 on Build GRCh38
rs1017596519 Chr.11: 123977346-123977346 on Build GRCh38
rs749358345 Chr.11: 123977319-123977319 on Build GRCh38
rs867980308 Chr.11: 123977329-123977329 on Build GRCh38
rs781634623 Chr.11: 123977649-123977649 on Build GRCh38
rs759124937 Chr.11: 123977678-123977678 on Build GRCh38
rs11219384 Chr.11: 123977403-123977403 on Build GRCh38
rs560485253 Chr.11: 123977385-123977385 on Build GRCh38
rs768110985 Chr.11: 123977563-123977563 on Build GRCh38
rs758006350 Chr.11: 123977366-123977366 on Build GRCh38
rs769187595 Chr.11: 123977420-123977420 on Build GRCh38
rs751315243 Chr.11: 123977384-123977384 on Build GRCh38
rs769099597 Chr.11: 123977522-123977522 on Build GRCh38
rs977652183 Chr.11: 123977393-123977393 on Build GRCh38
rs749834130 Chr.11: 123977668-123977668 on Build GRCh38
rs17759513 Chr.11: 123977505-123977505 on Build GRCh38
rs751051422 Chr.11: 123977554-123977562 on Build GRCh38
rs138137631 Chr.11: 123977575-123977575 on Build GRCh38
rs146636527 Chr.11: 123977438-123977438 on Build GRCh38
rs764689709 Chr.11: 123977356-123977356 on Build GRCh38
rs770428554 Chr.11: 123977529-123977529 on Build GRCh38
rs867042545 Chr.11: 123977607-123977607 on Build GRCh38
rs186017158 Chr.11: 123977593-123977593 on Build GRCh38
rs777156744 Chr.11: 123977527-123977527 on Build GRCh38
rs1047920342 Chr.11: 123977512-123977512 on Build GRCh38
rs780113022 Chr.11: 123977510-123977510 on Build GRCh38
rs368590825 Chr.11: 123977327-123977327 on Build GRCh38
rs371936332 Chr.11: 123977718-123977718 on Build GRCh38
rs754698227 Chr.11: 123977456-123977456 on Build GRCh38
rs201821946 Chr.11: 123977638-123977638 on Build GRCh38
rs142345245 Chr.11: 123977681-123977681 on Build GRCh38
rs140511439 Chr.11: 123977528-123977528 on Build GRCh38
rs372266906 Chr.11: 123977543-123977543 on Build GRCh38
rs983226208 Chr.11: 123977721-123977721 on Build GRCh38
rs773926642 Chr.11: 123977672-123977672 on Build GRCh38
rs752496595 Chr.11: 123977578-123977578 on Build GRCh38
rs367636323 Chr.11: 123977432-123977432 on Build GRCh38
rs760436923 Chr.11: 123977692-123977692 on Build GRCh38
rs781550775 Chr.11: 123977729-123977729 on Build GRCh38
rs200775631 Chr.11: 123977427-123977427 on Build GRCh38
rs1006514452 Chr.11: 123977330-123977330 on Build GRCh38
rs138853243 Chr.11: 123977710-123977710 on Build GRCh38
rs981134378 Chr.11: 123977476-123977476 on Build GRCh38
rs760930356 Chr.11: 123977419-123977419 on Build GRCh38
rs527686929 Chr.11: 123977712-123977712 on Build GRCh38
rs544829792 Chr.11: 123977651-123977651 on Build GRCh38
rs780998182 Chr.11: 123977461-123977461 on Build GRCh38
rs752408882 Chr.11: 123977609-123977609 on Build GRCh38
rs759210760 Chr.11: 123977549-123977549 on Build GRCh38
rs773561934 Chr.11: 123977659-123977659 on Build GRCh38
rs112409724 Chr.11: 123977514-123977514 on Build GRCh38
rs748517163 Chr.11: 123977650-123977650 on Build GRCh38
rs376676003 Chr.11: 123977413-123977413 on Build GRCh38
rs759025210 Chr.11: 123977553-123977554 on Build GRCh38
rs951883197 Chr.11: 123977350-123977350 on Build GRCh38
rs765746305 Chr.11: 123977554-123977556 on Build GRCh38
rs755175602 Chr.11: 123977727-123977727 on Build GRCh38
rs1057510419 Chr.11: 123977761-123977761 on Build GRCh38
rs767157382 Chr.11: 123977569-123977571 on Build GRCh38
rs17686210 Chr.11: 123977414-123977414 on Build GRCh38
rs767876382 Chr.11: 123977348-123977348 on Build GRCh38
rs778901930 Chr.11: 123977493-123977493 on Build GRCh38

View Full Product Details