Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124630250-124630774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBRG1
Entrez Gene ID
84897
Gene Name
transforming growth factor beta regulator 1
Gene Aliases
NIAM, TB-5
UniGene
Hs.436410
Gene Chromosome Location
Chr.11: 124622599-124635926 on Build GRCh38
Transcripts
NM_032811.2 NR_016021.1 XM_005271709.3 XM_017018442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7142756 c.802C>A p.H268N Chr.11: 124630451-124630451 on Build GRCh38
7210385 c.835A>G p.M279V Chr.11: 124630484-124630484 on Build GRCh38
1974763 c.413C>T p.A138V Chr.11: 124630774-124630774 on Build GRCh38
1135129 c.786T>C p.S262S Chr.11: 124630435-124630435 on Build GRCh38
247748 c.304G>C p.D102H Chr.11: 124630406-124630406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754264229 Chr.11: 124630749-124630749 on Build GRCh38
rs1025372065 Chr.11: 124630709-124630709 on Build GRCh38
rs747240766 Chr.11: 124630611-124630611 on Build GRCh38
rs764304985 Chr.11: 124630378-124630381 on Build GRCh38
rs773328654 Chr.11: 124630727-124630727 on Build GRCh38
rs768277281 Chr.11: 124630705-124630705 on Build GRCh38
rs919515667 Chr.11: 124630465-124630465 on Build GRCh38
rs79414234 Chr.11: 124630666-124630666 on Build GRCh38
rs749876670 Chr.11: 124630372-124630372 on Build GRCh38
rs903069630 Chr.11: 124630333-124630333 on Build GRCh38
rs757954245 Chr.11: 124630533-124630533 on Build GRCh38
rs147171432 Chr.11: 124630400-124630400 on Build GRCh38
rs1040791553 Chr.11: 124630293-124630293 on Build GRCh38
rs752531071 Chr.11: 124630298-124630298 on Build GRCh38
rs757786436 Chr.11: 124630500-124630500 on Build GRCh38
rs754246081 Chr.11: 124630487-124630487 on Build GRCh38
rs759671001 Chr.11: 124630734-124630734 on Build GRCh38
rs952256927 Chr.11: 124630497-124630497 on Build GRCh38
rs200809719 Chr.11: 124630718-124630718 on Build GRCh38
rs140282994 Chr.11: 124630425-124630425 on Build GRCh38
rs901827289 Chr.11: 124630297-124630297 on Build GRCh38
rs775164730 Chr.11: 124630420-124630420 on Build GRCh38
rs557306584 Chr.11: 124630286-124630286 on Build GRCh38
rs1034606073 Chr.11: 124630634-124630634 on Build GRCh38
rs373511727 Chr.11: 124630437-124630437 on Build GRCh38
rs372772691 Chr.11: 124630700-124630700 on Build GRCh38
rs943443284 Chr.11: 124630287-124630287 on Build GRCh38
rs562470816 Chr.11: 124630617-124630617 on Build GRCh38
rs758428976 Chr.11: 124630589-124630589 on Build GRCh38
rs764528371 Chr.11: 124630481-124630481 on Build GRCh38
rs768554708 Chr.11: 124630428-124630428 on Build GRCh38
rs774602125 Chr.11: 124630452-124630452 on Build GRCh38
rs375171804 Chr.11: 124630457-124630457 on Build GRCh38
rs755227505 Chr.11: 124630652-124630652 on Build GRCh38
rs750504503 Chr.11: 124630579-124630579 on Build GRCh38
rs377150923 Chr.11: 124630356-124630356 on Build GRCh38
rs753017374 Chr.11: 124630459-124630459 on Build GRCh38
rs763960664 Chr.11: 124630415-124630415 on Build GRCh38
rs760969833 Chr.11: 124630744-124630744 on Build GRCh38
rs765572443 Chr.11: 124630526-124630526 on Build GRCh38
rs775511656 Chr.11: 124630742-124630742 on Build GRCh38
rs762083108 Chr.11: 124630439-124630439 on Build GRCh38
rs200806695 Chr.11: 124630343-124630343 on Build GRCh38
rs200635425 Chr.11: 124630707-124630707 on Build GRCh38
rs530892513 Chr.11: 124630608-124630608 on Build GRCh38
rs765710763 Chr.11: 124630752-124630752 on Build GRCh38
rs759727193 Chr.11: 124630453-124630453 on Build GRCh38
rs992551209 Chr.11: 124630289-124630289 on Build GRCh38
rs202107879 Chr.11: 124630472-124630472 on Build GRCh38
rs756659677 Chr.11: 124630373-124630373 on Build GRCh38
rs142077518 Chr.11: 124630745-124630745 on Build GRCh38
rs771826956 Chr.11: 124630518-124630518 on Build GRCh38
rs760689920 Chr.11: 124630343-124630344 on Build GRCh38
rs772341635 Chr.11: 124630736-124630736 on Build GRCh38
rs1041650137 Chr.11: 124630317-124630317 on Build GRCh38
rs776906583 Chr.11: 124630385-124630385 on Build GRCh38
rs985381271 Chr.11: 124630510-124630510 on Build GRCh38
rs756547131 Chr.11: 124630363-124630363 on Build GRCh38
rs745352787 Chr.11: 124630521-124630521 on Build GRCh38
rs540031281 Chr.11: 124630517-124630517 on Build GRCh38
rs200993262 Chr.11: 124630535-124630535 on Build GRCh38
rs1000090925 Chr.11: 124630680-124630680 on Build GRCh38
rs766515101 Chr.11: 124630443-124630443 on Build GRCh38
rs1042374016 Chr.11: 124630660-124630660 on Build GRCh38
rs77814446 Chr.11: 124630360-124630360 on Build GRCh38
rs368533372 Chr.11: 124630735-124630735 on Build GRCh38
rs576850218 Chr.11: 124630553-124630553 on Build GRCh38
rs910692039 Chr.11: 124630532-124630532 on Build GRCh38
rs745477847 Chr.11: 124630414-124630414 on Build GRCh38
rs376298258 Chr.11: 124630706-124630706 on Build GRCh38
rs762205839 Chr.11: 124630750-124630750 on Build GRCh38
rs778125758 Chr.11: 124630365-124630365 on Build GRCh38
rs775569583 Chr.11: 124630341-124630344 on Build GRCh38
rs767823918 Chr.11: 124630455-124630455 on Build GRCh38
rs776365211 Chr.11: 124630432-124630432 on Build GRCh38

View Full Product Details