Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125596951-125597470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STT3A
Entrez Gene ID
3703
Gene Name
STT3A, catalytic subunit of the oligosaccharyltransferase complex
Gene Aliases
ITM1, STT3-A, TMC
UniGene
Hs.504237
Gene Chromosome Location
Chr.11: 125592795-125622759 on Build GRCh38
Transcripts
NM_001278503.1 NM_001278504.1 NM_152713.4 XM_011542807.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3444970 c.95C>A p.S32Y Chr.11: 125597065-125597065 on Build GRCh38
6792524 c.100C>T p.R34C Chr.11: 125597070-125597070 on Build GRCh38
6792529 c.101G>A p.R34H Chr.11: 125597071-125597071 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764165575 Chr.11: 125597138-125597138 on Build GRCh38
rs513209 Chr.11: 125597249-125597249 on Build GRCh38
rs372386670 Chr.11: 125597291-125597291 on Build GRCh38
rs554101190 Chr.11: 125597277-125597277 on Build GRCh38
rs764109996 Chr.11: 125597020-125597020 on Build GRCh38
rs975429535 Chr.11: 125597413-125597413 on Build GRCh38
rs902510132 Chr.11: 125597225-125597225 on Build GRCh38
rs568911834 Chr.11: 125597024-125597024 on Build GRCh38
rs76225995 Chr.11: 125597281-125597281 on Build GRCh38
rs774311073 Chr.11: 125597012-125597012 on Build GRCh38
rs754547148 Chr.11: 125597060-125597060 on Build GRCh38
rs909671887 Chr.11: 125597065-125597065 on Build GRCh38
rs929663643 Chr.11: 125597005-125597005 on Build GRCh38
rs370077215 Chr.11: 125597141-125597141 on Build GRCh38
rs572984370 Chr.11: 125597421-125597422 on Build GRCh38
rs540023383 Chr.11: 125597302-125597302 on Build GRCh38
rs760772350 Chr.11: 125597019-125597019 on Build GRCh38
rs201369680 Chr.11: 125597239-125597240 on Build GRCh38
rs1052025726 Chr.11: 125597213-125597213 on Build GRCh38
rs531278643 Chr.11: 125597053-125597053 on Build GRCh38
rs369018247 Chr.11: 125597016-125597016 on Build GRCh38
rs201391299 Chr.11: 125597075-125597075 on Build GRCh38
rs1025549715 Chr.11: 125597399-125597399 on Build GRCh38
rs369590985 Chr.11: 125597021-125597021 on Build GRCh38
rs1035191088 Chr.11: 125597203-125597203 on Build GRCh38
rs769623450 Chr.11: 125597071-125597071 on Build GRCh38
rs951116845 Chr.11: 125597442-125597442 on Build GRCh38
rs758759795 Chr.11: 125597042-125597042 on Build GRCh38
rs568115954 Chr.11: 125597240-125597240 on Build GRCh38
rs747652562 Chr.11: 125597070-125597070 on Build GRCh38
rs970535650 Chr.11: 125597339-125597339 on Build GRCh38
rs768371983 Chr.11: 125597299-125597299 on Build GRCh38
rs750801867 Chr.11: 125597031-125597031 on Build GRCh38
rs576493536 Chr.11: 125597324-125597324 on Build GRCh38
rs556258306 Chr.11: 125597307-125597307 on Build GRCh38
rs369191984 Chr.11: 125597421-125597421 on Build GRCh38
rs138235571 Chr.11: 125597063-125597063 on Build GRCh38
rs747499490 Chr.11: 125597055-125597055 on Build GRCh38
rs999574798 Chr.11: 125597235-125597235 on Build GRCh38
rs202151592 Chr.11: 125597135-125597135 on Build GRCh38
rs984389471 Chr.11: 125597292-125597292 on Build GRCh38
rs371968584 Chr.11: 125596994-125596994 on Build GRCh38
rs58836204 Chr.11: 125597422-125597423 on Build GRCh38
rs896230189 Chr.11: 125597240-125597240 on Build GRCh38
rs556433529 Chr.11: 125597287-125597287 on Build GRCh38
rs981958370 Chr.11: 125597353-125597353 on Build GRCh38
rs113884453 Chr.11: 125597251-125597251 on Build GRCh38
rs397813716 Chr.11: 125597437-125597438 on Build GRCh38
rs781106175 Chr.11: 125597316-125597316 on Build GRCh38
rs376957358 Chr.11: 125597140-125597140 on Build GRCh38
rs531971276 Chr.11: 125596991-125596991 on Build GRCh38
rs774181158 Chr.11: 125597136-125597136 on Build GRCh38
rs553075175 Chr.11: 125597249-125597249 on Build GRCh38
rs373498147 Chr.11: 125597183-125597183 on Build GRCh38
rs374068836 Chr.11: 125597030-125597030 on Build GRCh38
rs144743356 Chr.11: 125597139-125597139 on Build GRCh38

View Full Product Details