Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126429087-126429486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIRREL3
Entrez Gene ID
84623
Gene Name
kin of IRRE like 3 (Drosophila)
Gene Aliases
KIRRE, MRD4, NEPH2, PRO4502
UniGene
Hs.376015
Gene Chromosome Location
Chr.11: 126423493-127003460 on Build GRCh38
Transcripts
NM_001301097.1 NM_032531.3 XM_011543026.2 XM_011543027.2 XM_011543028.2 XM_011543030.2 XM_011543031.2 XM_011543032.2 XM_011543033.1 XM_017018418.1 XM_017018419.1 XM_017018420.1 XM_011542959.1 XM_011542960.2

Gene Symbol
ST3GAL4
Entrez Gene ID
6484
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Gene Aliases
CGS23, NANTA3, SAT3, SIAT4, SIAT4C, ST3GalIV, STZ
UniGene
Hs.591947
Gene Chromosome Location
Chr.11: 126355645-126459601 on Build GRCh38
Transcripts
NM_001301097.1 NM_032531.3 XM_011543026.2 XM_011543027.2 XM_011543028.2 XM_011543030.2 XM_011543031.2 XM_011543032.2 XM_011543033.1 XM_017018418.1 XM_017018419.1 XM_017018420.1 XM_011542959.1 XM_011542960.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838135 c.1706G>A p.G569D Chr.11: 126429279-126429279 on Build GRCh38
6295511 c.1718C>A p.A573D Chr.11: 126429267-126429267 on Build GRCh38
1212546 c.1574-2A>G p.? Chr.11: 126429290-126429290 on Build GRCh38
6696043 c.1732C>T p.R578* Chr.11: 126429253-126429253 on Build GRCh38
1146620 c.1768C>A p.R590R Chr.11: 126429217-126429217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895982611 Chr.11: 126429323-126429323 on Build GRCh38
rs533224256 Chr.11: 126429402-126429402 on Build GRCh38
rs750546184 Chr.11: 126429230-126429230 on Build GRCh38
rs531322165 Chr.11: 126429225-126429225 on Build GRCh38
rs527280644 Chr.11: 126429352-126429352 on Build GRCh38
rs777129407 Chr.11: 126429147-126429147 on Build GRCh38
rs987854177 Chr.11: 126429132-126429132 on Build GRCh38
rs372461074 Chr.11: 126429248-126429248 on Build GRCh38
rs375451547 Chr.11: 126429318-126429318 on Build GRCh38
rs770259323 Chr.11: 126429301-126429301 on Build GRCh38
rs367848509 Chr.11: 126429216-126429216 on Build GRCh38
rs547320046 Chr.11: 126429384-126429384 on Build GRCh38
rs748802604 Chr.11: 126429191-126429191 on Build GRCh38
rs973459158 Chr.11: 126429426-126429427 on Build GRCh38
rs755403705 Chr.11: 126429252-126429252 on Build GRCh38
rs919260746 Chr.11: 126429266-126429266 on Build GRCh38
rs759377225 Chr.11: 126429206-126429206 on Build GRCh38
rs372381320 Chr.11: 126429331-126429331 on Build GRCh38
rs150056103 Chr.11: 126429167-126429167 on Build GRCh38
rs773813451 Chr.11: 126429199-126429199 on Build GRCh38
rs796077047 Chr.11: 126429220-126429220 on Build GRCh38
rs773955915 Chr.11: 126429325-126429325 on Build GRCh38
rs758361889 Chr.11: 126429232-126429232 on Build GRCh38
rs780337340 Chr.11: 126429243-126429243 on Build GRCh38
rs1000516407 Chr.11: 126429115-126429115 on Build GRCh38
rs982216527 Chr.11: 126429443-126429443 on Build GRCh38
rs1031930412 Chr.11: 126429395-126429395 on Build GRCh38
rs892142201 Chr.11: 126429425-126429425 on Build GRCh38
rs748879865 Chr.11: 126429279-126429279 on Build GRCh38
rs1030537588 Chr.11: 126429445-126429445 on Build GRCh38
rs1000423891 Chr.11: 126429343-126429343 on Build GRCh38
rs769355375 Chr.11: 126429163-126429163 on Build GRCh38
rs956434962 Chr.11: 126429128-126429128 on Build GRCh38
rs765447391 Chr.11: 126429229-126429229 on Build GRCh38
rs578151570 Chr.11: 126429148-126429148 on Build GRCh38
rs772041154 Chr.11: 126429333-126429333 on Build GRCh38
rs375566225 Chr.11: 126429310-126429310 on Build GRCh38
rs74464311 Chr.11: 126429194-126429194 on Build GRCh38
rs954553054 Chr.11: 126429452-126429452 on Build GRCh38
rs923097481 Chr.11: 126429139-126429139 on Build GRCh38
rs368620051 Chr.11: 126429330-126429330 on Build GRCh38
rs541484222 Chr.11: 126429277-126429277 on Build GRCh38
rs954620654 Chr.11: 126429308-126429308 on Build GRCh38
rs954739230 Chr.11: 126429152-126429152 on Build GRCh38
rs772074108 Chr.11: 126429432-126429432 on Build GRCh38
rs778217107 Chr.11: 126429314-126429314 on Build GRCh38
rs748705912 Chr.11: 126429295-126429295 on Build GRCh38
rs950893861 Chr.11: 126429438-126429438 on Build GRCh38
rs564201836 Chr.11: 126429393-126429393 on Build GRCh38
rs1031982290 Chr.11: 126429124-126429124 on Build GRCh38
rs752738293 Chr.11: 126429219-126429219 on Build GRCh38

View Full Product Details