Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4590245-4590745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK4
Entrez Gene ID
8798
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 4
Gene Aliases
-
UniGene
Hs.439530
Gene Chromosome Location
Chr.12: 4590072-4613888 on Build GRCh38
Transcripts
NM_003845.2 NR_104115.1

SNP Detail

SNP ID Chromosome Location
rs147564041 Chr.12: 4590439-4590439 on Build GRCh38
rs930145386 Chr.12: 4590681-4590681 on Build GRCh38
rs906895448 Chr.12: 4590303-4590303 on Build GRCh38
rs765906089 Chr.12: 4590301-4590301 on Build GRCh38
rs994319899 Chr.12: 4590297-4590297 on Build GRCh38
rs898704595 Chr.12: 4590270-4590270 on Build GRCh38
rs751261732 Chr.12: 4590363-4590363 on Build GRCh38
rs752553422 Chr.12: 4590396-4590396 on Build GRCh38
rs919976028 Chr.12: 4590669-4590669 on Build GRCh38
rs938308857 Chr.12: 4590338-4590338 on Build GRCh38
rs541969056 Chr.12: 4590285-4590285 on Build GRCh38
rs567270762 Chr.12: 4590300-4590300 on Build GRCh38
rs569648413 Chr.12: 4590557-4590557 on Build GRCh38
rs975446063 Chr.12: 4590658-4590658 on Build GRCh38
rs891391046 Chr.12: 4590704-4590704 on Build GRCh38
rs2302250 Chr.12: 4590529-4590529 on Build GRCh38
rs1047357354 Chr.12: 4590321-4590321 on Build GRCh38
rs565750959 Chr.12: 4590659-4590659 on Build GRCh38
rs1017903682 Chr.12: 4590382-4590382 on Build GRCh38
rs780908544 Chr.12: 4590687-4590687 on Build GRCh38
rs948047513 Chr.12: 4590574-4590574 on Build GRCh38
rs532152477 Chr.12: 4590558-4590558 on Build GRCh38
rs928576194 Chr.12: 4590484-4590484 on Build GRCh38
rs1020078960 Chr.12: 4590542-4590542 on Build GRCh38
rs990458347 Chr.12: 4590706-4590706 on Build GRCh38
rs887246944 Chr.12: 4590453-4590453 on Build GRCh38
rs141935111 Chr.12: 4590452-4590452 on Build GRCh38
rs1047223088 Chr.12: 4590701-4590701 on Build GRCh38
rs992128161 Chr.12: 4590554-4590554 on Build GRCh38
rs953297779 Chr.12: 4590693-4590693 on Build GRCh38
rs775050225 Chr.12: 4590286-4590286 on Build GRCh38
rs894763533 Chr.12: 4590350-4590350 on Build GRCh38
rs112542588 Chr.12: 4590352-4590352 on Build GRCh38
rs371358359 Chr.12: 4590535-4590535 on Build GRCh38
rs1055976823 Chr.12: 4590343-4590343 on Build GRCh38
rs12296105 Chr.12: 4590617-4590617 on Build GRCh38
rs71579236 Chr.12: 4590398-4590398 on Build GRCh38
rs185875180 Chr.12: 4590280-4590280 on Build GRCh38
rs756808287 Chr.12: 4590676-4590676 on Build GRCh38

View Full Product Details